Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171553217-171553756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBRD1
Entrez Gene ID
79901
Gene Name
cytochrome b reductase 1
Gene Aliases
CYB561A2, DCYTB, FRRS3
UniGene
Hs.221941
Gene Chromosome Location
Chr.2: 171522247-171558133 on Build GRCh38
Transcripts
NM_001127383.1 NM_001256909.1 NM_024843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603333 c.434C>T p.P145L Chr.2: 171553377-171553377 on Build GRCh38
6534678 c.516A>G p.A172A Chr.2: 171553459-171553459 on Build GRCh38
4403519 c.532T>G p.L178V Chr.2: 171553475-171553475 on Build GRCh38
1009544 c.474A>G p.I158M Chr.2: 171553417-171553417 on Build GRCh38
5779933 c.534G>C p.L178F Chr.2: 171553477-171553477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558737857 Chr.2: 171553627-171553627 on Build GRCh38
rs765719282 Chr.2: 171553542-171553542 on Build GRCh38
rs968534552 Chr.2: 171553439-171553439 on Build GRCh38
rs759719738 Chr.2: 171553365-171553365 on Build GRCh38
rs762342107 Chr.2: 171553536-171553536 on Build GRCh38
rs1015317837 Chr.2: 171553635-171553639 on Build GRCh38
rs17288588 Chr.2: 171553676-171553676 on Build GRCh38
rs761578508 Chr.2: 171553356-171553356 on Build GRCh38
rs929361575 Chr.2: 171553571-171553571 on Build GRCh38
rs200712533 Chr.2: 171553471-171553471 on Build GRCh38
rs111227005 Chr.2: 171553576-171553576 on Build GRCh38
rs779722907 Chr.2: 171553411-171553411 on Build GRCh38
rs577309798 Chr.2: 171553643-171553643 on Build GRCh38
rs777046054 Chr.2: 171553528-171553528 on Build GRCh38
rs781047968 Chr.2: 171553421-171553421 on Build GRCh38
rs765666345 Chr.2: 171553392-171553392 on Build GRCh38
rs753292572 Chr.2: 171553514-171553516 on Build GRCh38
rs776489350 Chr.2: 171553331-171553331 on Build GRCh38
rs868251308 Chr.2: 171553252-171553252 on Build GRCh38
rs1034175461 Chr.2: 171553257-171553257 on Build GRCh38
rs202158377 Chr.2: 171553387-171553387 on Build GRCh38
rs1049084215 Chr.2: 171553583-171553583 on Build GRCh38
rs763795766 Chr.2: 171553365-171553365 on Build GRCh38
rs774602397 Chr.2: 171553363-171553363 on Build GRCh38
rs768214360 Chr.2: 171553483-171553483 on Build GRCh38
rs767386737 Chr.2: 171553306-171553311 on Build GRCh38
rs1041285062 Chr.2: 171553516-171553516 on Build GRCh38
rs16859487 Chr.2: 171553410-171553410 on Build GRCh38
rs1055637461 Chr.2: 171553642-171553642 on Build GRCh38
rs754505258 Chr.2: 171553415-171553415 on Build GRCh38
rs536929544 Chr.2: 171553323-171553323 on Build GRCh38
rs887762978 Chr.2: 171553592-171553592 on Build GRCh38
rs769615671 Chr.2: 171553360-171553360 on Build GRCh38
rs1024063209 Chr.2: 171553707-171553707 on Build GRCh38
rs760421926 Chr.2: 171553332-171553332 on Build GRCh38
rs970551205 Chr.2: 171553637-171553637 on Build GRCh38
rs986366837 Chr.2: 171553457-171553457 on Build GRCh38
rs775328160 Chr.2: 171553307-171553311 on Build GRCh38
rs1004078667 Chr.2: 171553269-171553269 on Build GRCh38
rs145340565 Chr.2: 171553470-171553470 on Build GRCh38
rs760727590 Chr.2: 171553515-171553515 on Build GRCh38
rs1003977429 Chr.2: 171553612-171553612 on Build GRCh38
rs200585127 Chr.2: 171553412-171553412 on Build GRCh38
rs958525533 Chr.2: 171553264-171553264 on Build GRCh38
rs569201460 Chr.2: 171553297-171553297 on Build GRCh38
rs1048523371 Chr.2: 171553592-171553592 on Build GRCh38
rs538394234 Chr.2: 171553660-171553660 on Build GRCh38
rs958771066 Chr.2: 171553326-171553326 on Build GRCh38
rs769614064 Chr.2: 171553435-171553435 on Build GRCh38
rs144115153 Chr.2: 171553397-171553397 on Build GRCh38
rs980165654 Chr.2: 171553606-171553606 on Build GRCh38
rs779671593 Chr.2: 171553318-171553318 on Build GRCh38
rs1014050050 Chr.2: 171553703-171553703 on Build GRCh38
rs775494355 Chr.2: 171553378-171553378 on Build GRCh38
rs959536403 Chr.2: 171553729-171553729 on Build GRCh38
rs1040532222 Chr.2: 171553499-171553499 on Build GRCh38
rs900082884 Chr.2: 171553519-171553519 on Build GRCh38
rs761252992 Chr.2: 171553379-171553379 on Build GRCh38
rs919356052 Chr.2: 171553564-171553564 on Build GRCh38
rs746106742 Chr.2: 171553461-171553461 on Build GRCh38
rs1024010524 Chr.2: 171553336-171553336 on Build GRCh38
rs925583496 Chr.2: 171553399-171553399 on Build GRCh38
rs941322262 Chr.2: 171553603-171553603 on Build GRCh38
rs143524630 Chr.2: 171553281-171553281 on Build GRCh38
rs764578336 Chr.2: 171553386-171553386 on Build GRCh38
rs746965513 Chr.2: 171553322-171553322 on Build GRCh38
rs764213877 Chr.2: 171553525-171553525 on Build GRCh38
rs767611054 Chr.2: 171553367-171553367 on Build GRCh38
rs772947456 Chr.2: 171553453-171553453 on Build GRCh38

View Full Product Details