Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171554798-171555294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBRD1
Entrez Gene ID
79901
Gene Name
cytochrome b reductase 1
Gene Aliases
CYB561A2, DCYTB, FRRS3
UniGene
Hs.221941
Gene Chromosome Location
Chr.2: 171522247-171558133 on Build GRCh38
Transcripts
NM_001127383.1 NM_001256909.1 NM_024843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4913310 c.833A>G p.D278G Chr.2: 171554799-171554799 on Build GRCh38
476220 c.858G>A p.M286I Chr.2: 171554824-171554824 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901751082 Chr.2: 171555108-171555108 on Build GRCh38
rs765285755 Chr.2: 171554854-171554854 on Build GRCh38
rs16859496 Chr.2: 171555176-171555176 on Build GRCh38
rs1017131802 Chr.2: 171554884-171554884 on Build GRCh38
rs776062353 Chr.2: 171554849-171554849 on Build GRCh38
rs138508811 Chr.2: 171555147-171555147 on Build GRCh38
rs550355580 Chr.2: 171555110-171555110 on Build GRCh38
rs6746474 Chr.2: 171555045-171555045 on Build GRCh38
rs1016995537 Chr.2: 171555154-171555154 on Build GRCh38
rs761745028 Chr.2: 171554850-171554850 on Build GRCh38
rs766055590 Chr.2: 171554872-171554872 on Build GRCh38
rs746305730 Chr.2: 171554823-171554823 on Build GRCh38
rs774579184 Chr.2: 171554821-171554821 on Build GRCh38
rs1025076404 Chr.2: 171555126-171555126 on Build GRCh38
rs772306050 Chr.2: 171554831-171554831 on Build GRCh38
rs775166396 Chr.2: 171554832-171554832 on Build GRCh38
rs758260372 Chr.2: 171554860-171554860 on Build GRCh38
rs923027814 Chr.2: 171555042-171555042 on Build GRCh38
rs552488923 Chr.2: 171554992-171554992 on Build GRCh38
rs978516990 Chr.2: 171554859-171554859 on Build GRCh38
rs895362013 Chr.2: 171555123-171555123 on Build GRCh38
rs1012376777 Chr.2: 171555124-171555124 on Build GRCh38
rs1037914169 Chr.2: 171555093-171555093 on Build GRCh38
rs960693739 Chr.2: 171554953-171554953 on Build GRCh38
rs1021562145 Chr.2: 171555235-171555235 on Build GRCh38
rs781448647 Chr.2: 171555245-171555245 on Build GRCh38
rs1035308162 Chr.2: 171555255-171555255 on Build GRCh38
rs750935470 Chr.2: 171554822-171554822 on Build GRCh38
rs886090714 Chr.2: 171555088-171555088 on Build GRCh38
rs1012324620 Chr.2: 171554829-171554829 on Build GRCh38
rs1047872335 Chr.2: 171555046-171555046 on Build GRCh38
rs923947633 Chr.2: 171554885-171554885 on Build GRCh38
rs946469031 Chr.2: 171555003-171555003 on Build GRCh38
rs1040306313 Chr.2: 171555101-171555101 on Build GRCh38
rs1060248 Chr.2: 171555134-171555134 on Build GRCh38
rs376023294 Chr.2: 171554844-171554844 on Build GRCh38
rs554263285 Chr.2: 171555152-171555152 on Build GRCh38
rs570827754 Chr.2: 171555012-171555012 on Build GRCh38
rs1028859645 Chr.2: 171554843-171554843 on Build GRCh38
rs560581890 Chr.2: 171554855-171554855 on Build GRCh38
rs942310011 Chr.2: 171555092-171555092 on Build GRCh38
rs561603161 Chr.2: 171555034-171555034 on Build GRCh38
rs943188628 Chr.2: 171555032-171555032 on Build GRCh38
rs758813938 Chr.2: 171554847-171554848 on Build GRCh38
rs6759970 Chr.2: 171554986-171554986 on Build GRCh38
rs956179972 Chr.2: 171554942-171554942 on Build GRCh38
rs961507255 Chr.2: 171554909-171554909 on Build GRCh38
rs928164231 Chr.2: 171554950-171554950 on Build GRCh38
rs751712917 Chr.2: 171555204-171555204 on Build GRCh38
rs960977394 Chr.2: 171555259-171555259 on Build GRCh38
rs950163 Chr.2: 171555227-171555227 on Build GRCh38
rs534063455 Chr.2: 171555194-171555194 on Build GRCh38
rs935126807 Chr.2: 171555097-171555097 on Build GRCh38
rs763619350 Chr.2: 171554845-171554845 on Build GRCh38
rs990494665 Chr.2: 171554957-171554957 on Build GRCh38
rs974542146 Chr.2: 171555038-171555038 on Build GRCh38
rs752995999 Chr.2: 171554879-171554879 on Build GRCh38

View Full Product Details