Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171965709-171966207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAT1
Entrez Gene ID
8520
Gene Name
histone acetyltransferase 1
Gene Aliases
KAT1
UniGene
Hs.632532
Gene Chromosome Location
Chr.2: 171922425-171983686 on Build GRCh38
Transcripts
NM_003642.3 NR_027862.1 XM_006712808.3 XM_011512027.2 XM_017005136.1 XM_017005137.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1325761 c.605T>G p.F202C Chr.2: 171965902-171965902 on Build GRCh38
2893384 c.497T>G p.M166R Chr.2: 171965794-171965794 on Build GRCh38
7262576 c.536A>G p.Q179R Chr.2: 171965833-171965833 on Build GRCh38
4689521 c.519A>G p.E173E Chr.2: 171965816-171965816 on Build GRCh38
1009562 c.550T>C p.W184R Chr.2: 171965847-171965847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747359397 Chr.2: 171965821-171965821 on Build GRCh38
rs751341862 Chr.2: 171965771-171965771 on Build GRCh38
rs376429104 Chr.2: 171965929-171965929 on Build GRCh38
rs1039177394 Chr.2: 171966046-171966046 on Build GRCh38
rs567232699 Chr.2: 171965876-171965876 on Build GRCh38
rs2304999 Chr.2: 171966081-171966081 on Build GRCh38
rs1043046583 Chr.2: 171965958-171965958 on Build GRCh38
rs765791740 Chr.2: 171965980-171965980 on Build GRCh38
rs138153000 Chr.2: 171966171-171966171 on Build GRCh38
rs772830902 Chr.2: 171965752-171965752 on Build GRCh38
rs780572505 Chr.2: 171965817-171965817 on Build GRCh38
rs141652201 Chr.2: 171966044-171966044 on Build GRCh38
rs762843864 Chr.2: 171965754-171965754 on Build GRCh38
rs778288022 Chr.2: 171965931-171965932 on Build GRCh38
rs147866736 Chr.2: 171965844-171965844 on Build GRCh38
rs534919587 Chr.2: 171965933-171965933 on Build GRCh38
rs747614202 Chr.2: 171965948-171965948 on Build GRCh38
rs146360586 Chr.2: 171965908-171965908 on Build GRCh38
rs984791274 Chr.2: 171966119-171966119 on Build GRCh38
rs764315094 Chr.2: 171965793-171965793 on Build GRCh38
rs780301274 Chr.2: 171966098-171966098 on Build GRCh38
rs369912305 Chr.2: 171965791-171965791 on Build GRCh38
rs937025260 Chr.2: 171966139-171966139 on Build GRCh38
rs762485046 Chr.2: 171965879-171965879 on Build GRCh38
rs371877066 Chr.2: 171965799-171965799 on Build GRCh38
rs766247676 Chr.2: 171965904-171965904 on Build GRCh38
rs758565495 Chr.2: 171965864-171965864 on Build GRCh38
rs994985499 Chr.2: 171966104-171966104 on Build GRCh38
rs376194532 Chr.2: 171966140-171966140 on Build GRCh38
rs755422648 Chr.2: 171965951-171965951 on Build GRCh38
rs571385824 Chr.2: 171966120-171966120 on Build GRCh38
rs918756166 Chr.2: 171966000-171966000 on Build GRCh38
rs972757116 Chr.2: 171965972-171965972 on Build GRCh38
rs372856571 Chr.2: 171965922-171965922 on Build GRCh38
rs758458717 Chr.2: 171965946-171965946 on Build GRCh38
rs530593330 Chr.2: 171965812-171965812 on Build GRCh38
rs569714151 Chr.2: 171965801-171965801 on Build GRCh38
rs367760195 Chr.2: 171965763-171965763 on Build GRCh38
rs747856422 Chr.2: 171965849-171965849 on Build GRCh38
rs1001408523 Chr.2: 171966183-171966183 on Build GRCh38
rs769545273 Chr.2: 171965865-171965865 on Build GRCh38
rs1048746028 Chr.2: 171966105-171966105 on Build GRCh38
rs749668458 Chr.2: 171965958-171965959 on Build GRCh38
rs745928896 Chr.2: 171965811-171965811 on Build GRCh38
rs371484531 Chr.2: 171965747-171965747 on Build GRCh38
rs756586898 Chr.2: 171965775-171965776 on Build GRCh38
rs750504274 Chr.2: 171965943-171965943 on Build GRCh38
rs757466935 Chr.2: 171965800-171965800 on Build GRCh38
rs895547832 Chr.2: 171965760-171965760 on Build GRCh38
rs34606494 Chr.2: 171965986-171965987 on Build GRCh38
rs764904338 Chr.2: 171965751-171965751 on Build GRCh38
rs1056796148 Chr.2: 171966168-171966168 on Build GRCh38
rs757524962 Chr.2: 171965938-171965938 on Build GRCh38
rs780442943 Chr.2: 171965947-171965947 on Build GRCh38
rs900292802 Chr.2: 171966065-171966065 on Build GRCh38
rs147062013 Chr.2: 171966124-171966124 on Build GRCh38
rs917038662 Chr.2: 171966169-171966169 on Build GRCh38
rs765417806 Chr.2: 171965942-171965942 on Build GRCh38
rs1006879819 Chr.2: 171965877-171965877 on Build GRCh38
rs768996123 Chr.2: 171965825-171965825 on Build GRCh38

View Full Product Details