Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176123810-176124348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD9
Entrez Gene ID
3235
Gene Name
homeobox D9
Gene Aliases
HOX4, HOX4C, Hox-4.3, Hox-5.2
UniGene
Hs.236646
Gene Chromosome Location
Chr.2: 176122685-176124917 on Build GRCh38
Transcripts
NM_014213.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1613728 c.930G>T p.R310S Chr.2: 176124076-176124076 on Build GRCh38
4402292 c.901A>G p.T301A Chr.2: 176124047-176124047 on Build GRCh38
4519969 c.1006G>A p.E336K Chr.2: 176124152-176124152 on Build GRCh38
1009882 c.840C>T p.I280I Chr.2: 176123956-176123956 on Build GRCh38
7106377 c.841C>T p.H281Y Chr.2: 176123957-176123957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375399643 Chr.2: 176124179-176124179 on Build GRCh38
rs769740616 Chr.2: 176123980-176123980 on Build GRCh38
rs1028440759 Chr.2: 176124264-176124264 on Build GRCh38
rs773113298 Chr.2: 176123981-176123981 on Build GRCh38
rs555645033 Chr.2: 176123853-176123853 on Build GRCh38
rs754192235 Chr.2: 176123897-176123897 on Build GRCh38
rs754535646 Chr.2: 176124040-176124040 on Build GRCh38
rs748231542 Chr.2: 176124189-176124189 on Build GRCh38
rs563878728 Chr.2: 176124067-176124067 on Build GRCh38
rs775503474 Chr.2: 176123991-176123991 on Build GRCh38
rs757633288 Chr.2: 176123947-176123947 on Build GRCh38
rs771718430 Chr.2: 176124216-176124218 on Build GRCh38
rs997814041 Chr.2: 176124268-176124268 on Build GRCh38
rs192165308 Chr.2: 176124298-176124298 on Build GRCh38
rs769651961 Chr.2: 176123925-176123925 on Build GRCh38
rs1010011987 Chr.2: 176123890-176123890 on Build GRCh38
rs767278150 Chr.2: 176124221-176124221 on Build GRCh38
rs1007737949 Chr.2: 176123937-176123937 on Build GRCh38
rs536881181 Chr.2: 176124291-176124292 on Build GRCh38
rs561988112 Chr.2: 176124212-176124212 on Build GRCh38
rs759076963 Chr.2: 176124151-176124151 on Build GRCh38
rs777145928 Chr.2: 176124013-176124013 on Build GRCh38
rs1026163691 Chr.2: 176124281-176124281 on Build GRCh38
rs568475051 Chr.2: 176123918-176123918 on Build GRCh38
rs767188288 Chr.2: 176124154-176124154 on Build GRCh38
rs966191055 Chr.2: 176124307-176124307 on Build GRCh38
rs753883753 Chr.2: 176124162-176124162 on Build GRCh38
rs762579550 Chr.2: 176124021-176124021 on Build GRCh38
rs926876402 Chr.2: 176124074-176124074 on Build GRCh38
rs762379004 Chr.2: 176124295-176124296 on Build GRCh38
rs711822 Chr.2: 176124295-176124295 on Build GRCh38
rs371557389 Chr.2: 176123999-176123999 on Build GRCh38
rs780337434 Chr.2: 176124082-176124082 on Build GRCh38
rs997340758 Chr.2: 176124032-176124032 on Build GRCh38
rs35299188 Chr.2: 176124103-176124103 on Build GRCh38
rs770671156 Chr.2: 176123884-176123885 on Build GRCh38
rs780806613 Chr.2: 176124052-176124052 on Build GRCh38
rs768756561 Chr.2: 176124088-176124088 on Build GRCh38
rs892535493 Chr.2: 176123880-176123880 on Build GRCh38
rs745465945 Chr.2: 176123913-176123913 on Build GRCh38
rs112934078 Chr.2: 176124076-176124076 on Build GRCh38
rs146173388 Chr.2: 176124084-176124084 on Build GRCh38
rs1044162736 Chr.2: 176124065-176124065 on Build GRCh38
rs202030114 Chr.2: 176124031-176124031 on Build GRCh38
rs765538916 Chr.2: 176123953-176123953 on Build GRCh38
rs138258352 Chr.2: 176124068-176124068 on Build GRCh38
rs1021389706 Chr.2: 176123836-176123836 on Build GRCh38
rs764023606 Chr.2: 176124008-176124008 on Build GRCh38
rs747958740 Chr.2: 176123924-176123924 on Build GRCh38
rs373038108 Chr.2: 176124292-176124293 on Build GRCh38
rs756970863 Chr.2: 176123907-176123907 on Build GRCh38
rs748046716 Chr.2: 176123973-176123973 on Build GRCh38
rs776353054 Chr.2: 176123923-176123923 on Build GRCh38
rs756101299 Chr.2: 176124058-176124058 on Build GRCh38
rs760814488 Chr.2: 176123994-176123994 on Build GRCh38
rs762269182 Chr.2: 176124016-176124016 on Build GRCh38
rs1026511263 Chr.2: 176124034-176124034 on Build GRCh38
rs764418768 Chr.2: 176123887-176123887 on Build GRCh38
rs775221055 Chr.2: 176124222-176124222 on Build GRCh38
rs746524430 Chr.2: 176123964-176123964 on Build GRCh38
rs914028361 Chr.2: 176124085-176124085 on Build GRCh38
rs567457035 Chr.2: 176123963-176123963 on Build GRCh38
rs73033729 Chr.2: 176123847-176123847 on Build GRCh38
rs773903248 Chr.2: 176123959-176123962 on Build GRCh38
rs1005933045 Chr.2: 176124294-176124295 on Build GRCh38
rs968010176 Chr.2: 176123837-176123837 on Build GRCh38
rs752724102 Chr.2: 176124053-176124053 on Build GRCh38
rs1000861220 Chr.2: 176124244-176124244 on Build GRCh38
rs541136840 Chr.2: 176124198-176124198 on Build GRCh38
rs751253380 Chr.2: 176124150-176124150 on Build GRCh38
rs372008744 Chr.2: 176124169-176124169 on Build GRCh38
rs752245598 Chr.2: 176124161-176124161 on Build GRCh38
rs201376389 Chr.2: 176124295-176124295 on Build GRCh38
rs775124999 Chr.2: 176124224-176124224 on Build GRCh38
rs776668727 Chr.2: 176124095-176124095 on Build GRCh38
rs866068404 Chr.2: 176124002-176124002 on Build GRCh38
rs759794740 Chr.2: 176123935-176123935 on Build GRCh38
rs367897393 Chr.2: 176124286-176124288 on Build GRCh38
rs754277883 Chr.2: 176123945-176123945 on Build GRCh38
rs35649704 Chr.2: 176124037-176124037 on Build GRCh38
rs921348889 Chr.2: 176123863-176123863 on Build GRCh38
rs767536951 Chr.2: 176123938-176123938 on Build GRCh38
rs761147520 Chr.2: 176123940-176123940 on Build GRCh38
rs542311904 Chr.2: 176124199-176124199 on Build GRCh38
rs766599876 Chr.2: 176124136-176124136 on Build GRCh38
rs768850703 Chr.2: 176124183-176124183 on Build GRCh38
rs368352732 Chr.2: 176124110-176124110 on Build GRCh38
rs747297770 Chr.2: 176124181-176124181 on Build GRCh38
rs781285150 Chr.2: 176123967-176123967 on Build GRCh38
rs753660337 Chr.2: 176124178-176124178 on Build GRCh38
rs775621367 Chr.2: 176123939-176123939 on Build GRCh38
rs376874077 Chr.2: 176123982-176123982 on Build GRCh38
rs772328942 Chr.2: 176123985-176123985 on Build GRCh38
rs759323422 Chr.2: 176124218-176124218 on Build GRCh38
rs572161051 Chr.2: 176124193-176124193 on Build GRCh38
rs778542236 Chr.2: 176123910-176123910 on Build GRCh38
rs62188694 Chr.2: 176124292-176124292 on Build GRCh38
rs878900300 Chr.2: 176124296-176124296 on Build GRCh38
rs774734700 Chr.2: 176123930-176123930 on Build GRCh38
rs764657611 Chr.2: 176123944-176123944 on Build GRCh38
rs757052465 Chr.2: 176124192-176124192 on Build GRCh38
rs1043760900 Chr.2: 176123908-176123908 on Build GRCh38
rs757871081 Chr.2: 176123914-176123914 on Build GRCh38
rs774560342 Chr.2: 176124216-176124216 on Build GRCh38
rs750772104 Chr.2: 176124018-176124018 on Build GRCh38
rs750114730 Chr.2: 176123959-176123959 on Build GRCh38
rs760622122 Chr.2: 176124099-176124099 on Build GRCh38
rs754375028 Chr.2: 176123965-176123965 on Build GRCh38
rs984922159 Chr.2: 176124039-176124039 on Build GRCh38
rs886230673 Chr.2: 176124283-176124283 on Build GRCh38
rs768624482 Chr.2: 176123920-176123920 on Build GRCh38
rs764121479 Chr.2: 176124135-176124135 on Build GRCh38
rs781314575 Chr.2: 176124185-176124185 on Build GRCh38
rs939529072 Chr.2: 176123838-176123838 on Build GRCh38
rs759158050 Chr.2: 176124159-176124160 on Build GRCh38

View Full Product Details