Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 182833198-182833697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRZB
Entrez Gene ID
2487
Gene Name
frizzled-related protein
Gene Aliases
FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, OS1, SFRP3, SRFP3, hFIZ
UniGene
Hs.128453
Gene Chromosome Location
Chr.2: 182833275-182866770 on Build GRCh38
Transcripts
NM_001463.3

SNP Detail

SNP ID Chromosome Location
rs943572819 Chr.2: 182833247-182833247 on Build GRCh38
rs561211006 Chr.2: 182833581-182833581 on Build GRCh38
rs574818487 Chr.2: 182833619-182833619 on Build GRCh38
rs1053856061 Chr.2: 182833254-182833254 on Build GRCh38
rs1024529944 Chr.2: 182833453-182833453 on Build GRCh38
rs1005824160 Chr.2: 182833344-182833344 on Build GRCh38
rs542211536 Chr.2: 182833611-182833612 on Build GRCh38
rs891946574 Chr.2: 182833437-182833437 on Build GRCh38
rs944637176 Chr.2: 182833537-182833537 on Build GRCh38
rs776125909 Chr.2: 182833572-182833572 on Build GRCh38
rs376840088 Chr.2: 182833499-182833499 on Build GRCh38
rs937775965 Chr.2: 182833224-182833224 on Build GRCh38
rs746723485 Chr.2: 182833472-182833472 on Build GRCh38
rs920449444 Chr.2: 182833501-182833501 on Build GRCh38
rs1029338851 Chr.2: 182833399-182833399 on Build GRCh38
rs778720984 Chr.2: 182833345-182833345 on Build GRCh38
rs955058449 Chr.2: 182833405-182833405 on Build GRCh38
rs187652697 Chr.2: 182833230-182833230 on Build GRCh38
rs900481824 Chr.2: 182833666-182833666 on Build GRCh38
rs572918546 Chr.2: 182833306-182833306 on Build GRCh38
rs1047613260 Chr.2: 182833291-182833291 on Build GRCh38
rs943536468 Chr.2: 182833470-182833470 on Build GRCh38
rs929247501 Chr.2: 182833259-182833259 on Build GRCh38
rs4666865 Chr.2: 182833364-182833364 on Build GRCh38
rs970386143 Chr.2: 182833465-182833465 on Build GRCh38
rs746318745 Chr.2: 182833610-182833610 on Build GRCh38
rs906538289 Chr.2: 182833515-182833515 on Build GRCh38
rs959235173 Chr.2: 182833333-182833333 on Build GRCh38
rs1034050441 Chr.2: 182833307-182833307 on Build GRCh38
rs771942767 Chr.2: 182833386-182833386 on Build GRCh38
rs777269003 Chr.2: 182833419-182833419 on Build GRCh38
rs1033340326 Chr.2: 182833480-182833480 on Build GRCh38
rs893910831 Chr.2: 182833272-182833272 on Build GRCh38
rs895085586 Chr.2: 182833565-182833565 on Build GRCh38
rs75221156 Chr.2: 182833427-182833427 on Build GRCh38
rs939395927 Chr.2: 182833517-182833517 on Build GRCh38
rs781485360 Chr.2: 182833240-182833240 on Build GRCh38
rs915303653 Chr.2: 182833481-182833481 on Build GRCh38
rs288320 Chr.2: 182833506-182833506 on Build GRCh38
rs1009496578 Chr.2: 182833283-182833283 on Build GRCh38
rs1039120111 Chr.2: 182833355-182833355 on Build GRCh38
rs987738571 Chr.2: 182833407-182833407 on Build GRCh38
rs754977526 Chr.2: 182833298-182833298 on Build GRCh38
rs190968687 Chr.2: 182833520-182833520 on Build GRCh38
rs753760794 Chr.2: 182833293-182833293 on Build GRCh38

View Full Product Details