Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 182833780-182834323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRZB
Entrez Gene ID
2487
Gene Name
frizzled-related protein
Gene Aliases
FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, OS1, SFRP3, SRFP3, hFIZ
UniGene
Hs.128453
Gene Chromosome Location
Chr.2: 182833275-182866770 on Build GRCh38
Transcripts
NM_001463.3

SNP Detail

SNP ID Chromosome Location
rs950505020 Chr.2: 182834140-182834140 on Build GRCh38
rs531132600 Chr.2: 182833813-182833813 on Build GRCh38
rs921939917 Chr.2: 182833914-182833914 on Build GRCh38
rs184162598 Chr.2: 182834155-182834155 on Build GRCh38
rs540484795 Chr.2: 182834277-182834277 on Build GRCh38
rs978261195 Chr.2: 182834271-182834271 on Build GRCh38
rs765054568 Chr.2: 182834299-182834299 on Build GRCh38
rs891922071 Chr.2: 182833961-182833961 on Build GRCh38
rs1011567835 Chr.2: 182834180-182834180 on Build GRCh38
rs1804713 Chr.2: 182834166-182834166 on Build GRCh38
rs941280849 Chr.2: 182833993-182833993 on Build GRCh38
rs796993095 Chr.2: 182834009-182834009 on Build GRCh38
rs1045826685 Chr.2: 182834287-182834287 on Build GRCh38
rs944644081 Chr.2: 182833884-182833884 on Build GRCh38
rs16823799 Chr.2: 182833964-182833964 on Build GRCh38
rs990800960 Chr.2: 182834298-182834298 on Build GRCh38
rs950556273 Chr.2: 182833858-182833859 on Build GRCh38
rs896988022 Chr.2: 182834063-182834063 on Build GRCh38
rs61235169 Chr.2: 182834001-182834001 on Build GRCh38
rs113414222 Chr.2: 182833970-182833970 on Build GRCh38
rs368734908 Chr.2: 182834268-182834268 on Build GRCh38
rs143600540 Chr.2: 182833834-182833834 on Build GRCh38
rs398102385 Chr.2: 182834003-182834003 on Build GRCh38
rs16823797 Chr.2: 182833872-182833872 on Build GRCh38
rs961075794 Chr.2: 182834285-182834285 on Build GRCh38
rs112035616 Chr.2: 182833865-182833865 on Build GRCh38
rs1056024049 Chr.2: 182834001-182834002 on Build GRCh38
rs989626142 Chr.2: 182833865-182833865 on Build GRCh38
rs935981129 Chr.2: 182833941-182833941 on Build GRCh38
rs547205904 Chr.2: 182833859-182833860 on Build GRCh38
rs538027984 Chr.2: 182834184-182834184 on Build GRCh38
rs895022429 Chr.2: 182833972-182833972 on Build GRCh38
rs894812492 Chr.2: 182834071-182834071 on Build GRCh38
rs774708256 Chr.2: 182833943-182833943 on Build GRCh38
rs150220037 Chr.2: 182834255-182834255 on Build GRCh38
rs994423926 Chr.2: 182834104-182834104 on Build GRCh38
rs762129004 Chr.2: 182833952-182833952 on Build GRCh38
rs1022544593 Chr.2: 182834187-182834187 on Build GRCh38
rs964957282 Chr.2: 182834172-182834172 on Build GRCh38
rs1018361444 Chr.2: 182834171-182834171 on Build GRCh38
rs536514866 Chr.2: 182833946-182833946 on Build GRCh38
rs975073125 Chr.2: 182834188-182834188 on Build GRCh38
rs111764346 Chr.2: 182833932-182833932 on Build GRCh38
rs376582388 Chr.2: 182833859-182833859 on Build GRCh38
rs750552766 Chr.2: 182834111-182834111 on Build GRCh38
rs538686307 Chr.2: 182833992-182833992 on Build GRCh38
rs990181238 Chr.2: 182834272-182834273 on Build GRCh38
rs529736746 Chr.2: 182833931-182833931 on Build GRCh38
rs1038327524 Chr.2: 182834028-182834028 on Build GRCh38
rs111718123 Chr.2: 182834158-182834158 on Build GRCh38
rs763202011 Chr.2: 182833845-182833845 on Build GRCh38
rs554432959 Chr.2: 182834225-182834225 on Build GRCh38
rs946139397 Chr.2: 182834280-182834280 on Build GRCh38
rs978147187 Chr.2: 182833942-182833942 on Build GRCh38
rs974663113 Chr.2: 182833886-182833886 on Build GRCh38
rs994025438 Chr.2: 182834101-182834101 on Build GRCh38
rs1027275760 Chr.2: 182834131-182834131 on Build GRCh38

View Full Product Details