Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 186743393-186743896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM171B
Entrez Gene ID
165215
Gene Name
family with sequence similarity 171 member B
Gene Aliases
KIAA1946
UniGene
Hs.28872
Gene Chromosome Location
Chr.2: 186694062-186763787 on Build GRCh38
Transcripts
NM_177454.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233767 c.503C>T p.P168L Chr.2: 186743513-186743513 on Build GRCh38
3575199 c.494C>T p.S165L Chr.2: 186743504-186743504 on Build GRCh38
6567294 c.479C>T p.S160L Chr.2: 186743489-186743489 on Build GRCh38
3838115 c.515C>G p.A172G Chr.2: 186743525-186743525 on Build GRCh38
6863280 c.565+6T>C p.? Chr.2: 186743581-186743581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755969537 Chr.2: 186743513-186743513 on Build GRCh38
rs545808435 Chr.2: 186743703-186743703 on Build GRCh38
rs184037266 Chr.2: 186743659-186743659 on Build GRCh38
rs748874709 Chr.2: 186743444-186743444 on Build GRCh38
rs555856843 Chr.2: 186743483-186743483 on Build GRCh38
rs763964246 Chr.2: 186743607-186743612 on Build GRCh38
rs767415189 Chr.2: 186743638-186743638 on Build GRCh38
rs752546021 Chr.2: 186743511-186743511 on Build GRCh38
rs759462338 Chr.2: 186743629-186743629 on Build GRCh38
rs771319738 Chr.2: 186743427-186743427 on Build GRCh38
rs767515720 Chr.2: 186743621-186743621 on Build GRCh38
rs776882387 Chr.2: 186743460-186743460 on Build GRCh38
rs756865734 Chr.2: 186743548-186743548 on Build GRCh38
rs752584826 Chr.2: 186743438-186743438 on Build GRCh38
rs762103498 Chr.2: 186743465-186743465 on Build GRCh38
rs756082727 Chr.2: 186743591-186743594 on Build GRCh38
rs1030020413 Chr.2: 186743808-186743808 on Build GRCh38
rs753520734 Chr.2: 186743545-186743545 on Build GRCh38
rs977844329 Chr.2: 186743859-186743859 on Build GRCh38
rs769056285 Chr.2: 186743563-186743563 on Build GRCh38
rs576950718 Chr.2: 186743828-186743830 on Build GRCh38
rs780662259 Chr.2: 186743451-186743451 on Build GRCh38
rs974785764 Chr.2: 186743701-186743701 on Build GRCh38
rs745333347 Chr.2: 186743555-186743555 on Build GRCh38
rs767436066 Chr.2: 186743508-186743508 on Build GRCh38
rs937660234 Chr.2: 186743824-186743824 on Build GRCh38
rs965994711 Chr.2: 186743788-186743788 on Build GRCh38
rs370401384 Chr.2: 186743752-186743752 on Build GRCh38
rs998420129 Chr.2: 186743794-186743794 on Build GRCh38
rs778316060 Chr.2: 186743550-186743550 on Build GRCh38
rs539944674 Chr.2: 186743837-186743837 on Build GRCh38
rs979732977 Chr.2: 186743814-186743814 on Build GRCh38
rs534836678 Chr.2: 186743472-186743472 on Build GRCh38
rs74519381 Chr.2: 186743827-186743827 on Build GRCh38
rs747441131 Chr.2: 186743452-186743452 on Build GRCh38
rs994229225 Chr.2: 186743645-186743645 on Build GRCh38
rs925111498 Chr.2: 186743438-186743438 on Build GRCh38
rs770553304 Chr.2: 186743446-186743446 on Build GRCh38
rs557308183 Chr.2: 186743620-186743620 on Build GRCh38
rs1046367228 Chr.2: 186743528-186743528 on Build GRCh38
rs767403000 Chr.2: 186743437-186743438 on Build GRCh38
rs913700500 Chr.2: 186743762-186743762 on Build GRCh38
rs750292049 Chr.2: 186743660-186743660 on Build GRCh38
rs888628000 Chr.2: 186743671-186743671 on Build GRCh38
rs74449843 Chr.2: 186743604-186743604 on Build GRCh38
rs202060142 Chr.2: 186743457-186743457 on Build GRCh38
rs372016071 Chr.2: 186743737-186743737 on Build GRCh38
rs751621711 Chr.2: 186743503-186743503 on Build GRCh38
rs925018192 Chr.2: 186743865-186743865 on Build GRCh38
rs375020464 Chr.2: 186743599-186743599 on Build GRCh38
rs774692129 Chr.2: 186743467-186743467 on Build GRCh38
rs374624191 Chr.2: 186743866-186743866 on Build GRCh38
rs1030820628 Chr.2: 186743712-186743712 on Build GRCh38
rs770128461 Chr.2: 186743603-186743603 on Build GRCh38
rs542293336 Chr.2: 186743618-186743618 on Build GRCh38
rs767422183 Chr.2: 186743625-186743625 on Build GRCh38
rs370337149 Chr.2: 186743474-186743474 on Build GRCh38
rs865815859 Chr.2: 186743733-186743733 on Build GRCh38
rs759418419 Chr.2: 186743505-186743505 on Build GRCh38
rs781672286 Chr.2: 186743598-186743598 on Build GRCh38
rs1016520789 Chr.2: 186743675-186743675 on Build GRCh38
rs965047960 Chr.2: 186743685-186743685 on Build GRCh38
rs777627299 Chr.2: 186743529-186743529 on Build GRCh38
rs777500589 Chr.2: 186743440-186743440 on Build GRCh38
rs1018568199 Chr.2: 186743757-186743757 on Build GRCh38
rs201718626 Chr.2: 186743487-186743487 on Build GRCh38

View Full Product Details