Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 188988398-188988897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL3A1
Entrez Gene ID
1281
Gene Name
collagen type III alpha 1 chain
Gene Aliases
EDS4A
UniGene
Hs.443625
Gene Chromosome Location
Chr.2: 188974373-189012746 on Build GRCh38
Transcripts
NM_000090.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5937990 c.620G>A p.G207E Chr.2: 188988627-188988627 on Build GRCh38
3372568 c.606C>A p.P202P Chr.2: 188988613-188988613 on Build GRCh38
5937989 c.620G>A p.G207E Chr.2: 188988627-188988627 on Build GRCh38
3575345 c.607C>T p.P203S Chr.2: 188988614-188988614 on Build GRCh38
3575344 c.607C>T p.P203S Chr.2: 188988614-188988614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527415370 Chr.2: 188988631-188988631 on Build GRCh38
rs3134659 Chr.2: 188988688-188988688 on Build GRCh38
rs587779592 Chr.2: 188988627-188988627 on Build GRCh38
rs756128490 Chr.2: 188988423-188988423 on Build GRCh38
rs986626006 Chr.2: 188988770-188988770 on Build GRCh38
rs587779681 Chr.2: 188988638-188988649 on Build GRCh38
rs560255534 Chr.2: 188988616-188988616 on Build GRCh38
rs10166835 Chr.2: 188988582-188988582 on Build GRCh38
rs879259575 Chr.2: 188988736-188988736 on Build GRCh38
rs587779688 Chr.2: 188988648-188988648 on Build GRCh38
rs560984169 Chr.2: 188988677-188988677 on Build GRCh38
rs751350498 Chr.2: 188988593-188988593 on Build GRCh38
rs754708730 Chr.2: 188988595-188988595 on Build GRCh38
rs780921226 Chr.2: 188988606-188988606 on Build GRCh38
rs946397344 Chr.2: 188988454-188988454 on Build GRCh38
rs375362835 Chr.2: 188988675-188988675 on Build GRCh38
rs56094115 Chr.2: 188988734-188988734 on Build GRCh38
rs746358771 Chr.2: 188988629-188988629 on Build GRCh38
rs779352711 Chr.2: 188988628-188988628 on Build GRCh38
rs926285757 Chr.2: 188988829-188988829 on Build GRCh38
rs573870374 Chr.2: 188988864-188988864 on Build GRCh38
rs768871160 Chr.2: 188988670-188988670 on Build GRCh38
rs966308127 Chr.2: 188988783-188988783 on Build GRCh38
rs767333683 Chr.2: 188988692-188988692 on Build GRCh38
rs979067911 Chr.2: 188988820-188988820 on Build GRCh38
rs751697157 Chr.2: 188988529-188988529 on Build GRCh38
rs587779436 Chr.2: 188988608-188988608 on Build GRCh38
rs886055330 Chr.2: 188988583-188988583 on Build GRCh38
rs747338007 Chr.2: 188988655-188988655 on Build GRCh38
rs538800273 Chr.2: 188988767-188988767 on Build GRCh38
rs761886651 Chr.2: 188988542-188988542 on Build GRCh38
rs587779676 Chr.2: 188988644-188988644 on Build GRCh38
rs1027631874 Chr.2: 188988744-188988744 on Build GRCh38
rs587779601 Chr.2: 188988599-188988599 on Build GRCh38
rs921067518 Chr.2: 188988498-188988498 on Build GRCh38
rs913848809 Chr.2: 188988815-188988815 on Build GRCh38
rs587779626 Chr.2: 188988618-188988618 on Build GRCh38
rs755622839 Chr.2: 188988695-188988695 on Build GRCh38
rs377380502 Chr.2: 188988676-188988676 on Build GRCh38
rs550009137 Chr.2: 188988729-188988729 on Build GRCh38
rs975290892 Chr.2: 188988834-188988840 on Build GRCh38
rs531497438 Chr.2: 188988678-188988678 on Build GRCh38
rs755634638 Chr.2: 188988621-188988621 on Build GRCh38
rs776724064 Chr.2: 188988540-188988540 on Build GRCh38
rs587779628 Chr.2: 188988636-188988637 on Build GRCh38
rs776777474 Chr.2: 188988671-188988671 on Build GRCh38
rs765101672 Chr.2: 188988543-188988543 on Build GRCh38
rs988065775 Chr.2: 188988811-188988811 on Build GRCh38
rs750363113 Chr.2: 188988568-188988568 on Build GRCh38
rs370561283 Chr.2: 188988672-188988672 on Build GRCh38
rs587779641 Chr.2: 188988600-188988600 on Build GRCh38
rs962629478 Chr.2: 188988832-188988832 on Build GRCh38
rs587779711 Chr.2: 188988617-188988617 on Build GRCh38
rs1055934953 Chr.2: 188988660-188988660 on Build GRCh38
rs566612250 Chr.2: 188988788-188988788 on Build GRCh38
rs41272809 Chr.2: 188988769-188988769 on Build GRCh38
rs1018740005 Chr.2: 188988774-188988774 on Build GRCh38
rs587779515 Chr.2: 188988636-188988636 on Build GRCh38
rs763535262 Chr.2: 188988464-188988464 on Build GRCh38
rs758167432 Chr.2: 188988576-188988576 on Build GRCh38
rs937736996 Chr.2: 188988868-188988868 on Build GRCh38
rs909832476 Chr.2: 188988812-188988812 on Build GRCh38
rs764484048 Chr.2: 188988483-188988483 on Build GRCh38
rs780423624 Chr.2: 188988649-188988649 on Build GRCh38
rs1043480253 Chr.2: 188988473-188988473 on Build GRCh38
rs267599120 Chr.2: 188988590-188988590 on Build GRCh38
rs752387616 Chr.2: 188988693-188988693 on Build GRCh38
rs35435280 Chr.2: 188988739-188988740 on Build GRCh38
rs967687762 Chr.2: 188988817-188988817 on Build GRCh38
rs897819411 Chr.2: 188988719-188988719 on Build GRCh38

View Full Product Details