Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 188989310-188989822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL3A1
Entrez Gene ID
1281
Gene Name
collagen type III alpha 1 chain
Gene Aliases
EDS4A
UniGene
Hs.443625
Gene Chromosome Location
Chr.2: 188974373-189012746 on Build GRCh38
Transcripts
NM_000090.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6031719 c.682G>T p.G228* Chr.2: 188989441-188989441 on Build GRCh38
230950 c.667C>T p.P223S Chr.2: 188989426-188989426 on Build GRCh38
3838126 c.667C>T p.P223S Chr.2: 188989426-188989426 on Build GRCh38
3575346 c.644C>T p.P215L Chr.2: 188989403-188989403 on Build GRCh38
5035354 c.637-1G>A p.? Chr.2: 188989395-188989395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049337831 Chr.2: 188989338-188989338 on Build GRCh38
rs1009667574 Chr.2: 188989380-188989380 on Build GRCh38
rs587779470 Chr.2: 188989441-188989441 on Build GRCh38
rs959054633 Chr.2: 188989440-188989440 on Build GRCh38
rs771473939 Chr.2: 188989564-188989564 on Build GRCh38
rs1028285260 Chr.2: 188989538-188989538 on Build GRCh38
rs776043531 Chr.2: 188989637-188989637 on Build GRCh38
rs986701856 Chr.2: 188989571-188989573 on Build GRCh38
rs12619321 Chr.2: 188989611-188989611 on Build GRCh38
rs930845858 Chr.2: 188989619-188989619 on Build GRCh38
rs759468533 Chr.2: 188989498-188989498 on Build GRCh38
rs1020764737 Chr.2: 188989393-188989393 on Build GRCh38
rs58220378 Chr.2: 188989623-188989623 on Build GRCh38
rs41272815 Chr.2: 188989708-188989708 on Build GRCh38
rs898529214 Chr.2: 188989511-188989511 on Build GRCh38
rs1004725785 Chr.2: 188989478-188989478 on Build GRCh38
rs587779497 Chr.2: 188989451-188989451 on Build GRCh38
rs901900335 Chr.2: 188989377-188989377 on Build GRCh38
rs587779624 Chr.2: 188989414-188989414 on Build GRCh38
rs747397336 Chr.2: 188989373-188989374 on Build GRCh38
rs1042530574 Chr.2: 188989657-188989657 on Build GRCh38
rs13388534 Chr.2: 188989733-188989733 on Build GRCh38
rs11886142 Chr.2: 188989473-188989473 on Build GRCh38
rs587779557 Chr.2: 188989396-188989396 on Build GRCh38
rs79949505 Chr.2: 188989548-188989548 on Build GRCh38
rs12616391 Chr.2: 188989622-188989622 on Build GRCh38
rs543247004 Chr.2: 188989791-188989791 on Build GRCh38
rs1007999040 Chr.2: 188989359-188989359 on Build GRCh38
rs769093858 Chr.2: 188989449-188989449 on Build GRCh38
rs587779518 Chr.2: 188989424-188989424 on Build GRCh38
rs41272813 Chr.2: 188989705-188989705 on Build GRCh38
rs889112083 Chr.2: 188989344-188989344 on Build GRCh38
rs747387258 Chr.2: 188989381-188989381 on Build GRCh38
rs386653662 Chr.2: 188989622-188989623 on Build GRCh38
rs41272811 Chr.2: 188989629-188989629 on Build GRCh38
rs972447403 Chr.2: 188989735-188989735 on Build GRCh38
rs370451250 Chr.2: 188989458-188989458 on Build GRCh38
rs774413781 Chr.2: 188989477-188989477 on Build GRCh38
rs368321073 Chr.2: 188989623-188989623 on Build GRCh38
rs866341501 Chr.2: 188989427-188989427 on Build GRCh38
rs587779555 Chr.2: 188989442-188989442 on Build GRCh38
rs780472970 Chr.2: 188989363-188989363 on Build GRCh38
rs1034042392 Chr.2: 188989428-188989428 on Build GRCh38
rs967913772 Chr.2: 188989400-188989400 on Build GRCh38
rs768997695 Chr.2: 188989435-188989435 on Build GRCh38
rs748378490 Chr.2: 188989460-188989460 on Build GRCh38
rs1040441323 Chr.2: 188989373-188989373 on Build GRCh38
rs1025023340 Chr.2: 188989716-188989716 on Build GRCh38
rs764725713 Chr.2: 188989737-188989737 on Build GRCh38
rs536044672 Chr.2: 188989614-188989614 on Build GRCh38
rs903675523 Chr.2: 188989662-188989662 on Build GRCh38
rs1000724773 Chr.2: 188989669-188989669 on Build GRCh38
rs572631735 Chr.2: 188989781-188989781 on Build GRCh38
rs961385848 Chr.2: 188989509-188989509 on Build GRCh38
rs910733165 Chr.2: 188989796-188989796 on Build GRCh38
rs113535225 Chr.2: 188989642-188989642 on Build GRCh38
rs1026717795 Chr.2: 188989745-188989745 on Build GRCh38
rs758938813 Chr.2: 188989361-188989361 on Build GRCh38
rs867258281 Chr.2: 188989418-188989418 on Build GRCh38
rs1021224796 Chr.2: 188989577-188989577 on Build GRCh38
rs763191944 Chr.2: 188989680-188989680 on Build GRCh38
rs753243435 Chr.2: 188989355-188989355 on Build GRCh38
rs374426800 Chr.2: 188989464-188989464 on Build GRCh38
rs565154375 Chr.2: 188989506-188989506 on Build GRCh38
rs996833144 Chr.2: 188989537-188989538 on Build GRCh38
rs894726056 Chr.2: 188989340-188989340 on Build GRCh38
rs188048066 Chr.2: 188989696-188989696 on Build GRCh38
rs1000777138 Chr.2: 188989404-188989404 on Build GRCh38
rs547171357 Chr.2: 188989522-188989522 on Build GRCh38
rs532309541 Chr.2: 188989507-188989507 on Build GRCh38
rs587779441 Chr.2: 188989415-188989415 on Build GRCh38
rs587779533 Chr.2: 188989433-188989433 on Build GRCh38
rs587779596 Chr.2: 188989406-188989406 on Build GRCh38
rs979178697 Chr.2: 188989620-188989620 on Build GRCh38
rs531290981 Chr.2: 188989462-188989462 on Build GRCh38
rs891104001 Chr.2: 188989788-188989788 on Build GRCh38
rs996883680 Chr.2: 188989762-188989762 on Build GRCh38
rs556315634 Chr.2: 188989633-188989633 on Build GRCh38
rs587779660 Chr.2: 188989450-188989450 on Build GRCh38

View Full Product Details