Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189666308-189666844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASNSD1
Entrez Gene ID
54529
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
NBLA00058, NS3TP1
UniGene
Hs.101364
Gene Chromosome Location
Chr.2: 189661399-189670831 on Build GRCh38
Transcripts
NM_019048.2 XM_017004381.1 XM_017004382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1404042 c.407_408insT p.G139fs*4 Chr.2: 189666539-189666540 on Build GRCh38
6444979 c.667C>T p.P223S Chr.2: 189666799-189666799 on Build GRCh38
4845705 c.309G>A p.K103K Chr.2: 189666441-189666441 on Build GRCh38
4879173 c.466C>G p.L156V Chr.2: 189666598-189666598 on Build GRCh38
6448559 c.422G>A p.R141H Chr.2: 189666554-189666554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745896187 Chr.2: 189666655-189666655 on Build GRCh38
rs781141260 Chr.2: 189666652-189666652 on Build GRCh38
rs776929142 Chr.2: 189666409-189666409 on Build GRCh38
rs776758861 Chr.2: 189666416-189666416 on Build GRCh38
rs896217136 Chr.2: 189666563-189666563 on Build GRCh38
rs143704216 Chr.2: 189666471-189666471 on Build GRCh38
rs748399229 Chr.2: 189666755-189666755 on Build GRCh38
rs751276134 Chr.2: 189666774-189666774 on Build GRCh38
rs966113884 Chr.2: 189666817-189666817 on Build GRCh38
rs772012444 Chr.2: 189666377-189666377 on Build GRCh38
rs761620700 Chr.2: 189666801-189666801 on Build GRCh38
rs778836315 Chr.2: 189666366-189666366 on Build GRCh38
rs759146718 Chr.2: 189666602-189666602 on Build GRCh38
rs201813790 Chr.2: 189666478-189666478 on Build GRCh38
rs146351895 Chr.2: 189666734-189666734 on Build GRCh38
rs553670623 Chr.2: 189666492-189666492 on Build GRCh38
rs146979789 Chr.2: 189666463-189666463 on Build GRCh38
rs759722534 Chr.2: 189666539-189666540 on Build GRCh38
rs774842030 Chr.2: 189666568-189666568 on Build GRCh38
rs373960766 Chr.2: 189666661-189666661 on Build GRCh38
rs756106982 Chr.2: 189666811-189666811 on Build GRCh38
rs779791970 Chr.2: 189666338-189666338 on Build GRCh38
rs1052058238 Chr.2: 189666355-189666355 on Build GRCh38
rs758143139 Chr.2: 189666509-189666509 on Build GRCh38
rs762214044 Chr.2: 189666589-189666589 on Build GRCh38
rs752490005 Chr.2: 189666503-189666503 on Build GRCh38
rs1022194153 Chr.2: 189666423-189666423 on Build GRCh38
rs748584822 Chr.2: 189666704-189666704 on Build GRCh38
rs759050855 Chr.2: 189666735-189666735 on Build GRCh38
rs745991037 Chr.2: 189666541-189666541 on Build GRCh38
rs752297560 Chr.2: 189666608-189666608 on Build GRCh38
rs371537991 Chr.2: 189666418-189666418 on Build GRCh38
rs754743584 Chr.2: 189666363-189666363 on Build GRCh38
rs750362135 Chr.2: 189666806-189666806 on Build GRCh38
rs749463576 Chr.2: 189666682-189666682 on Build GRCh38
rs140748135 Chr.2: 189666804-189666804 on Build GRCh38
rs535848516 Chr.2: 189666373-189666373 on Build GRCh38
rs909013950 Chr.2: 189666738-189666738 on Build GRCh38
rs763784040 Chr.2: 189666620-189666620 on Build GRCh38
rs548573736 Chr.2: 189666762-189666762 on Build GRCh38
rs1437880 Chr.2: 189666700-189666700 on Build GRCh38
rs768902902 Chr.2: 189666554-189666554 on Build GRCh38
rs776106961 Chr.2: 189666334-189666334 on Build GRCh38
rs746917768 Chr.2: 189666525-189666525 on Build GRCh38
rs758137583 Chr.2: 189666611-189666611 on Build GRCh38
rs770920386 Chr.2: 189666415-189666415 on Build GRCh38
rs763256540 Chr.2: 189666441-189666441 on Build GRCh38
rs751424792 Chr.2: 189666625-189666625 on Build GRCh38
rs200125189 Chr.2: 189666551-189666551 on Build GRCh38
rs377319888 Chr.2: 189666747-189666747 on Build GRCh38
rs763026988 Chr.2: 189666455-189666455 on Build GRCh38
rs974787806 Chr.2: 189666788-189666788 on Build GRCh38
rs769424635 Chr.2: 189666739-189666739 on Build GRCh38
rs753541123 Chr.2: 189666340-189666340 on Build GRCh38
rs1043613339 Chr.2: 189666529-189666529 on Build GRCh38
rs765020623 Chr.2: 189666482-189666482 on Build GRCh38
rs115346256 Chr.2: 189666812-189666812 on Build GRCh38
rs775803023 Chr.2: 189666437-189666437 on Build GRCh38
rs765982507 Chr.2: 189666469-189666469 on Build GRCh38
rs139770698 Chr.2: 189666802-189666802 on Build GRCh38
rs754653621 Chr.2: 189666480-189666480 on Build GRCh38
rs781229792 Chr.2: 189666535-189666535 on Build GRCh38
rs1437879 Chr.2: 189666687-189666687 on Build GRCh38
rs780251750 Chr.2: 189666681-189666681 on Build GRCh38
rs200920806 Chr.2: 189666553-189666553 on Build GRCh38
rs953531793 Chr.2: 189666330-189666330 on Build GRCh38
rs766433525 Chr.2: 189666519-189666522 on Build GRCh38
rs188664014 Chr.2: 189666468-189666468 on Build GRCh38
rs767888340 Chr.2: 189666796-189666796 on Build GRCh38
rs757158815 Chr.2: 189666631-189666631 on Build GRCh38
rs76917094 Chr.2: 189666348-189666348 on Build GRCh38
rs769966669 Chr.2: 189666548-189666548 on Build GRCh38
rs201324299 Chr.2: 189666540-189666540 on Build GRCh38
rs200095933 Chr.2: 189666539-189666539 on Build GRCh38
rs764926809 Chr.2: 189666605-189666605 on Build GRCh38
rs757224399 Chr.2: 189666429-189666429 on Build GRCh38
rs768980049 Chr.2: 189666688-189666688 on Build GRCh38
rs777700209 Chr.2: 189666511-189666511 on Build GRCh38
rs747054433 Chr.2: 189666406-189666406 on Build GRCh38
rs1036710021 Chr.2: 189666597-189666597 on Build GRCh38
rs193167266 Chr.2: 189666534-189666534 on Build GRCh38
rs768186476 Chr.2: 189666590-189666590 on Build GRCh38
rs776248492 Chr.2: 189666594-189666594 on Build GRCh38
rs564425055 Chr.2: 189666753-189666753 on Build GRCh38
rs148737403 Chr.2: 189666717-189666717 on Build GRCh38
rs377372887 Chr.2: 189666496-189666496 on Build GRCh38
rs777595485 Chr.2: 189666385-189666385 on Build GRCh38
rs138256489 Chr.2: 189666443-189666443 on Build GRCh38
rs1010434591 Chr.2: 189666380-189666380 on Build GRCh38
rs753143035 Chr.2: 189666813-189666815 on Build GRCh38
rs774615459 Chr.2: 189666540-189666540 on Build GRCh38

View Full Product Details