Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190244808-190245308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIBCH
Entrez Gene ID
26275
Gene Name
3-hydroxyisobutyryl-CoA hydrolase
Gene Aliases
HIBYLCOAH
UniGene
Hs.656685
Gene Chromosome Location
Chr.2: 190204634-190320045 on Build GRCh38
Transcripts
NM_014362.3 NM_198047.2 XM_011510953.1 XM_011510954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1014091 c.891+1G>A p.? Chr.2: 190244886-190244886 on Build GRCh38
6392668 c.820G>T p.A274S Chr.2: 190244958-190244958 on Build GRCh38
6487174 c.891+1G>C p.? Chr.2: 190244886-190244886 on Build GRCh38
5861360 c.875C>T p.A292V Chr.2: 190244903-190244903 on Build GRCh38
320817 c.866C>T p.S289L Chr.2: 190244912-190244912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757899313 Chr.2: 190244966-190244966 on Build GRCh38
rs937435447 Chr.2: 190245186-190245186 on Build GRCh38
rs924136882 Chr.2: 190245184-190245184 on Build GRCh38
rs760590354 Chr.2: 190245006-190245006 on Build GRCh38
rs369575301 Chr.2: 190245014-190245014 on Build GRCh38
rs527426569 Chr.2: 190245269-190245269 on Build GRCh38
rs747515363 Chr.2: 190244952-190244952 on Build GRCh38
rs976941064 Chr.2: 190245219-190245219 on Build GRCh38
rs922288490 Chr.2: 190244964-190244964 on Build GRCh38
rs111910389 Chr.2: 190245046-190245046 on Build GRCh38
rs778451600 Chr.2: 190244848-190244848 on Build GRCh38
rs549618573 Chr.2: 190245240-190245240 on Build GRCh38
rs138786617 Chr.2: 190244925-190244925 on Build GRCh38
rs755975088 Chr.2: 190244931-190244931 on Build GRCh38
rs768124839 Chr.2: 190245053-190245056 on Build GRCh38
rs375349694 Chr.2: 190244841-190244841 on Build GRCh38
rs913481669 Chr.2: 190245052-190245052 on Build GRCh38
rs573302664 Chr.2: 190244972-190244972 on Build GRCh38
rs769400158 Chr.2: 190244855-190244855 on Build GRCh38
rs558122884 Chr.2: 190244842-190244842 on Build GRCh38
rs773897332 Chr.2: 190244999-190244999 on Build GRCh38
rs377648933 Chr.2: 190245259-190245259 on Build GRCh38
rs369232582 Chr.2: 190244986-190244986 on Build GRCh38
rs974451994 Chr.2: 190245031-190245031 on Build GRCh38
rs979186292 Chr.2: 190245139-190245139 on Build GRCh38
rs577861252 Chr.2: 190245218-190245218 on Build GRCh38
rs1035021408 Chr.2: 190245092-190245092 on Build GRCh38
rs915412371 Chr.2: 190244886-190244886 on Build GRCh38
rs570799904 Chr.2: 190245029-190245029 on Build GRCh38
rs753060412 Chr.2: 190244935-190244935 on Build GRCh38
rs991514611 Chr.2: 190244880-190244880 on Build GRCh38
rs752916501 Chr.2: 190244972-190244973 on Build GRCh38
rs879203473 Chr.2: 190245074-190245074 on Build GRCh38
rs776703180 Chr.2: 190245017-190245017 on Build GRCh38
rs568264065 Chr.2: 190245172-190245172 on Build GRCh38
rs959676588 Chr.2: 190245101-190245101 on Build GRCh38
rs775208899 Chr.2: 190244859-190244859 on Build GRCh38
rs756444230 Chr.2: 190245002-190245005 on Build GRCh38
rs187705610 Chr.2: 190245055-190245055 on Build GRCh38
rs556749864 Chr.2: 190245265-190245265 on Build GRCh38
rs773055524 Chr.2: 190245004-190245004 on Build GRCh38
rs755220161 Chr.2: 190244837-190244837 on Build GRCh38
rs535227988 Chr.2: 190245241-190245241 on Build GRCh38
rs955776020 Chr.2: 190245208-190245208 on Build GRCh38
rs747881400 Chr.2: 190245000-190245000 on Build GRCh38
rs778325186 Chr.2: 190244946-190244946 on Build GRCh38
rs758213487 Chr.2: 190245028-190245028 on Build GRCh38
rs750268138 Chr.2: 190244850-190244850 on Build GRCh38
rs750337514 Chr.2: 190244915-190244915 on Build GRCh38
rs543956560 Chr.2: 190245102-190245102 on Build GRCh38
rs755166207 Chr.2: 190244928-190244928 on Build GRCh38
rs2303827 Chr.2: 190245144-190245144 on Build GRCh38
rs778683855 Chr.2: 190244893-190244893 on Build GRCh38
rs534285157 Chr.2: 190245002-190245002 on Build GRCh38
rs778147785 Chr.2: 190245004-190245004 on Build GRCh38
rs16832465 Chr.2: 190245257-190245257 on Build GRCh38
rs954872213 Chr.2: 190245048-190245048 on Build GRCh38
rs774129841 Chr.2: 190244904-190244904 on Build GRCh38
rs748753472 Chr.2: 190244975-190244975 on Build GRCh38
rs754662471 Chr.2: 190245115-190245115 on Build GRCh38
rs766312695 Chr.2: 190245007-190245007 on Build GRCh38
rs528509249 Chr.2: 190245131-190245131 on Build GRCh38
rs762656578 Chr.2: 190244862-190244862 on Build GRCh38
rs747570655 Chr.2: 190244854-190244854 on Build GRCh38
rs1003659734 Chr.2: 190245090-190245090 on Build GRCh38
rs1022980195 Chr.2: 190245129-190245129 on Build GRCh38
rs767392058 Chr.2: 190244910-190244910 on Build GRCh38
rs987716572 Chr.2: 190245049-190245049 on Build GRCh38
rs959638999 Chr.2: 190244869-190244869 on Build GRCh38

View Full Product Details