Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190368973-190369490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP1
Entrez Gene ID
3628
Gene Name
inositol polyphosphate-1-phosphatase
Gene Aliases
-
UniGene
Hs.32309
Gene Chromosome Location
Chr.2: 190343470-190371665 on Build GRCh38
Transcripts
NM_001128928.1 NM_002194.3 XM_005246532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
75014 c.559G>T p.G187C Chr.2: 190369195-190369195 on Build GRCh38
3709280 c.605A>G p.N202S Chr.2: 190369241-190369241 on Build GRCh38
6156236 c.580G>T p.G194W Chr.2: 190369216-190369216 on Build GRCh38
6236930 c.641+8G>A p.? Chr.2: 190369285-190369285 on Build GRCh38
5635179 c.533G>C p.G178A Chr.2: 190369169-190369169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371964835 Chr.2: 190369157-190369157 on Build GRCh38
rs374948332 Chr.2: 190369194-190369194 on Build GRCh38
rs755560479 Chr.2: 190369310-190369310 on Build GRCh38
rs1030434795 Chr.2: 190369035-190369035 on Build GRCh38
rs771754946 Chr.2: 190369216-190369216 on Build GRCh38
rs958059060 Chr.2: 190369308-190369308 on Build GRCh38
rs79432845 Chr.2: 190369230-190369230 on Build GRCh38
rs111525525 Chr.2: 190369028-190369028 on Build GRCh38
rs150587539 Chr.2: 190369109-190369109 on Build GRCh38
rs1051494238 Chr.2: 190369434-190369434 on Build GRCh38
rs747739941 Chr.2: 190369084-190369084 on Build GRCh38
rs936249490 Chr.2: 190369346-190369346 on Build GRCh38
rs867216212 Chr.2: 190369152-190369152 on Build GRCh38
rs141259736 Chr.2: 190369408-190369408 on Build GRCh38
rs775457185 Chr.2: 190369259-190369259 on Build GRCh38
rs574069840 Chr.2: 190369240-190369240 on Build GRCh38
rs771877889 Chr.2: 190369100-190369100 on Build GRCh38
rs891945174 Chr.2: 190369361-190369361 on Build GRCh38
rs767192789 Chr.2: 190369113-190369113 on Build GRCh38
rs751770359 Chr.2: 190369280-190369280 on Build GRCh38
rs762990235 Chr.2: 190369266-190369266 on Build GRCh38
rs749044482 Chr.2: 190369224-190369224 on Build GRCh38
rs1108939 Chr.2: 190369464-190369464 on Build GRCh38
rs558627848 Chr.2: 190369079-190369079 on Build GRCh38
rs757560460 Chr.2: 190369185-190369185 on Build GRCh38
rs199940421 Chr.2: 190369218-190369218 on Build GRCh38
rs754517367 Chr.2: 190369258-190369258 on Build GRCh38
rs768042021 Chr.2: 190369083-190369083 on Build GRCh38
rs778529736 Chr.2: 190369204-190369204 on Build GRCh38
rs954379018 Chr.2: 190369056-190369056 on Build GRCh38
rs766234311 Chr.2: 190369112-190369112 on Build GRCh38
rs1007191709 Chr.2: 190369448-190369448 on Build GRCh38
rs746741158 Chr.2: 190369222-190369222 on Build GRCh38
rs368182510 Chr.2: 190369085-190369085 on Build GRCh38
rs751821440 Chr.2: 190369184-190369184 on Build GRCh38
rs189513230 Chr.2: 190369312-190369312 on Build GRCh38
rs764280063 Chr.2: 190369174-190369174 on Build GRCh38
rs541556251 Chr.2: 190369370-190369370 on Build GRCh38
rs144983855 Chr.2: 190369451-190369451 on Build GRCh38
rs752778596 Chr.2: 190369158-190369158 on Build GRCh38
rs184896073 Chr.2: 190369193-190369193 on Build GRCh38
rs892245136 Chr.2: 190369015-190369015 on Build GRCh38
rs1007902392 Chr.2: 190369066-190369066 on Build GRCh38
rs890125318 Chr.2: 190369447-190369447 on Build GRCh38
rs776497864 Chr.2: 190369248-190369248 on Build GRCh38
rs1020088597 Chr.2: 190369122-190369122 on Build GRCh38
rs764227271 Chr.2: 190369269-190369269 on Build GRCh38
rs780681608 Chr.2: 190369323-190369323 on Build GRCh38
rs773088923 Chr.2: 190369106-190369106 on Build GRCh38
rs759324610 Chr.2: 190369249-190369249 on Build GRCh38
rs747680282 Chr.2: 190369214-190369214 on Build GRCh38
rs1026142955 Chr.2: 190369183-190369183 on Build GRCh38
rs759575263 Chr.2: 190369148-190369148 on Build GRCh38
rs1053247260 Chr.2: 190369355-190369355 on Build GRCh38
rs776426679 Chr.2: 190369137-190369137 on Build GRCh38
rs762111295 Chr.2: 190369289-190369289 on Build GRCh38
rs537236281 Chr.2: 190369018-190369018 on Build GRCh38
rs750835778 Chr.2: 190369309-190369309 on Build GRCh38
rs758567676 Chr.2: 190369164-190369164 on Build GRCh38
rs948500000 Chr.2: 190369430-190369430 on Build GRCh38
rs756652194 Chr.2: 190369202-190369202 on Build GRCh38
rs926248482 Chr.2: 190369345-190369345 on Build GRCh38
rs767710798 Chr.2: 190369296-190369296 on Build GRCh38
rs749936358 Chr.2: 190369274-190369274 on Build GRCh38

View Full Product Details