Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197488943-197489443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPD1
Entrez Gene ID
3329
Gene Name
heat shock protein family D (Hsp60) member 1
Gene Aliases
CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13
UniGene
Hs.595053
Gene Chromosome Location
Chr.2: 197486584-197500274 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3909305 c.1127A>G p.E376G Chr.2: 197489090-197489090 on Build GRCh38
6484598 c.1177C>A p.R393R Chr.2: 197489040-197489040 on Build GRCh38
3909304 c.1208T>G p.V403G Chr.2: 197489009-197489009 on Build GRCh38
442072 c.1084G>T p.G362C Chr.2: 197489133-197489133 on Build GRCh38
6682709 c.1208T>C p.V403A Chr.2: 197489009-197489009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200655333 Chr.2: 197489224-197489224 on Build GRCh38
rs938920949 Chr.2: 197489372-197489372 on Build GRCh38
rs199877481 Chr.2: 197489291-197489291 on Build GRCh38
rs950458504 Chr.2: 197489266-197489266 on Build GRCh38
rs755920309 Chr.2: 197489208-197489208 on Build GRCh38
rs61736612 Chr.2: 197489077-197489077 on Build GRCh38
rs768557320 Chr.2: 197489109-197489109 on Build GRCh38
rs763822262 Chr.2: 197489028-197489028 on Build GRCh38
rs778839028 Chr.2: 197489234-197489234 on Build GRCh38
rs751104797 Chr.2: 197489039-197489039 on Build GRCh38
rs781119204 Chr.2: 197489062-197489062 on Build GRCh38
rs758652542 Chr.2: 197489245-197489245 on Build GRCh38
rs564748273 Chr.2: 197489108-197489108 on Build GRCh38
rs762528177 Chr.2: 197489164-197489164 on Build GRCh38
rs767458046 Chr.2: 197489203-197489203 on Build GRCh38
rs760990707 Chr.2: 197489148-197489148 on Build GRCh38
rs745990880 Chr.2: 197489284-197489285 on Build GRCh38
rs756138484 Chr.2: 197489186-197489196 on Build GRCh38
rs1020108381 Chr.2: 197489060-197489060 on Build GRCh38
rs61751219 Chr.2: 197489077-197489077 on Build GRCh38
rs764997042 Chr.2: 197489149-197489149 on Build GRCh38
rs750272634 Chr.2: 197489204-197489204 on Build GRCh38
rs1038458497 Chr.2: 197489315-197489315 on Build GRCh38
rs201460108 Chr.2: 197489011-197489011 on Build GRCh38
rs747387316 Chr.2: 197489255-197489255 on Build GRCh38
rs886055386 Chr.2: 197489010-197489010 on Build GRCh38
rs752591564 Chr.2: 197489049-197489049 on Build GRCh38
rs750434057 Chr.2: 197489069-197489069 on Build GRCh38
rs918785953 Chr.2: 197489342-197489342 on Build GRCh38
rs868828254 Chr.2: 197489218-197489218 on Build GRCh38
rs758599676 Chr.2: 197489296-197489296 on Build GRCh38
rs980676314 Chr.2: 197489355-197489355 on Build GRCh38
rs751207405 Chr.2: 197489418-197489418 on Build GRCh38
rs777261563 Chr.2: 197489277-197489277 on Build GRCh38
rs182333371 Chr.2: 197489265-197489265 on Build GRCh38
rs771451010 Chr.2: 197489151-197489151 on Build GRCh38
rs762320773 Chr.2: 197488969-197488969 on Build GRCh38
rs1045994497 Chr.2: 197489348-197489348 on Build GRCh38
rs763507243 Chr.2: 197489171-197489171 on Build GRCh38
rs531377590 Chr.2: 197489414-197489414 on Build GRCh38
rs773536520 Chr.2: 197489146-197489146 on Build GRCh38
rs776946007 Chr.2: 197489158-197489158 on Build GRCh38
rs779265741 Chr.2: 197489110-197489110 on Build GRCh38
rs773701264 Chr.2: 197489293-197489293 on Build GRCh38
rs746347682 Chr.2: 197489280-197489280 on Build GRCh38
rs761534155 Chr.2: 197489195-197489195 on Build GRCh38
rs755147825 Chr.2: 197489227-197489227 on Build GRCh38
rs67523958 Chr.2: 197489081-197489081 on Build GRCh38
rs187751424 Chr.2: 197489313-197489313 on Build GRCh38
rs778859402 Chr.2: 197489252-197489253 on Build GRCh38
rs370073151 Chr.2: 197489240-197489240 on Build GRCh38
rs562559221 Chr.2: 197489413-197489413 on Build GRCh38
rs780050389 Chr.2: 197489209-197489209 on Build GRCh38
rs529369453 Chr.2: 197489371-197489371 on Build GRCh38
rs549419365 Chr.2: 197489412-197489412 on Build GRCh38
rs751281922 Chr.2: 197489176-197489176 on Build GRCh38
rs947476885 Chr.2: 197489415-197489415 on Build GRCh38
rs758160937 Chr.2: 197489323-197489323 on Build GRCh38
rs948902471 Chr.2: 197489347-197489347 on Build GRCh38
rs772478050 Chr.2: 197489128-197489128 on Build GRCh38
rs757141779 Chr.2: 197489042-197489042 on Build GRCh38
rs930121822 Chr.2: 197489343-197489343 on Build GRCh38
rs749499239 Chr.2: 197489253-197489253 on Build GRCh38
rs748307894 Chr.2: 197489117-197489117 on Build GRCh38

View Full Product Details