Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200490273-200490785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD18
Entrez Gene ID
130535
Gene Name
potassium channel tetramerization domain containing 18
Gene Aliases
6530404F10Rik
UniGene
Hs.605775
Gene Chromosome Location
Chr.2: 200488952-200510094 on Build GRCh38
Transcripts
NM_001321547.1 NM_001321548.1 NM_001321550.1 NM_152387.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4090087 c.926C>T p.A309V Chr.2: 200490455-200490455 on Build GRCh38
1683886 c.832delT p.L279fs*98 Chr.2: 200490549-200490549 on Build GRCh38
1404398 c.1037C>T p.S346F Chr.2: 200490344-200490344 on Build GRCh38
5525131 c.1000C>T p.L334L Chr.2: 200490381-200490381 on Build GRCh38
5518366 c.917C>T p.S306L Chr.2: 200490464-200490464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534708442 Chr.2: 200490625-200490625 on Build GRCh38
rs769903059 Chr.2: 200490626-200490626 on Build GRCh38
rs763517829 Chr.2: 200490580-200490580 on Build GRCh38
rs545679088 Chr.2: 200490424-200490424 on Build GRCh38
rs200898789 Chr.2: 200490604-200490604 on Build GRCh38
rs768275163 Chr.2: 200490447-200490447 on Build GRCh38
rs557619871 Chr.2: 200490738-200490738 on Build GRCh38
rs780719949 Chr.2: 200490494-200490494 on Build GRCh38
rs756549001 Chr.2: 200490321-200490321 on Build GRCh38
rs370785664 Chr.2: 200490592-200490592 on Build GRCh38
rs185813104 Chr.2: 200490444-200490444 on Build GRCh38
rs199722039 Chr.2: 200490386-200490386 on Build GRCh38
rs765680919 Chr.2: 200490575-200490575 on Build GRCh38
rs764954958 Chr.2: 200490301-200490301 on Build GRCh38
rs376505965 Chr.2: 200490646-200490646 on Build GRCh38
rs201870494 Chr.2: 200490425-200490425 on Build GRCh38
rs761799873 Chr.2: 200490452-200490452 on Build GRCh38
rs975489651 Chr.2: 200490464-200490464 on Build GRCh38
rs764570627 Chr.2: 200490585-200490585 on Build GRCh38
rs765454221 Chr.2: 200490487-200490487 on Build GRCh38
rs200653506 Chr.2: 200490331-200490331 on Build GRCh38
rs13018579 Chr.2: 200490383-200490383 on Build GRCh38
rs748821162 Chr.2: 200490509-200490509 on Build GRCh38
rs369656416 Chr.2: 200490505-200490505 on Build GRCh38
rs543025211 Chr.2: 200490455-200490455 on Build GRCh38
rs771459770 Chr.2: 200490650-200490650 on Build GRCh38
rs754829938 Chr.2: 200490520-200490520 on Build GRCh38
rs1002347034 Chr.2: 200490573-200490573 on Build GRCh38
rs1045274395 Chr.2: 200490480-200490480 on Build GRCh38
rs779779147 Chr.2: 200490437-200490437 on Build GRCh38
rs930942858 Chr.2: 200490717-200490717 on Build GRCh38
rs150419396 Chr.2: 200490335-200490335 on Build GRCh38
rs750702180 Chr.2: 200490385-200490385 on Build GRCh38
rs748148780 Chr.2: 200490526-200490526 on Build GRCh38
rs115629407 Chr.2: 200490593-200490593 on Build GRCh38
rs979079956 Chr.2: 200490429-200490429 on Build GRCh38
rs1004135506 Chr.2: 200490749-200490749 on Build GRCh38
rs774348783 Chr.2: 200490382-200490382 on Build GRCh38
rs776526393 Chr.2: 200490616-200490616 on Build GRCh38
rs201678106 Chr.2: 200490535-200490535 on Build GRCh38
rs931785255 Chr.2: 200490402-200490402 on Build GRCh38
rs779463028 Chr.2: 200490408-200490408 on Build GRCh38
rs758879282 Chr.2: 200490493-200490493 on Build GRCh38
rs200761596 Chr.2: 200490466-200490466 on Build GRCh38
rs536670939 Chr.2: 200490327-200490327 on Build GRCh38
rs200312807 Chr.2: 200490633-200490633 on Build GRCh38
rs866190955 Chr.2: 200490496-200490496 on Build GRCh38
rs964181885 Chr.2: 200490461-200490461 on Build GRCh38
rs766902417 Chr.2: 200490397-200490397 on Build GRCh38
rs370941047 Chr.2: 200490506-200490506 on Build GRCh38
rs374611164 Chr.2: 200490615-200490615 on Build GRCh38
rs368692549 Chr.2: 200490632-200490632 on Build GRCh38
rs749913269 Chr.2: 200490406-200490411 on Build GRCh38
rs775798899 Chr.2: 200490577-200490577 on Build GRCh38
rs908548372 Chr.2: 200490725-200490725 on Build GRCh38
rs149091190 Chr.2: 200490492-200490492 on Build GRCh38
rs373381202 Chr.2: 200490370-200490370 on Build GRCh38
rs761369148 Chr.2: 200490426-200490426 on Build GRCh38
rs1009532746 Chr.2: 200490549-200490549 on Build GRCh38
rs578090485 Chr.2: 200490654-200490654 on Build GRCh38
rs569262259 Chr.2: 200490310-200490310 on Build GRCh38
rs371332768 Chr.2: 200490297-200490297 on Build GRCh38
rs988080269 Chr.2: 200490358-200490358 on Build GRCh38
rs776918225 Chr.2: 200490553-200490553 on Build GRCh38
rs370973122 Chr.2: 200490607-200490607 on Build GRCh38
rs569631375 Chr.2: 200490567-200490567 on Build GRCh38
rs749784775 Chr.2: 200490328-200490328 on Build GRCh38
rs147353210 Chr.2: 200490416-200490416 on Build GRCh38
rs770858936 Chr.2: 200490542-200490542 on Build GRCh38
rs899422813 Chr.2: 200490463-200490463 on Build GRCh38
rs368024909 Chr.2: 200490652-200490652 on Build GRCh38
rs777866408 Chr.2: 200490605-200490605 on Build GRCh38
rs752339754 Chr.2: 200490657-200490657 on Build GRCh38
rs766622885 Chr.2: 200490469-200490469 on Build GRCh38
rs747956684 Chr.2: 200490599-200490599 on Build GRCh38
rs200202417 Chr.2: 200490482-200490482 on Build GRCh38
rs144995654 Chr.2: 200490296-200490296 on Build GRCh38
rs775436756 Chr.2: 200490302-200490302 on Build GRCh38
rs982820866 Chr.2: 200490675-200490675 on Build GRCh38
rs922422319 Chr.2: 200490634-200490634 on Build GRCh38
rs746524614 Chr.2: 200490337-200490337 on Build GRCh38
rs10203154 Chr.2: 200490375-200490375 on Build GRCh38
rs778520508 Chr.2: 200490524-200490524 on Build GRCh38
rs372891645 Chr.2: 200490534-200490534 on Build GRCh38
rs767635791 Chr.2: 200490384-200490384 on Build GRCh38
rs760876279 Chr.2: 200490435-200490435 on Build GRCh38
rs539248170 Chr.2: 200490668-200490668 on Build GRCh38
rs754204639 Chr.2: 200490407-200490407 on Build GRCh38
rs757911192 Chr.2: 200490426-200490426 on Build GRCh38
rs760932856 Chr.2: 200490393-200490393 on Build GRCh38
rs181273720 Chr.2: 200490589-200490589 on Build GRCh38
rs774866286 Chr.2: 200490645-200490645 on Build GRCh38
rs528581353 Chr.2: 200490475-200490475 on Build GRCh38
rs762837663 Chr.2: 200490371-200490371 on Build GRCh38
rs780836873 Chr.2: 200490365-200490365 on Build GRCh38
rs376356569 Chr.2: 200490588-200490588 on Build GRCh38
rs745459410 Chr.2: 200490366-200490366 on Build GRCh38
rs770629295 Chr.2: 200490352-200490352 on Build GRCh38
rs745567181 Chr.2: 200490431-200490431 on Build GRCh38
rs774347784 Chr.2: 200490450-200490450 on Build GRCh38
rs374734919 Chr.2: 200490303-200490303 on Build GRCh38
rs188668776 Chr.2: 200490541-200490541 on Build GRCh38
rs765961264 Chr.2: 200490648-200490648 on Build GRCh38
rs551272636 Chr.2: 200490309-200490309 on Build GRCh38

View Full Product Details