Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200493094-200493546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD18
Entrez Gene ID
130535
Gene Name
potassium channel tetramerization domain containing 18
Gene Aliases
6530404F10Rik
UniGene
Hs.605775
Gene Chromosome Location
Chr.2: 200488952-200510094 on Build GRCh38
Transcripts
NM_001321547.1 NM_001321548.1 NM_001321550.1 NM_152387.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4090088 c.760A>G p.K254E Chr.2: 200493176-200493176 on Build GRCh38
1752238 c.751C>G p.P251A Chr.2: 200493185-200493185 on Build GRCh38
6256513 c.669C>A p.S223R Chr.2: 200493267-200493267 on Build GRCh38
1211686 c.740G>A p.R247H Chr.2: 200493196-200493196 on Build GRCh38
6457101 c.753A>G p.P251P Chr.2: 200493183-200493183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762424043 Chr.2: 200493190-200493190 on Build GRCh38
rs894637760 Chr.2: 200493429-200493429 on Build GRCh38
rs758982580 Chr.2: 200493197-200493197 on Build GRCh38
rs903249840 Chr.2: 200493451-200493451 on Build GRCh38
rs755252564 Chr.2: 200493311-200493311 on Build GRCh38
rs999271833 Chr.2: 200493484-200493484 on Build GRCh38
rs776263342 Chr.2: 200493196-200493196 on Build GRCh38
rs764031587 Chr.2: 200493216-200493216 on Build GRCh38
rs116004805 Chr.2: 200493379-200493379 on Build GRCh38
rs377566618 Chr.2: 200493218-200493218 on Build GRCh38
rs541515686 Chr.2: 200493399-200493399 on Build GRCh38
rs202096745 Chr.2: 200493260-200493260 on Build GRCh38
rs761208197 Chr.2: 200493257-200493257 on Build GRCh38
rs1055931630 Chr.2: 200493219-200493219 on Build GRCh38
rs373524771 Chr.2: 200493183-200493183 on Build GRCh38
rs768832472 Chr.2: 200493244-200493244 on Build GRCh38
rs748449097 Chr.2: 200493252-200493252 on Build GRCh38
rs545973995 Chr.2: 200493459-200493459 on Build GRCh38
rs111302441 Chr.2: 200493170-200493170 on Build GRCh38
rs781001321 Chr.2: 200493225-200493225 on Build GRCh38
rs78882630 Chr.2: 200493381-200493381 on Build GRCh38
rs906217533 Chr.2: 200493480-200493480 on Build GRCh38
rs769455001 Chr.2: 200493259-200493259 on Build GRCh38
rs753912975 Chr.2: 200493305-200493305 on Build GRCh38
rs574384773 Chr.2: 200493397-200493397 on Build GRCh38
rs749471099 Chr.2: 200493182-200493182 on Build GRCh38
rs1003590808 Chr.2: 200493487-200493487 on Build GRCh38
rs564370388 Chr.2: 200493461-200493461 on Build GRCh38
rs947791628 Chr.2: 200493433-200493433 on Build GRCh38
rs756014650 Chr.2: 200493148-200493148 on Build GRCh38
rs752431948 Chr.2: 200493203-200493203 on Build GRCh38
rs780260009 Chr.2: 200493179-200493179 on Build GRCh38
rs1049295882 Chr.2: 200493418-200493418 on Build GRCh38
rs777672131 Chr.2: 200493122-200493122 on Build GRCh38
rs780934812 Chr.2: 200493210-200493211 on Build GRCh38
rs780665159 Chr.2: 200493130-200493130 on Build GRCh38
rs754693447 Chr.2: 200493152-200493154 on Build GRCh38
rs745919041 Chr.2: 200493168-200493168 on Build GRCh38
rs374865907 Chr.2: 200493166-200493166 on Build GRCh38
rs779017211 Chr.2: 200493315-200493315 on Build GRCh38
rs757274344 Chr.2: 200493220-200493220 on Build GRCh38
rs770738702 Chr.2: 200493328-200493328 on Build GRCh38
rs750303948 Chr.2: 200493226-200493226 on Build GRCh38
rs779402868 Chr.2: 200493250-200493250 on Build GRCh38
rs982439739 Chr.2: 200493405-200493405 on Build GRCh38
rs1003464438 Chr.2: 200493234-200493234 on Build GRCh38
rs929646240 Chr.2: 200493400-200493400 on Build GRCh38
rs779627463 Chr.2: 200493134-200493134 on Build GRCh38
rs185888456 Chr.2: 200493410-200493410 on Build GRCh38
rs527849244 Chr.2: 200493426-200493426 on Build GRCh38
rs989302206 Chr.2: 200493133-200493134 on Build GRCh38
rs372255855 Chr.2: 200493291-200493291 on Build GRCh38
rs751163160 Chr.2: 200493263-200493263 on Build GRCh38
rs1045814890 Chr.2: 200493446-200493446 on Build GRCh38
rs772424911 Chr.2: 200493254-200493254 on Build GRCh38
rs761513336 Chr.2: 200493265-200493265 on Build GRCh38
rs762649444 Chr.2: 200493211-200493211 on Build GRCh38
rs953498285 Chr.2: 200493371-200493371 on Build GRCh38
rs879660384 Chr.2: 200493501-200493501 on Build GRCh38
rs749559061 Chr.2: 200493242-200493242 on Build GRCh38
rs769849868 Chr.2: 200493176-200493176 on Build GRCh38
rs751201739 Chr.2: 200493151-200493154 on Build GRCh38
rs963822650 Chr.2: 200493338-200493338 on Build GRCh38
rs181056732 Chr.2: 200493129-200493129 on Build GRCh38
rs772454771 Chr.2: 200493193-200493193 on Build GRCh38
rs758725632 Chr.2: 200493316-200493316 on Build GRCh38
rs146790839 Chr.2: 200493185-200493185 on Build GRCh38
rs374863949 Chr.2: 200493299-200493299 on Build GRCh38
rs750349397 Chr.2: 200493294-200493294 on Build GRCh38
rs112932012 Chr.2: 200493256-200493256 on Build GRCh38
rs765668334 Chr.2: 200493210-200493210 on Build GRCh38
rs888680298 Chr.2: 200493215-200493215 on Build GRCh38
rs950162588 Chr.2: 200493443-200493443 on Build GRCh38
rs986145309 Chr.2: 200493378-200493378 on Build GRCh38
rs780059272 Chr.2: 200493241-200493241 on Build GRCh38
rs764777566 Chr.2: 200493198-200493198 on Build GRCh38
rs375870938 Chr.2: 200493425-200493425 on Build GRCh38
rs919533069 Chr.2: 200493407-200493407 on Build GRCh38
rs756217145 Chr.2: 200493239-200493239 on Build GRCh38
rs1022083573 Chr.2: 200493357-200493357 on Build GRCh38
rs866354560 Chr.2: 200493237-200493238 on Build GRCh38
rs866353306 Chr.2: 200493177-200493177 on Build GRCh38
rs752678498 Chr.2: 200493143-200493143 on Build GRCh38

View Full Product Details