Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 205995897-205996408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INO80D
Entrez Gene ID
54891
Gene Name
INO80 complex subunit D
Gene Aliases
-
UniGene
Hs.445036
Gene Chromosome Location
Chr.2: 205993721-206086182 on Build GRCh38
Transcripts
NM_017759.4 XM_011511369.2 XM_011511375.2 XM_011511376.2

SNP Detail

SNP ID Chromosome Location
rs927723683 Chr.2: 205995997-205995997 on Build GRCh38
rs182643963 Chr.2: 205996235-205996235 on Build GRCh38
rs981762570 Chr.2: 205995990-205995990 on Build GRCh38
rs887353834 Chr.2: 205996101-205996101 on Build GRCh38
rs937764226 Chr.2: 205996000-205996000 on Build GRCh38
rs972519260 Chr.2: 205996300-205996300 on Build GRCh38
rs71035431 Chr.2: 205996342-205996343 on Build GRCh38
rs934013876 Chr.2: 205995928-205995928 on Build GRCh38
rs893310780 Chr.2: 205996080-205996080 on Build GRCh38
rs1004192549 Chr.2: 205996110-205996110 on Build GRCh38
rs77340848 Chr.2: 205996327-205996327 on Build GRCh38
rs528539485 Chr.2: 205996330-205996331 on Build GRCh38
rs150476250 Chr.2: 205996207-205996207 on Build GRCh38
rs771176508 Chr.2: 205996093-205996093 on Build GRCh38
rs544361879 Chr.2: 205996346-205996346 on Build GRCh38
rs1049153836 Chr.2: 205996089-205996089 on Build GRCh38
rs933885714 Chr.2: 205996304-205996304 on Build GRCh38
rs575435536 Chr.2: 205996271-205996271 on Build GRCh38
rs962532256 Chr.2: 205996280-205996280 on Build GRCh38
rs958985357 Chr.2: 205996311-205996311 on Build GRCh38
rs918530108 Chr.2: 205996304-205996304 on Build GRCh38
rs996895703 Chr.2: 205996170-205996170 on Build GRCh38
rs544782590 Chr.2: 205996026-205996026 on Build GRCh38
rs1034699278 Chr.2: 205996243-205996243 on Build GRCh38
rs575868021 Chr.2: 205996327-205996330 on Build GRCh38
rs901151544 Chr.2: 205996119-205996119 on Build GRCh38
rs570993271 Chr.2: 205996327-205996328 on Build GRCh38
rs889904262 Chr.2: 205995958-205995958 on Build GRCh38
rs913817357 Chr.2: 205995969-205995969 on Build GRCh38
rs959192784 Chr.2: 205996205-205996205 on Build GRCh38
rs79758042 Chr.2: 205996326-205996326 on Build GRCh38
rs1003240070 Chr.2: 205996182-205996182 on Build GRCh38
rs911209759 Chr.2: 205996375-205996375 on Build GRCh38
rs1010732403 Chr.2: 205996193-205996193 on Build GRCh38
rs796658457 Chr.2: 205995933-205995933 on Build GRCh38
rs1010374481 Chr.2: 205996098-205996098 on Build GRCh38
rs74994696 Chr.2: 205996105-205996105 on Build GRCh38
rs1041040608 Chr.2: 205996114-205996114 on Build GRCh38
rs931493865 Chr.2: 205996041-205996041 on Build GRCh38
rs558855378 Chr.2: 205996380-205996380 on Build GRCh38
rs966814466 Chr.2: 205995971-205995971 on Build GRCh38
rs71410851 Chr.2: 205996326-205996327 on Build GRCh38
rs879219986 Chr.2: 205996327-205996327 on Build GRCh38
rs1054845690 Chr.2: 205996015-205996015 on Build GRCh38
rs1028369712 Chr.2: 205996172-205996172 on Build GRCh38
rs916065651 Chr.2: 205996036-205996036 on Build GRCh38
rs1021321003 Chr.2: 205996200-205996200 on Build GRCh38
rs751041742 Chr.2: 205996189-205996189 on Build GRCh38

View Full Product Details