Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 205996301-205996808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INO80D
Entrez Gene ID
54891
Gene Name
INO80 complex subunit D
Gene Aliases
-
UniGene
Hs.445036
Gene Chromosome Location
Chr.2: 205993721-206086182 on Build GRCh38
Transcripts
NM_017759.4 XM_011511369.2 XM_011511375.2 XM_011511376.2

SNP Detail

SNP ID Chromosome Location
rs1050303814 Chr.2: 205996590-205996590 on Build GRCh38
rs187666318 Chr.2: 205996512-205996512 on Build GRCh38
rs77340848 Chr.2: 205996327-205996327 on Build GRCh38
rs946214739 Chr.2: 205996463-205996463 on Build GRCh38
rs528539485 Chr.2: 205996330-205996331 on Build GRCh38
rs544361879 Chr.2: 205996346-205996346 on Build GRCh38
rs777485337 Chr.2: 205996620-205996620 on Build GRCh38
rs372853822 Chr.2: 205996434-205996434 on Build GRCh38
rs1056012730 Chr.2: 205996549-205996551 on Build GRCh38
rs570993271 Chr.2: 205996327-205996328 on Build GRCh38
rs79758042 Chr.2: 205996326-205996326 on Build GRCh38
rs763410505 Chr.2: 205996656-205996659 on Build GRCh38
rs1016659647 Chr.2: 205996612-205996612 on Build GRCh38
rs911209759 Chr.2: 205996375-205996375 on Build GRCh38
rs1020806322 Chr.2: 205996617-205996617 on Build GRCh38
rs558855378 Chr.2: 205996380-205996380 on Build GRCh38
rs1011182181 Chr.2: 205996611-205996611 on Build GRCh38
rs532331973 Chr.2: 205996648-205996648 on Build GRCh38
rs71410851 Chr.2: 205996326-205996327 on Build GRCh38
rs879219986 Chr.2: 205996327-205996327 on Build GRCh38
rs1003462043 Chr.2: 205996641-205996641 on Build GRCh38
rs560104771 Chr.2: 205996609-205996609 on Build GRCh38
rs867694722 Chr.2: 205996446-205996446 on Build GRCh38
rs529980597 Chr.2: 205996489-205996489 on Build GRCh38
rs920347165 Chr.2: 205996438-205996438 on Build GRCh38
rs71035431 Chr.2: 205996342-205996343 on Build GRCh38
rs1041248578 Chr.2: 205996544-205996544 on Build GRCh38
rs368852150 Chr.2: 205996558-205996558 on Build GRCh38
rs143871658 Chr.2: 205996585-205996586 on Build GRCh38
rs764455528 Chr.2: 205996766-205996766 on Build GRCh38
rs112697858 Chr.2: 205996662-205996662 on Build GRCh38
rs1027840778 Chr.2: 205996679-205996679 on Build GRCh38
rs757807952 Chr.2: 205996548-205996548 on Build GRCh38
rs990484807 Chr.2: 205996672-205996672 on Build GRCh38
rs575868021 Chr.2: 205996327-205996330 on Build GRCh38
rs752941102 Chr.2: 205996629-205996629 on Build GRCh38
rs938831538 Chr.2: 205996782-205996782 on Build GRCh38
rs1041948403 Chr.2: 205996769-205996769 on Build GRCh38
rs987123906 Chr.2: 205996651-205996654 on Build GRCh38
rs79911593 Chr.2: 205996647-205996647 on Build GRCh38
rs1041895977 Chr.2: 205996486-205996486 on Build GRCh38
rs908722522 Chr.2: 205996776-205996776 on Build GRCh38
rs908679888 Chr.2: 205996523-205996523 on Build GRCh38
rs747717799 Chr.2: 205996543-205996543 on Build GRCh38
rs570452503 Chr.2: 205996506-205996506 on Build GRCh38
rs911106000 Chr.2: 205996653-205996653 on Build GRCh38
rs932652148 Chr.2: 205996551-205996551 on Build GRCh38
rs532655678 Chr.2: 205996443-205996447 on Build GRCh38
rs962420139 Chr.2: 205996627-205996627 on Build GRCh38
rs951848063 Chr.2: 205996685-205996685 on Build GRCh38

View Full Product Details