Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206001661-206002172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INO80D
Entrez Gene ID
54891
Gene Name
INO80 complex subunit D
Gene Aliases
-
UniGene
Hs.445036
Gene Chromosome Location
Chr.2: 205993721-206086182 on Build GRCh38
Transcripts
NM_017759.4 XM_011511369.2 XM_011511375.2 XM_011511376.2

SNP Detail

SNP ID Chromosome Location
rs971907832 Chr.2: 206001944-206001944 on Build GRCh38
rs927126704 Chr.2: 206001953-206001953 on Build GRCh38
rs912204880 Chr.2: 206002093-206002093 on Build GRCh38
rs755283714 Chr.2: 206001698-206001698 on Build GRCh38
rs559948932 Chr.2: 206001696-206001696 on Build GRCh38
rs1046820703 Chr.2: 206001726-206001726 on Build GRCh38
rs1019547110 Chr.2: 206001862-206001862 on Build GRCh38
rs768024185 Chr.2: 206002027-206002027 on Build GRCh38
rs996868632 Chr.2: 206001867-206001867 on Build GRCh38
rs1039449202 Chr.2: 206002120-206002120 on Build GRCh38
rs546974412 Chr.2: 206001952-206001952 on Build GRCh38
rs535592730 Chr.2: 206002109-206002109 on Build GRCh38
rs1036483306 Chr.2: 206001825-206001825 on Build GRCh38
rs375410055 Chr.2: 206001757-206001757 on Build GRCh38
rs755696304 Chr.2: 206001818-206001818 on Build GRCh38
rs1020268984 Chr.2: 206001934-206001934 on Build GRCh38
rs549140304 Chr.2: 206002066-206002066 on Build GRCh38
rs1033262024 Chr.2: 206001905-206001905 on Build GRCh38
rs1010229961 Chr.2: 206001921-206001921 on Build GRCh38
rs187097268 Chr.2: 206002003-206002003 on Build GRCh38
rs900637389 Chr.2: 206001866-206001866 on Build GRCh38
rs532861012 Chr.2: 206002038-206002038 on Build GRCh38
rs1007947352 Chr.2: 206001785-206001785 on Build GRCh38
rs981510692 Chr.2: 206001950-206001950 on Build GRCh38
rs1048702476 Chr.2: 206001705-206001705 on Build GRCh38
rs890861715 Chr.2: 206001781-206001781 on Build GRCh38
rs766248869 Chr.2: 206001977-206001977 on Build GRCh38
rs973999220 Chr.2: 206002047-206002047 on Build GRCh38
rs887558669 Chr.2: 206001780-206001780 on Build GRCh38
rs373560599 Chr.2: 206002139-206002139 on Build GRCh38
rs528776736 Chr.2: 206001878-206001878 on Build GRCh38
rs943786591 Chr.2: 206002119-206002119 on Build GRCh38
rs957383256 Chr.2: 206001913-206001913 on Build GRCh38
rs148972756 Chr.2: 206002121-206002121 on Build GRCh38
rs4675582 Chr.2: 206002056-206002056 on Build GRCh38
rs1000901964 Chr.2: 206002057-206002057 on Build GRCh38
rs901685581 Chr.2: 206001830-206001830 on Build GRCh38
rs753598047 Chr.2: 206002029-206002029 on Build GRCh38
rs956880129 Chr.2: 206002099-206002099 on Build GRCh38
rs919824209 Chr.2: 206002051-206002051 on Build GRCh38

View Full Product Details