Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207766588-207767079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD5
Entrez Gene ID
7855
Gene Name
frizzled class receptor 5
Gene Aliases
C2orf31, HFZ5
UniGene
Hs.17631
Gene Chromosome Location
Chr.2: 207762586-207769419 on Build GRCh38
Transcripts
NM_003468.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3576903 c.1748C>T p.S583L Chr.2: 207766992-207766992 on Build GRCh38
5522734 c.1710G>A p.P570P Chr.2: 207767030-207767030 on Build GRCh38
3768925 c.1670C>T p.P557L Chr.2: 207767070-207767070 on Build GRCh38
6448206 c.1753G>A p.V585M Chr.2: 207766987-207766987 on Build GRCh38
6895330 c.1716C>A p.P572P Chr.2: 207767024-207767024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920567138 Chr.2: 207766953-207766953 on Build GRCh38
rs757877030 Chr.2: 207767018-207767018 on Build GRCh38
rs893570016 Chr.2: 207766737-207766737 on Build GRCh38
rs770398696 Chr.2: 207766967-207766967 on Build GRCh38
rs970905061 Chr.2: 207766803-207766803 on Build GRCh38
rs1004234744 Chr.2: 207766841-207766841 on Build GRCh38
rs953186356 Chr.2: 207766865-207766865 on Build GRCh38
rs781487290 Chr.2: 207766943-207766943 on Build GRCh38
rs539119927 Chr.2: 207766750-207766750 on Build GRCh38
rs778520376 Chr.2: 207766956-207766956 on Build GRCh38
rs938909008 Chr.2: 207767017-207767017 on Build GRCh38
rs756247200 Chr.2: 207767029-207767029 on Build GRCh38
rs754326568 Chr.2: 207767027-207767027 on Build GRCh38
rs555846140 Chr.2: 207766778-207766778 on Build GRCh38
rs989209733 Chr.2: 207766961-207766961 on Build GRCh38
rs192374431 Chr.2: 207766638-207766638 on Build GRCh38
rs757398217 Chr.2: 207766942-207766942 on Build GRCh38
rs981664137 Chr.2: 207767001-207767001 on Build GRCh38
rs763388244 Chr.2: 207766992-207766992 on Build GRCh38
rs749535241 Chr.2: 207767034-207767034 on Build GRCh38
rs576449228 Chr.2: 207766678-207766678 on Build GRCh38
rs752937292 Chr.2: 207766988-207766988 on Build GRCh38
rs901743327 Chr.2: 207766811-207766811 on Build GRCh38
rs541760352 Chr.2: 207766871-207766871 on Build GRCh38
rs1009247882 Chr.2: 207766748-207766748 on Build GRCh38
rs776168873 Chr.2: 207766970-207766970 on Build GRCh38
rs752106678 Chr.2: 207766999-207766999 on Build GRCh38
rs543895505 Chr.2: 207767036-207767036 on Build GRCh38
rs3739161 Chr.2: 207766628-207766628 on Build GRCh38
rs779488180 Chr.2: 207766814-207766815 on Build GRCh38
rs770081021 Chr.2: 207766972-207766980 on Build GRCh38
rs927423698 Chr.2: 207766662-207766662 on Build GRCh38
rs887538135 Chr.2: 207766740-207766740 on Build GRCh38
rs535280944 Chr.2: 207766945-207766945 on Build GRCh38
rs773125033 Chr.2: 207766966-207766966 on Build GRCh38
rs973411336 Chr.2: 207766952-207766952 on Build GRCh38
rs542453735 Chr.2: 207766820-207766820 on Build GRCh38
rs937508891 Chr.2: 207766670-207766670 on Build GRCh38
rs746929697 Chr.2: 207767046-207767046 on Build GRCh38
rs553386789 Chr.2: 207766672-207766672 on Build GRCh38
rs572731070 Chr.2: 207766842-207766842 on Build GRCh38
rs576549360 Chr.2: 207766807-207766807 on Build GRCh38
rs913470198 Chr.2: 207766630-207766630 on Build GRCh38
rs1036064740 Chr.2: 207766808-207766810 on Build GRCh38
rs751631220 Chr.2: 207766937-207766937 on Build GRCh38
rs746031009 Chr.2: 207766955-207766958 on Build GRCh38
rs13416271 Chr.2: 207767024-207767024 on Build GRCh38
rs764609939 Chr.2: 207766998-207766998 on Build GRCh38
rs771853726 Chr.2: 207767039-207767039 on Build GRCh38
rs971407718 Chr.2: 207766987-207766987 on Build GRCh38
rs1000660229 Chr.2: 207766878-207766878 on Build GRCh38
rs1021564569 Chr.2: 207766782-207766782 on Build GRCh38
rs13429572 Chr.2: 207766980-207766980 on Build GRCh38
rs555759953 Chr.2: 207766831-207766831 on Build GRCh38
rs761373854 Chr.2: 207766733-207766733 on Build GRCh38
rs369602029 Chr.2: 207767042-207767042 on Build GRCh38
rs908349969 Chr.2: 207766640-207766640 on Build GRCh38
rs748002513 Chr.2: 207766960-207766960 on Build GRCh38
rs770101168 Chr.2: 207766951-207766951 on Build GRCh38
rs996467441 Chr.2: 207766728-207766728 on Build GRCh38
rs766882565 Chr.2: 207766877-207766877 on Build GRCh38
rs780323223 Chr.2: 207767030-207767030 on Build GRCh38
rs1040955274 Chr.2: 207766654-207766654 on Build GRCh38
rs58365448 Chr.2: 207767060-207767060 on Build GRCh38
rs943977568 Chr.2: 207766642-207766642 on Build GRCh38
rs764973670 Chr.2: 207766986-207766986 on Build GRCh38

View Full Product Details