Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210020889-210021432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1L
Entrez Gene ID
151050
Gene Name
KAT8 regulatory NSL complex subunit 1 like
Gene Aliases
C2orf67, MSL1v2
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210021421-210171370 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 NM_001278282.1 NM_001278283.1 NM_001278285.1 NM_001278286.1 NM_001278288.1 NM_001278289.1 NM_001318926.1 NM_001318927.1 NM_001318928.1 NM_001318929.1 NM_001318930.1 NM_001318931.1 NM_006916.2 NM_199229.2 XM_006712677.3

Gene Symbol
RPE
Entrez Gene ID
6120
Gene Name
ribulose-5-phosphate-3-epimerase
Gene Aliases
RPE2-1
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210002565-210029156 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 NM_001278282.1 NM_001278283.1 NM_001278285.1 NM_001278286.1 NM_001278288.1 NM_001278289.1 NM_001318926.1 NM_001318927.1 NM_001318928.1 NM_001318929.1 NM_001318930.1 NM_001318931.1 NM_006916.2 NM_199229.2 XM_006712677.3

SNP Detail

SNP ID Chromosome Location
rs754706434 Chr.2: 210021396-210021396 on Build GRCh38
rs146335650 Chr.2: 210021388-210021388 on Build GRCh38
rs992537706 Chr.2: 210021067-210021067 on Build GRCh38
rs966816635 Chr.2: 210020977-210020977 on Build GRCh38
rs924271097 Chr.2: 210021039-210021041 on Build GRCh38
rs1008169097 Chr.2: 210020940-210020940 on Build GRCh38
rs763558436 Chr.2: 210021000-210021000 on Build GRCh38
rs756562320 Chr.2: 210021157-210021157 on Build GRCh38
rs969021974 Chr.2: 210021020-210021020 on Build GRCh38
rs200301166 Chr.2: 210021243-210021243 on Build GRCh38
rs181512592 Chr.2: 210020942-210020942 on Build GRCh38
rs996714779 Chr.2: 210021298-210021298 on Build GRCh38
rs919688840 Chr.2: 210021270-210021270 on Build GRCh38
rs1038265228 Chr.2: 210021138-210021138 on Build GRCh38
rs1008138004 Chr.2: 210021393-210021393 on Build GRCh38
rs577976227 Chr.2: 210021379-210021379 on Build GRCh38
rs939780754 Chr.2: 210021245-210021245 on Build GRCh38
rs948926467 Chr.2: 210021131-210021131 on Build GRCh38
rs532070422 Chr.2: 210021167-210021167 on Build GRCh38
rs754185300 Chr.2: 210021347-210021347 on Build GRCh38
rs779278964 Chr.2: 210021283-210021284 on Build GRCh38
rs1032433420 Chr.2: 210021017-210021017 on Build GRCh38
rs1017668641 Chr.2: 210020952-210020956 on Build GRCh38
rs190079017 Chr.2: 210021249-210021249 on Build GRCh38
rs756437486 Chr.2: 210021397-210021397 on Build GRCh38
rs929543317 Chr.2: 210021182-210021182 on Build GRCh38
rs185713154 Chr.2: 210021021-210021021 on Build GRCh38
rs963914824 Chr.2: 210020954-210020954 on Build GRCh38
rs968697568 Chr.2: 210021313-210021313 on Build GRCh38
rs532388996 Chr.2: 210020945-210020945 on Build GRCh38
rs757718772 Chr.2: 210021034-210021034 on Build GRCh38
rs932454196 Chr.2: 210021325-210021325 on Build GRCh38
rs751004939 Chr.2: 210021037-210021037 on Build GRCh38
rs780577477 Chr.2: 210021289-210021290 on Build GRCh38
rs915619135 Chr.2: 210021089-210021089 on Build GRCh38
rs890585743 Chr.2: 210020917-210020917 on Build GRCh38
rs547530286 Chr.2: 210021366-210021366 on Build GRCh38
rs1020934347 Chr.2: 210020951-210020951 on Build GRCh38
rs1049563861 Chr.2: 210021331-210021331 on Build GRCh38
rs781680088 Chr.2: 210021104-210021104 on Build GRCh38
rs991020161 Chr.2: 210021063-210021063 on Build GRCh38
rs370797277 Chr.2: 210021330-210021330 on Build GRCh38
rs1008979225 Chr.2: 210020950-210020950 on Build GRCh38
rs1017720847 Chr.2: 210021268-210021268 on Build GRCh38
rs908249849 Chr.2: 210021227-210021227 on Build GRCh38
rs981087485 Chr.2: 210021183-210021183 on Build GRCh38
rs1008864668 Chr.2: 210021263-210021263 on Build GRCh38
rs752026449 Chr.2: 210021030-210021030 on Build GRCh38
rs753830491 Chr.2: 210021048-210021048 on Build GRCh38
rs546049414 Chr.2: 210020965-210020965 on Build GRCh38
rs572259253 Chr.2: 210021225-210021225 on Build GRCh38
rs1021874046 Chr.2: 210021302-210021302 on Build GRCh38

View Full Product Details