Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 211378324-211378833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB4
Entrez Gene ID
2066
Gene Name
erb-b2 receptor tyrosine kinase 4
Gene Aliases
ALS19, HER4, p180erbB4
UniGene
Hs.390729
Gene Chromosome Location
Chr.2: 211375717-212539147 on Build GRCh38
Transcripts
NM_001042599.1 NM_005235.2 XM_005246376.2 XM_005246377.2 XM_006712364.2 XM_017003577.1 XM_017003578.1 XM_017003579.1 XM_017003580.1 XM_017003581.1 XM_017003582.1

SNP Detail

SNP ID Chromosome Location
rs561645469 Chr.2: 211378808-211378808 on Build GRCh38
rs906288107 Chr.2: 211378548-211378548 on Build GRCh38
rs1972820 Chr.2: 211378697-211378697 on Build GRCh38
rs989449995 Chr.2: 211378784-211378784 on Build GRCh38
rs1029657072 Chr.2: 211378641-211378641 on Build GRCh38
rs546283804 Chr.2: 211378350-211378350 on Build GRCh38
rs1054096238 Chr.2: 211378474-211378474 on Build GRCh38
rs901544441 Chr.2: 211378712-211378712 on Build GRCh38
rs956305602 Chr.2: 211378535-211378535 on Build GRCh38
rs187712206 Chr.2: 211378521-211378521 on Build GRCh38
rs940607126 Chr.2: 211378674-211378674 on Build GRCh38
rs146457608 Chr.2: 211378735-211378735 on Build GRCh38
rs1049009641 Chr.2: 211378368-211378368 on Build GRCh38
rs749561502 Chr.2: 211378662-211378662 on Build GRCh38
rs967455656 Chr.2: 211378673-211378673 on Build GRCh38
rs796877951 Chr.2: 211378610-211378610 on Build GRCh38
rs911970866 Chr.2: 211378428-211378428 on Build GRCh38
rs776042471 Chr.2: 211378400-211378400 on Build GRCh38
rs898797638 Chr.2: 211378614-211378614 on Build GRCh38
rs901568897 Chr.2: 211378471-211378471 on Build GRCh38
rs193099507 Chr.2: 211378525-211378525 on Build GRCh38
rs761497196 Chr.2: 211378617-211378617 on Build GRCh38
rs1004595150 Chr.2: 211378597-211378597 on Build GRCh38
rs568224152 Chr.2: 211378592-211378592 on Build GRCh38
rs1009029290 Chr.2: 211378659-211378659 on Build GRCh38
rs182536063 Chr.2: 211378461-211378461 on Build GRCh38
rs770149519 Chr.2: 211378388-211378388 on Build GRCh38
rs554850967 Chr.2: 211378542-211378542 on Build GRCh38
rs77670483 Chr.2: 211378747-211378747 on Build GRCh38
rs7569828 Chr.2: 211378809-211378809 on Build GRCh38
rs987555685 Chr.2: 211378415-211378415 on Build GRCh38
rs774211685 Chr.2: 211378746-211378746 on Build GRCh38
rs111290928 Chr.2: 211378528-211378528 on Build GRCh38
rs957580028 Chr.2: 211378773-211378773 on Build GRCh38
rs767200924 Chr.2: 211378626-211378626 on Build GRCh38
rs995963141 Chr.2: 211378489-211378489 on Build GRCh38
rs977396829 Chr.2: 211378455-211378455 on Build GRCh38
rs1029191071 Chr.2: 211378369-211378369 on Build GRCh38
rs946102249 Chr.2: 211378446-211378446 on Build GRCh38
rs12473268 Chr.2: 211378739-211378739 on Build GRCh38
rs1028903599 Chr.2: 211378494-211378494 on Build GRCh38
rs566995166 Chr.2: 211378387-211378387 on Build GRCh38
rs751168504 Chr.2: 211378546-211378546 on Build GRCh38
rs116395564 Chr.2: 211378666-211378666 on Build GRCh38
rs927115538 Chr.2: 211378644-211378644 on Build GRCh38
rs759795147 Chr.2: 211378803-211378803 on Build GRCh38
rs551671560 Chr.2: 211378706-211378706 on Build GRCh38
rs144018296 Chr.2: 211378581-211378581 on Build GRCh38
rs1014563551 Chr.2: 211378346-211378346 on Build GRCh38
rs58853206 Chr.2: 211378655-211378655 on Build GRCh38
rs576416411 Chr.2: 211378682-211378682 on Build GRCh38
rs114473068 Chr.2: 211378367-211378367 on Build GRCh38
rs936616374 Chr.2: 211378470-211378470 on Build GRCh38

View Full Product Details