Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 211382858-211383368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB4
Entrez Gene ID
2066
Gene Name
erb-b2 receptor tyrosine kinase 4
Gene Aliases
ALS19, HER4, p180erbB4
UniGene
Hs.390729
Gene Chromosome Location
Chr.2: 211375717-212539147 on Build GRCh38
Transcripts
NM_001042599.1 NM_005235.2 XM_005246376.2 XM_005246377.2 XM_006712364.2 XM_017003577.1 XM_017003578.1 XM_017003579.1 XM_017003580.1 XM_017003581.1 XM_017003582.1

SNP Detail

SNP ID Chromosome Location
rs958630732 Chr.2: 211382914-211382914 on Build GRCh38
rs920898764 Chr.2: 211383074-211383074 on Build GRCh38
rs150624055 Chr.2: 211382883-211382883 on Build GRCh38
rs72399367 Chr.2: 211382995-211382995 on Build GRCh38
rs530659148 Chr.2: 211383027-211383027 on Build GRCh38
rs1012224079 Chr.2: 211382946-211382946 on Build GRCh38
rs1047555351 Chr.2: 211383309-211383309 on Build GRCh38
rs1057078032 Chr.2: 211382910-211382910 on Build GRCh38
rs866695435 Chr.2: 211383298-211383298 on Build GRCh38
rs79652465 Chr.2: 211383259-211383259 on Build GRCh38
rs975033402 Chr.2: 211383050-211383051 on Build GRCh38
rs564996263 Chr.2: 211383010-211383010 on Build GRCh38
rs79310613 Chr.2: 211383135-211383135 on Build GRCh38
rs1044249856 Chr.2: 211383311-211383311 on Build GRCh38
rs191220247 Chr.2: 211382893-211382893 on Build GRCh38
rs187665528 Chr.2: 211383099-211383099 on Build GRCh38
rs1043857977 Chr.2: 211382888-211382888 on Build GRCh38
rs551332240 Chr.2: 211383189-211383189 on Build GRCh38
rs571527737 Chr.2: 211383231-211383231 on Build GRCh38
rs982971906 Chr.2: 211382953-211382953 on Build GRCh38
rs964014932 Chr.2: 211383042-211383042 on Build GRCh38
rs1033640772 Chr.2: 211382895-211382895 on Build GRCh38
rs1056648344 Chr.2: 211383258-211383258 on Build GRCh38
rs3748960 Chr.2: 211383109-211383109 on Build GRCh38
rs565989842 Chr.2: 211383121-211383121 on Build GRCh38
rs757784281 Chr.2: 211383046-211383046 on Build GRCh38
rs569229342 Chr.2: 211382947-211382947 on Build GRCh38
rs139701250 Chr.2: 211382976-211382976 on Build GRCh38
rs948307997 Chr.2: 211383283-211383288 on Build GRCh38
rs550390045 Chr.2: 211383085-211383085 on Build GRCh38
rs183072498 Chr.2: 211382940-211382940 on Build GRCh38
rs778244617 Chr.2: 211382932-211382932 on Build GRCh38
rs878944435 Chr.2: 211383243-211383243 on Build GRCh38
rs776241458 Chr.2: 211382891-211382891 on Build GRCh38
rs1055143596 Chr.2: 211383344-211383344 on Build GRCh38
rs945560040 Chr.2: 211383303-211383303 on Build GRCh38
rs1019465604 Chr.2: 211383342-211383342 on Build GRCh38
rs975066154 Chr.2: 211383063-211383063 on Build GRCh38
rs572887009 Chr.2: 211382973-211382973 on Build GRCh38
rs1002266824 Chr.2: 211383332-211383332 on Build GRCh38
rs750472663 Chr.2: 211383140-211383140 on Build GRCh38
rs903941996 Chr.2: 211383330-211383330 on Build GRCh38
rs935465590 Chr.2: 211383206-211383206 on Build GRCh38
rs1045074898 Chr.2: 211383297-211383298 on Build GRCh38
rs201883972 Chr.2: 211383049-211383049 on Build GRCh38
rs537585645 Chr.2: 211383334-211383334 on Build GRCh38
rs13405545 Chr.2: 211383143-211383143 on Build GRCh38
rs903891374 Chr.2: 211382886-211382886 on Build GRCh38
rs918140497 Chr.2: 211383278-211383278 on Build GRCh38
rs529464394 Chr.2: 211383103-211383103 on Build GRCh38
rs951250330 Chr.2: 211382979-211382979 on Build GRCh38

View Full Product Details