Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216673199-216673694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP5
Entrez Gene ID
3488
Gene Name
insulin like growth factor binding protein 5
Gene Aliases
IBP5
UniGene
Hs.607212
Gene Chromosome Location
Chr.2: 216672105-216695549 on Build GRCh38
Transcripts
NM_000599.3

SNP Detail

SNP ID Chromosome Location
rs576639827 Chr.2: 216673612-216673612 on Build GRCh38
rs574555808 Chr.2: 216673518-216673518 on Build GRCh38
rs113555497 Chr.2: 216673423-216673423 on Build GRCh38
rs543825082 Chr.2: 216673654-216673654 on Build GRCh38
rs932085068 Chr.2: 216673243-216673243 on Build GRCh38
rs956651985 Chr.2: 216673590-216673590 on Build GRCh38
rs969761612 Chr.2: 216673455-216673455 on Build GRCh38
rs570707973 Chr.2: 216673334-216673334 on Build GRCh38
rs760366055 Chr.2: 216673546-216673546 on Build GRCh38
rs747913696 Chr.2: 216673486-216673486 on Build GRCh38
rs375949024 Chr.2: 216673502-216673502 on Build GRCh38
rs73989582 Chr.2: 216673587-216673587 on Build GRCh38
rs139713554 Chr.2: 216673661-216673661 on Build GRCh38
rs201058817 Chr.2: 216673272-216673272 on Build GRCh38
rs866046995 Chr.2: 216673583-216673583 on Build GRCh38
rs1049582740 Chr.2: 216673628-216673628 on Build GRCh38
rs1015556461 Chr.2: 216673384-216673384 on Build GRCh38
rs565297676 Chr.2: 216673660-216673660 on Build GRCh38
rs553104194 Chr.2: 216673397-216673397 on Build GRCh38
rs113393667 Chr.2: 216673565-216673566 on Build GRCh38
rs977791540 Chr.2: 216673296-216673296 on Build GRCh38
rs73989581 Chr.2: 216673534-216673534 on Build GRCh38
rs1040273779 Chr.2: 216673230-216673230 on Build GRCh38
rs927667615 Chr.2: 216673498-216673498 on Build GRCh38
rs536644614 Chr.2: 216673323-216673323 on Build GRCh38
rs944484602 Chr.2: 216673229-216673229 on Build GRCh38
rs150870578 Chr.2: 216673327-216673327 on Build GRCh38
rs534627733 Chr.2: 216673387-216673387 on Build GRCh38
rs913162129 Chr.2: 216673222-216673222 on Build GRCh38
rs1023787988 Chr.2: 216673436-216673436 on Build GRCh38
rs900582755 Chr.2: 216673239-216673239 on Build GRCh38
rs956602289 Chr.2: 216673328-216673328 on Build GRCh38
rs113020203 Chr.2: 216673396-216673396 on Build GRCh38
rs776171460 Chr.2: 216673594-216673594 on Build GRCh38
rs569450453 Chr.2: 216673232-216673232 on Build GRCh38
rs980237566 Chr.2: 216673474-216673474 on Build GRCh38
rs1029754916 Chr.2: 216673569-216673569 on Build GRCh38
rs931850702 Chr.2: 216673617-216673617 on Build GRCh38
rs1004897011 Chr.2: 216673379-216673379 on Build GRCh38
rs540239276 Chr.2: 216673522-216673522 on Build GRCh38
rs774311788 Chr.2: 216673470-216673470 on Build GRCh38
rs887874207 Chr.2: 216673649-216673649 on Build GRCh38
rs1013703739 Chr.2: 216673420-216673420 on Build GRCh38
rs899996507 Chr.2: 216673547-216673547 on Build GRCh38

View Full Product Details