Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218262651-218263150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPBAR1
Entrez Gene ID
151306
Gene Name
G protein-coupled bile acid receptor 1
Gene Aliases
BG37, GPCR19, GPR131, M-BAR, TGR5
UniGene
Hs.160954
Gene Chromosome Location
Chr.2: 218259496-218263861 on Build GRCh38
Transcripts
NM_001077191.1 NM_001077194.1 NM_001321950.1 NM_170699.2 XM_011510743.1 XM_017003467.1 XM_017003468.1 XM_017003469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
476905 c.198G>T p.L66L Chr.2: 218262922-218262922 on Build GRCh38
476906 c.198G>T p.L66L Chr.2: 218262922-218262922 on Build GRCh38
7207362 c.56G>C p.G19A Chr.2: 218262780-218262780 on Build GRCh38
7207361 c.56G>C p.G19A Chr.2: 218262780-218262780 on Build GRCh38
4687744 c.220C>T p.L74L Chr.2: 218262944-218262944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565722792 Chr.2: 218262938-218262938 on Build GRCh38
rs749795150 Chr.2: 218263060-218263060 on Build GRCh38
rs201157758 Chr.2: 218263098-218263098 on Build GRCh38
rs200372604 Chr.2: 218262814-218262814 on Build GRCh38
rs920194853 Chr.2: 218262977-218262977 on Build GRCh38
rs201118648 Chr.2: 218262735-218262735 on Build GRCh38
rs374268179 Chr.2: 218263106-218263106 on Build GRCh38
rs201091893 Chr.2: 218263048-218263048 on Build GRCh38
rs759881264 Chr.2: 218262792-218262792 on Build GRCh38
rs200126718 Chr.2: 218262962-218262962 on Build GRCh38
rs907641202 Chr.2: 218262852-218262852 on Build GRCh38
rs778422127 Chr.2: 218262757-218262757 on Build GRCh38
rs750336003 Chr.2: 218262738-218262738 on Build GRCh38
rs201598384 Chr.2: 218262932-218262932 on Build GRCh38
rs778821198 Chr.2: 218262817-218262817 on Build GRCh38
rs762080489 Chr.2: 218263114-218263114 on Build GRCh38
rs775590022 Chr.2: 218262941-218262943 on Build GRCh38
rs140737899 Chr.2: 218262776-218262776 on Build GRCh38
rs146271918 Chr.2: 218262907-218262907 on Build GRCh38
rs202080111 Chr.2: 218262772-218262772 on Build GRCh38
rs200516871 Chr.2: 218263059-218263059 on Build GRCh38
rs199556363 Chr.2: 218263129-218263129 on Build GRCh38
rs981080494 Chr.2: 218262726-218262726 on Build GRCh38
rs199550995 Chr.2: 218263071-218263071 on Build GRCh38
rs199941566 Chr.2: 218263045-218263045 on Build GRCh38
rs766437738 Chr.2: 218263088-218263088 on Build GRCh38
rs202193441 Chr.2: 218262960-218262960 on Build GRCh38
rs752840888 Chr.2: 218262806-218262806 on Build GRCh38
rs770486496 Chr.2: 218262841-218262841 on Build GRCh38
rs772430608 Chr.2: 218263072-218263072 on Build GRCh38
rs958597089 Chr.2: 218262966-218262966 on Build GRCh38
rs749647848 Chr.2: 218262973-218262973 on Build GRCh38
rs370152013 Chr.2: 218262881-218262881 on Build GRCh38
rs759934564 Chr.2: 218262876-218262878 on Build GRCh38
rs771464145 Chr.2: 218262760-218262760 on Build GRCh38
rs370878053 Chr.2: 218263096-218263096 on Build GRCh38
rs202233050 Chr.2: 218262736-218262736 on Build GRCh38
rs201530699 Chr.2: 218262815-218262815 on Build GRCh38
rs750106315 Chr.2: 218263123-218263123 on Build GRCh38
rs749522380 Chr.2: 218262842-218262842 on Build GRCh38
rs749106499 Chr.2: 218262746-218262746 on Build GRCh38
rs747708687 Chr.2: 218262819-218262819 on Build GRCh38
rs200909291 Chr.2: 218262860-218262860 on Build GRCh38
rs974704702 Chr.2: 218262965-218262965 on Build GRCh38
rs765699564 Chr.2: 218262802-218262802 on Build GRCh38
rs773374681 Chr.2: 218263020-218263020 on Build GRCh38
rs199759333 Chr.2: 218262851-218262851 on Build GRCh38
rs1017094219 Chr.2: 218262888-218262888 on Build GRCh38
rs201459930 Chr.2: 218262840-218262840 on Build GRCh38
rs750098209 Chr.2: 218263099-218263107 on Build GRCh38
rs201272591 Chr.2: 218263062-218263062 on Build GRCh38
rs779995184 Chr.2: 218262855-218262855 on Build GRCh38
rs201145373 Chr.2: 218262745-218262745 on Build GRCh38
rs754969786 Chr.2: 218262920-218262920 on Build GRCh38
rs763662195 Chr.2: 218262861-218262861 on Build GRCh38
rs578194069 Chr.2: 218262682-218262682 on Build GRCh38
rs778529941 Chr.2: 218263006-218263008 on Build GRCh38
rs757932369 Chr.2: 218262941-218262941 on Build GRCh38
rs756760558 Chr.2: 218262979-218262981 on Build GRCh38
rs576819656 Chr.2: 218263046-218263046 on Build GRCh38
rs200624663 Chr.2: 218262928-218262928 on Build GRCh38
rs773111690 Chr.2: 218263073-218263073 on Build GRCh38
rs1039057957 Chr.2: 218263117-218263117 on Build GRCh38
rs758377744 Chr.2: 218262730-218262730 on Build GRCh38
rs756055296 Chr.2: 218262809-218262809 on Build GRCh38
rs754234858 Chr.2: 218262709-218262709 on Build GRCh38
rs200389207 Chr.2: 218262820-218262820 on Build GRCh38
rs777393013 Chr.2: 218262944-218262944 on Build GRCh38
rs759328571 Chr.2: 218263039-218263039 on Build GRCh38
rs181236250 Chr.2: 218262986-218262986 on Build GRCh38
rs561380828 Chr.2: 218262781-218262781 on Build GRCh38
rs753283468 Chr.2: 218262945-218262945 on Build GRCh38
rs772733931 Chr.2: 218262773-218262773 on Build GRCh38
rs1010249544 Chr.2: 218262941-218262944 on Build GRCh38
rs200010375 Chr.2: 218262890-218262890 on Build GRCh38
rs558257242 Chr.2: 218263007-218263007 on Build GRCh38
rs186702546 Chr.2: 218263053-218263053 on Build GRCh38
rs777199090 Chr.2: 218262837-218262837 on Build GRCh38
rs905721001 Chr.2: 218262893-218262893 on Build GRCh38
rs751885784 Chr.2: 218262882-218262882 on Build GRCh38
rs199688937 Chr.2: 218262916-218262916 on Build GRCh38
rs767660471 Chr.2: 218263017-218263017 on Build GRCh38
rs201627569 Chr.2: 218262987-218262987 on Build GRCh38
rs761046924 Chr.2: 218262932-218262933 on Build GRCh38
rs199817350 Chr.2: 218262737-218262737 on Build GRCh38
rs202032682 Chr.2: 218263058-218263058 on Build GRCh38
rs111925653 Chr.2: 218262913-218262913 on Build GRCh38
rs201129128 Chr.2: 218263057-218263057 on Build GRCh38
rs766683052 Chr.2: 218262701-218262701 on Build GRCh38
rs376686452 Chr.2: 218262935-218262935 on Build GRCh38
rs547700545 Chr.2: 218262915-218262915 on Build GRCh38
rs200558126 Chr.2: 218262724-218262724 on Build GRCh38
rs375107430 Chr.2: 218262708-218262709 on Build GRCh38
rs201260550 Chr.2: 218262985-218262985 on Build GRCh38
rs397695422 Chr.2: 218262931-218262931 on Build GRCh38
rs1047433769 Chr.2: 218262828-218262828 on Build GRCh38
rs762928432 Chr.2: 218263091-218263091 on Build GRCh38
rs550649048 Chr.2: 218262711-218262711 on Build GRCh38
rs200915643 Chr.2: 218263124-218263124 on Build GRCh38
rs373073683 Chr.2: 218263001-218263001 on Build GRCh38
rs536606884 Chr.2: 218262992-218262992 on Build GRCh38
rs199765208 Chr.2: 218263122-218263122 on Build GRCh38
rs776924820 Chr.2: 218262782-218262782 on Build GRCh38
rs776671006 Chr.2: 218263093-218263093 on Build GRCh38
rs753215679 Chr.2: 218262921-218262926 on Build GRCh38
rs531707356 Chr.2: 218262808-218262808 on Build GRCh38
rs201747442 Chr.2: 218262963-218262963 on Build GRCh38
rs202136704 Chr.2: 218263052-218263052 on Build GRCh38
rs747879212 Chr.2: 218262758-218262758 on Build GRCh38
rs949839397 Chr.2: 218262854-218262854 on Build GRCh38
rs200257647 Chr.2: 218263097-218263097 on Build GRCh38
rs760796376 Chr.2: 218263080-218263080 on Build GRCh38
rs5838688 Chr.2: 218262931-218262932 on Build GRCh38
rs771789619 Chr.2: 218262830-218262830 on Build GRCh38
rs56192869 Chr.2: 218262694-218262694 on Build GRCh38
rs764902875 Chr.2: 218262926-218262926 on Build GRCh38
rs994702901 Chr.2: 218263127-218263127 on Build GRCh38
rs398105345 Chr.2: 218262929-218262929 on Build GRCh38
rs1020329611 Chr.2: 218262945-218262945 on Build GRCh38
rs369671903 Chr.2: 218262957-218262957 on Build GRCh38
rs200717262 Chr.2: 218262687-218262687 on Build GRCh38
rs933947869 Chr.2: 218262826-218262826 on Build GRCh38
rs927090977 Chr.2: 218263084-218263084 on Build GRCh38
rs746731355 Chr.2: 218262946-218262946 on Build GRCh38
rs57621524 Chr.2: 218262686-218262686 on Build GRCh38
rs145881577 Chr.2: 218262971-218262971 on Build GRCh38
rs554480858 Chr.2: 218263120-218263120 on Build GRCh38
rs767819043 Chr.2: 218262880-218262882 on Build GRCh38
rs1051084190 Chr.2: 218263083-218263083 on Build GRCh38
rs769115943 Chr.2: 218263061-218263061 on Build GRCh38
rs869146113 Chr.2: 218262930-218262930 on Build GRCh38
rs199822804 Chr.2: 218262959-218262959 on Build GRCh38
rs371444837 Chr.2: 218262749-218262749 on Build GRCh38
rs927173480 Chr.2: 218262729-218262729 on Build GRCh38
rs201963856 Chr.2: 218262816-218262816 on Build GRCh38
rs761133945 Chr.2: 218262778-218262778 on Build GRCh38
rs993922751 Chr.2: 218262919-218262919 on Build GRCh38

View Full Product Details