Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218357448-218357950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATIP
Entrez Gene ID
375307
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C2orf62
UniGene
Hs.645453
Gene Chromosome Location
Chr.2: 218356757-218368393 on Build GRCh38
Transcripts
NM_001320865.1 NM_198559.1 XM_005246539.4 XM_005246541.4 XM_011511148.2 XM_011511149.2 XM_011511150.2 XM_017004053.1 NR_125777.1

Gene Symbol
CATIP-AS2
Entrez Gene ID
103689911
Gene Name
CATIP antisense RNA 2
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 218326241-218357966 on Build GRCh38
Transcripts
NM_001320865.1 NM_198559.1 XM_005246539.4 XM_005246541.4 XM_011511148.2 XM_011511149.2 XM_011511150.2 XM_017004053.1 NR_125777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3043394 c.311C>T p.S104F Chr.2: 218357726-218357726 on Build GRCh38
1318110 c.261C>T p.F87F Chr.2: 218357676-218357676 on Build GRCh38
3731718 c.177C>T p.T59T Chr.2: 218357592-218357592 on Build GRCh38
1177306 c.237C>T p.L79L Chr.2: 218357652-218357652 on Build GRCh38
5003683 c.205G>A p.E69K Chr.2: 218357620-218357620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768289246 Chr.2: 218357557-218357559 on Build GRCh38
rs935454082 Chr.2: 218357910-218357910 on Build GRCh38
rs781447676 Chr.2: 218357593-218357593 on Build GRCh38
rs992065921 Chr.2: 218357868-218357868 on Build GRCh38
rs962784240 Chr.2: 218357827-218357827 on Build GRCh38
rs761316092 Chr.2: 218357677-218357677 on Build GRCh38
rs776215081 Chr.2: 218357718-218357719 on Build GRCh38
rs769832037 Chr.2: 218357506-218357506 on Build GRCh38
rs1033391653 Chr.2: 218357539-218357539 on Build GRCh38
rs573757108 Chr.2: 218357857-218357857 on Build GRCh38
rs773224783 Chr.2: 218357653-218357653 on Build GRCh38
rs760579695 Chr.2: 218357667-218357667 on Build GRCh38
rs192851347 Chr.2: 218357571-218357571 on Build GRCh38
rs780446363 Chr.2: 218357779-218357779 on Build GRCh38
rs1011093884 Chr.2: 218357757-218357757 on Build GRCh38
rs775257872 Chr.2: 218357735-218357735 on Build GRCh38
rs771957959 Chr.2: 218357629-218357629 on Build GRCh38
rs146310684 Chr.2: 218357477-218357477 on Build GRCh38
rs568366738 Chr.2: 218357747-218357747 on Build GRCh38
rs369378162 Chr.2: 218357570-218357570 on Build GRCh38
rs142868592 Chr.2: 218357856-218357856 on Build GRCh38
rs977112382 Chr.2: 218357598-218357598 on Build GRCh38
rs200838081 Chr.2: 218357518-218357518 on Build GRCh38
rs1002344706 Chr.2: 218357712-218357712 on Build GRCh38
rs759722732 Chr.2: 218357490-218357490 on Build GRCh38
rs759205178 Chr.2: 218357683-218357683 on Build GRCh38
rs775917465 Chr.2: 218357633-218357633 on Build GRCh38
rs144662667 Chr.2: 218357592-218357592 on Build GRCh38
rs750122627 Chr.2: 218357517-218357517 on Build GRCh38
rs150544491 Chr.2: 218357718-218357718 on Build GRCh38
rs922503699 Chr.2: 218357603-218357603 on Build GRCh38
rs1031210038 Chr.2: 218357726-218357726 on Build GRCh38
rs865811462 Chr.2: 218357528-218357528 on Build GRCh38
rs139498285 Chr.2: 218357760-218357760 on Build GRCh38
rs916509415 Chr.2: 218357877-218357877 on Build GRCh38
rs137894462 Chr.2: 218357703-218357703 on Build GRCh38
rs771251364 Chr.2: 218357572-218357572 on Build GRCh38
rs373595974 Chr.2: 218357906-218357906 on Build GRCh38
rs757726894 Chr.2: 218357753-218357753 on Build GRCh38
rs375181104 Chr.2: 218357702-218357702 on Build GRCh38
rs766168040 Chr.2: 218357676-218357676 on Build GRCh38
rs142253999 Chr.2: 218357701-218357701 on Build GRCh38
rs199998012 Chr.2: 218357740-218357740 on Build GRCh38
rs770785298 Chr.2: 218357728-218357728 on Build GRCh38
rs953917504 Chr.2: 218357895-218357895 on Build GRCh38
rs747963336 Chr.2: 218357757-218357768 on Build GRCh38
rs777124954 Chr.2: 218357835-218357836 on Build GRCh38
rs374390488 Chr.2: 218357696-218357696 on Build GRCh38
rs764985775 Chr.2: 218357687-218357687 on Build GRCh38
rs1044125759 Chr.2: 218357907-218357907 on Build GRCh38
rs539311895 Chr.2: 218357744-218357744 on Build GRCh38
rs371891084 Chr.2: 218357682-218357682 on Build GRCh38
rs778455674 Chr.2: 218357576-218357576 on Build GRCh38
rs145097529 Chr.2: 218357698-218357698 on Build GRCh38
rs770061389 Chr.2: 218357595-218357595 on Build GRCh38
rs976407472 Chr.2: 218357824-218357824 on Build GRCh38
rs764934635 Chr.2: 218357733-218357733 on Build GRCh38
rs140422711 Chr.2: 218357652-218357652 on Build GRCh38
rs371607639 Chr.2: 218357508-218357508 on Build GRCh38
rs376076590 Chr.2: 218357546-218357546 on Build GRCh38
rs142157169 Chr.2: 218357585-218357585 on Build GRCh38
rs777471545 Chr.2: 218357693-218357693 on Build GRCh38
rs376424752 Chr.2: 218357488-218357488 on Build GRCh38
rs188527626 Chr.2: 218357783-218357783 on Build GRCh38
rs147879826 Chr.2: 218357614-218357614 on Build GRCh38
rs771014396 Chr.2: 218357485-218357485 on Build GRCh38
rs966292365 Chr.2: 218357581-218357581 on Build GRCh38
rs957759208 Chr.2: 218357544-218357544 on Build GRCh38
rs985269206 Chr.2: 218357679-218357679 on Build GRCh38
rs918456652 Chr.2: 218357856-218357857 on Build GRCh38
rs769940242 Chr.2: 218357706-218357706 on Build GRCh38
rs543325743 Chr.2: 218357885-218357885 on Build GRCh38
rs755262499 Chr.2: 218357785-218357785 on Build GRCh38
rs775494004 Chr.2: 218357608-218357608 on Build GRCh38
rs746096715 Chr.2: 218357594-218357594 on Build GRCh38
rs756493179 Chr.2: 218357770-218357770 on Build GRCh38
rs754628518 Chr.2: 218357534-218357534 on Build GRCh38
rs200499642 Chr.2: 218357719-218357719 on Build GRCh38
rs910655532 Chr.2: 218357680-218357680 on Build GRCh38
rs753507964 Chr.2: 218357522-218357522 on Build GRCh38
rs762642450 Chr.2: 218357791-218357791 on Build GRCh38
rs780290660 Chr.2: 218357716-218357716 on Build GRCh38
rs188567053 Chr.2: 218357472-218357472 on Build GRCh38
rs201917148 Chr.2: 218357759-218357759 on Build GRCh38
rs767459688 Chr.2: 218357511-218357511 on Build GRCh38
rs149309207 Chr.2: 218357725-218357725 on Build GRCh38
rs1052587751 Chr.2: 218357918-218357918 on Build GRCh38
rs78470620 Chr.2: 218357692-218357692 on Build GRCh38
rs544369407 Chr.2: 218357908-218357908 on Build GRCh38
rs147403100 Chr.2: 218357732-218357732 on Build GRCh38
rs749514603 Chr.2: 218357554-218357554 on Build GRCh38

View Full Product Details