Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218959550-218960055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5R2
Entrez Gene ID
8941
Gene Name
cyclin dependent kinase 5 regulatory subunit 2
Gene Aliases
NCK5AI, P39, p39nck5ai
UniGene
Hs.158460
Gene Chromosome Location
Chr.2: 218959628-218962155 on Build GRCh38
Transcripts
NM_003936.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5692745 c.84G>A p.A28A Chr.2: 218959904-218959904 on Build GRCh38
6362238 c.92C>A p.A31E Chr.2: 218959912-218959912 on Build GRCh38
6392064 c.68A>T p.E23V Chr.2: 218959888-218959888 on Build GRCh38
5716674 c.94G>A p.G32R Chr.2: 218959914-218959914 on Build GRCh38
3380045 c.120G>A p.G40G Chr.2: 218959940-218959940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995381580 Chr.2: 218959669-218959669 on Build GRCh38
rs963291866 Chr.2: 218959810-218959810 on Build GRCh38
rs781635214 Chr.2: 218959963-218959963 on Build GRCh38
rs954717941 Chr.2: 218959597-218959597 on Build GRCh38
rs912179935 Chr.2: 218959590-218959590 on Build GRCh38
rs775738948 Chr.2: 218959847-218959847 on Build GRCh38
rs764803522 Chr.2: 218959943-218959943 on Build GRCh38
rs929174082 Chr.2: 218959660-218959660 on Build GRCh38
rs1030462103 Chr.2: 218959589-218959589 on Build GRCh38
rs35803444 Chr.2: 218959682-218959682 on Build GRCh38
rs746174739 Chr.2: 218959981-218959981 on Build GRCh38
rs199733283 Chr.2: 218960013-218960013 on Build GRCh38
rs542541754 Chr.2: 218959874-218959874 on Build GRCh38
rs894701843 Chr.2: 218959697-218959697 on Build GRCh38
rs1033601855 Chr.2: 218959844-218959844 on Build GRCh38
rs375734597 Chr.2: 218959591-218959591 on Build GRCh38
rs1048146506 Chr.2: 218959672-218959672 on Build GRCh38
rs34275515 Chr.2: 218959925-218959925 on Build GRCh38
rs144368604 Chr.2: 218959986-218959986 on Build GRCh38
rs999050469 Chr.2: 218959753-218959753 on Build GRCh38
rs967423312 Chr.2: 218959583-218959583 on Build GRCh38
rs572831666 Chr.2: 218959607-218959607 on Build GRCh38
rs201718137 Chr.2: 218959849-218959849 on Build GRCh38
rs1002144360 Chr.2: 218959695-218959696 on Build GRCh38
rs184241197 Chr.2: 218959674-218959674 on Build GRCh38
rs943337282 Chr.2: 218959722-218959722 on Build GRCh38
rs769092100 Chr.2: 218959592-218959592 on Build GRCh38
rs1021086808 Chr.2: 218959701-218959701 on Build GRCh38
rs753190569 Chr.2: 218959878-218959878 on Build GRCh38
rs986566815 Chr.2: 218959605-218959605 on Build GRCh38
rs890549077 Chr.2: 218959760-218959760 on Build GRCh38
rs937676493 Chr.2: 218959693-218959693 on Build GRCh38
rs899346295 Chr.2: 218959655-218959655 on Build GRCh38
rs774552967 Chr.2: 218960033-218960033 on Build GRCh38
rs753529638 Chr.2: 218959910-218959910 on Build GRCh38
rs367886212 Chr.2: 218959743-218959743 on Build GRCh38
rs540261251 Chr.2: 218959612-218959612 on Build GRCh38
rs777239345 Chr.2: 218959891-218959893 on Build GRCh38
rs963644577 Chr.2: 218959608-218959608 on Build GRCh38
rs557882480 Chr.2: 218959576-218959576 on Build GRCh38
rs964744539 Chr.2: 218959962-218959962 on Build GRCh38
rs946405757 Chr.2: 218959991-218959991 on Build GRCh38
rs939753677 Chr.2: 218959795-218959795 on Build GRCh38
rs893709658 Chr.2: 218959848-218959848 on Build GRCh38
rs1002241882 Chr.2: 218959838-218959838 on Build GRCh38
rs906482667 Chr.2: 218959690-218959691 on Build GRCh38
rs930866502 Chr.2: 218959765-218959765 on Build GRCh38
rs757960284 Chr.2: 218959948-218959948 on Build GRCh38
rs983607841 Chr.2: 218960008-218960008 on Build GRCh38
rs989591800 Chr.2: 218959581-218959581 on Build GRCh38
rs947619160 Chr.2: 218959705-218959705 on Build GRCh38
rs756638550 Chr.2: 218959904-218959904 on Build GRCh38
rs867582509 Chr.2: 218959857-218959857 on Build GRCh38
rs1048651262 Chr.2: 218959695-218959695 on Build GRCh38
rs746148262 Chr.2: 218959805-218959805 on Build GRCh38
rs773353083 Chr.2: 218959875-218959875 on Build GRCh38
rs943616821 Chr.2: 218959648-218959648 on Build GRCh38
rs919098334 Chr.2: 218959642-218959642 on Build GRCh38
rs930878539 Chr.2: 218960000-218960000 on Build GRCh38
rs987494648 Chr.2: 218959707-218959707 on Build GRCh38
rs200705478 Chr.2: 218959945-218959945 on Build GRCh38
rs769856179 Chr.2: 218959809-218959809 on Build GRCh38
rs886887259 Chr.2: 218959792-218959792 on Build GRCh38
rs975031216 Chr.2: 218959748-218959748 on Build GRCh38
rs35910919 Chr.2: 218959696-218959696 on Build GRCh38
rs757681013 Chr.2: 218959960-218959960 on Build GRCh38
rs1007535574 Chr.2: 218959789-218959789 on Build GRCh38
rs78141300 Chr.2: 218960009-218960009 on Build GRCh38
rs768901003 Chr.2: 218960022-218960022 on Build GRCh38
rs766248382 Chr.2: 218959900-218959900 on Build GRCh38
rs759878477 Chr.2: 218959801-218959801 on Build GRCh38
rs774796924 Chr.2: 218959785-218959785 on Build GRCh38
rs747589497 Chr.2: 218959954-218959954 on Build GRCh38
rs777259963 Chr.2: 218959952-218959952 on Build GRCh38
rs950535523 Chr.2: 218959867-218959867 on Build GRCh38
rs1043365403 Chr.2: 218959716-218959716 on Build GRCh38
rs998934222 Chr.2: 218959587-218959587 on Build GRCh38
rs1030409726 Chr.2: 218959759-218959759 on Build GRCh38
rs1011716265 Chr.2: 218959574-218959574 on Build GRCh38
rs760501749 Chr.2: 218959886-218959886 on Build GRCh38

View Full Product Details