Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218961246-218961783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5R2
Entrez Gene ID
8941
Gene Name
cyclin dependent kinase 5 regulatory subunit 2
Gene Aliases
NCK5AI, P39, p39nck5ai
UniGene
Hs.158460
Gene Chromosome Location
Chr.2: 218959628-218962155 on Build GRCh38
Transcripts
NM_003936.4

SNP Detail

SNP ID Chromosome Location
rs999269228 Chr.2: 218961443-218961443 on Build GRCh38
rs188341731 Chr.2: 218961306-218961306 on Build GRCh38
rs930846095 Chr.2: 218961666-218961666 on Build GRCh38
rs369265011 Chr.2: 218961298-218961299 on Build GRCh38
rs983483450 Chr.2: 218961710-218961710 on Build GRCh38
rs944325611 Chr.2: 218961487-218961487 on Build GRCh38
rs931845953 Chr.2: 218961568-218961568 on Build GRCh38
rs1019586390 Chr.2: 218961543-218961543 on Build GRCh38
rs887889764 Chr.2: 218961588-218961588 on Build GRCh38
rs976412497 Chr.2: 218961540-218961541 on Build GRCh38
rs1030795992 Chr.2: 218961471-218961471 on Build GRCh38
rs921832748 Chr.2: 218961547-218961547 on Build GRCh38
rs1022140434 Chr.2: 218961402-218961402 on Build GRCh38
rs756219902 Chr.2: 218961298-218961301 on Build GRCh38
rs142470311 Chr.2: 218961681-218961681 on Build GRCh38
rs548843309 Chr.2: 218961747-218961747 on Build GRCh38
rs920770197 Chr.2: 218961663-218961663 on Build GRCh38
rs1021784560 Chr.2: 218961738-218961738 on Build GRCh38
rs955195081 Chr.2: 218961522-218961522 on Build GRCh38
rs967777859 Chr.2: 218961395-218961395 on Build GRCh38
rs1003571835 Chr.2: 218961331-218961331 on Build GRCh38
rs895016764 Chr.2: 218961753-218961753 on Build GRCh38
rs894719961 Chr.2: 218961728-218961728 on Build GRCh38
rs895078213 Chr.2: 218961360-218961360 on Build GRCh38
rs959132053 Chr.2: 218961329-218961332 on Build GRCh38
rs990332891 Chr.2: 218961353-218961353 on Build GRCh38
rs193030166 Chr.2: 218961423-218961423 on Build GRCh38
rs1056774483 Chr.2: 218961355-218961355 on Build GRCh38
rs1012471956 Chr.2: 218961389-218961389 on Build GRCh38
rs977777056 Chr.2: 218961432-218961433 on Build GRCh38
rs1012188178 Chr.2: 218961730-218961731 on Build GRCh38
rs1035054732 Chr.2: 218961672-218961672 on Build GRCh38
rs965006119 Chr.2: 218961597-218961597 on Build GRCh38
rs940799062 Chr.2: 218961626-218961626 on Build GRCh38
rs879012266 Chr.2: 218961311-218961311 on Build GRCh38
rs1036378352 Chr.2: 218961653-218961653 on Build GRCh38
rs986511980 Chr.2: 218961534-218961534 on Build GRCh38
rs1034574971 Chr.2: 218961329-218961330 on Build GRCh38
rs750474076 Chr.2: 218961298-218961298 on Build GRCh38
rs967756608 Chr.2: 218961429-218961429 on Build GRCh38
rs552890700 Chr.2: 218961310-218961310 on Build GRCh38
rs939320998 Chr.2: 218961732-218961732 on Build GRCh38
rs533946267 Chr.2: 218961630-218961630 on Build GRCh38
rs533095071 Chr.2: 218961299-218961299 on Build GRCh38
rs1048875789 Chr.2: 218961569-218961569 on Build GRCh38
rs1022499540 Chr.2: 218961372-218961372 on Build GRCh38
rs796912458 Chr.2: 218961358-218961358 on Build GRCh38
rs1002838498 Chr.2: 218961664-218961664 on Build GRCh38
rs907997034 Chr.2: 218961712-218961712 on Build GRCh38
rs764651435 Chr.2: 218961300-218961301 on Build GRCh38

View Full Product Details