Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219384032-219384528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNPEP
Entrez Gene ID
23549
Gene Name
aspartyl aminopeptidase
Gene Aliases
ASPEP, DAP
UniGene
Hs.258551
Gene Chromosome Location
Chr.2: 219372029-219400022 on Build GRCh38
Transcripts
NM_001319116.1 NM_001319117.1 NM_001319118.1 NM_001319119.1 NM_001319120.1 NM_001319121.1 NM_001319122.1 NM_012100.3 NR_134970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752369 c.790T>A p.Y264N Chr.2: 219384428-219384428 on Build GRCh38
6307738 c.846C>A p.A282A Chr.2: 219384372-219384372 on Build GRCh38
442346 c.786delT p.A263fs*24 Chr.2: 219384432-219384432 on Build GRCh38
6864975 c.850C>T p.Q284* Chr.2: 219384368-219384368 on Build GRCh38
6647882 c.788C>A p.A263D Chr.2: 219384430-219384430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981687627 Chr.2: 219384253-219384253 on Build GRCh38
rs373669687 Chr.2: 219384478-219384478 on Build GRCh38
rs941470991 Chr.2: 219384090-219384090 on Build GRCh38
rs751807198 Chr.2: 219384203-219384203 on Build GRCh38
rs370318556 Chr.2: 219384473-219384473 on Build GRCh38
rs999429029 Chr.2: 219384189-219384189 on Build GRCh38
rs773401674 Chr.2: 219384132-219384134 on Build GRCh38
rs759449946 Chr.2: 219384360-219384360 on Build GRCh38
rs1005477295 Chr.2: 219384368-219384368 on Build GRCh38
rs930207227 Chr.2: 219384098-219384098 on Build GRCh38
rs764935323 Chr.2: 219384092-219384092 on Build GRCh38
rs769728005 Chr.2: 219384479-219384479 on Build GRCh38
rs184116311 Chr.2: 219384140-219384140 on Build GRCh38
rs377653451 Chr.2: 219384447-219384447 on Build GRCh38
rs529464642 Chr.2: 219384277-219384277 on Build GRCh38
rs937648542 Chr.2: 219384303-219384303 on Build GRCh38
rs923865766 Chr.2: 219384232-219384232 on Build GRCh38
rs567948180 Chr.2: 219384251-219384251 on Build GRCh38
rs367618988 Chr.2: 219384488-219384488 on Build GRCh38
rs758525950 Chr.2: 219384131-219384131 on Build GRCh38
rs372276024 Chr.2: 219384489-219384489 on Build GRCh38
rs60712769 Chr.2: 219384281-219384281 on Build GRCh38
rs752488599 Chr.2: 219384403-219384403 on Build GRCh38
rs11539911 Chr.2: 219384411-219384411 on Build GRCh38
rs758123570 Chr.2: 219384404-219384404 on Build GRCh38
rs763667668 Chr.2: 219384408-219384408 on Build GRCh38
rs971596326 Chr.2: 219384130-219384130 on Build GRCh38
rs762760569 Chr.2: 219384486-219384486 on Build GRCh38
rs955347733 Chr.2: 219384192-219384192 on Build GRCh38
rs73085218 Chr.2: 219384350-219384350 on Build GRCh38
rs374335152 Chr.2: 219384438-219384438 on Build GRCh38
rs777973824 Chr.2: 219384358-219384358 on Build GRCh38
rs10804272 Chr.2: 219384205-219384205 on Build GRCh38
rs574862095 Chr.2: 219384356-219384356 on Build GRCh38
rs551878374 Chr.2: 219384157-219384157 on Build GRCh38
rs923728332 Chr.2: 219384497-219384497 on Build GRCh38
rs962686209 Chr.2: 219384466-219384466 on Build GRCh38
rs559099942 Chr.2: 219384218-219384218 on Build GRCh38
rs981998663 Chr.2: 219384471-219384471 on Build GRCh38
rs1054789578 Chr.2: 219384307-219384307 on Build GRCh38
rs774570513 Chr.2: 219384355-219384355 on Build GRCh38
rs756048332 Chr.2: 219384468-219384468 on Build GRCh38
rs995971504 Chr.2: 219384304-219384304 on Build GRCh38
rs775349091 Chr.2: 219384484-219384484 on Build GRCh38
rs143090933 Chr.2: 219384215-219384215 on Build GRCh38
rs930154291 Chr.2: 219384252-219384252 on Build GRCh38
rs374131877 Chr.2: 219384462-219384462 on Build GRCh38
rs200396385 Chr.2: 219384425-219384425 on Build GRCh38
rs751177542 Chr.2: 219384422-219384422 on Build GRCh38
rs560352729 Chr.2: 219384073-219384073 on Build GRCh38
rs571654019 Chr.2: 219384190-219384190 on Build GRCh38
rs763366996 Chr.2: 219384268-219384268 on Build GRCh38
rs557260243 Chr.2: 219384064-219384066 on Build GRCh38
rs775982398 Chr.2: 219384337-219384337 on Build GRCh38
rs768999516 Chr.2: 219384351-219384351 on Build GRCh38
rs181316361 Chr.2: 219384431-219384431 on Build GRCh38
rs780270082 Chr.2: 219384171-219384171 on Build GRCh38
rs775626386 Chr.2: 219384220-219384220 on Build GRCh38
rs944581103 Chr.2: 219384241-219384241 on Build GRCh38
rs1018195183 Chr.2: 219384464-219384464 on Build GRCh38
rs1015024063 Chr.2: 219384373-219384373 on Build GRCh38
rs1054824142 Chr.2: 219384122-219384122 on Build GRCh38
rs549393948 Chr.2: 219384104-219384104 on Build GRCh38
rs918716501 Chr.2: 219384250-219384250 on Build GRCh38
rs951524116 Chr.2: 219384470-219384470 on Build GRCh38
rs765078445 Chr.2: 219384367-219384367 on Build GRCh38
rs902347036 Chr.2: 219384175-219384175 on Build GRCh38
rs553390506 Chr.2: 219384097-219384097 on Build GRCh38
rs370962125 Chr.2: 219384458-219384458 on Build GRCh38
rs542578437 Chr.2: 219384185-219384185 on Build GRCh38
rs780729824 Chr.2: 219384427-219384427 on Build GRCh38
rs367985800 Chr.2: 219384467-219384467 on Build GRCh38
rs573531741 Chr.2: 219384111-219384111 on Build GRCh38
rs939026540 Chr.2: 219384112-219384112 on Build GRCh38
rs761147142 Chr.2: 219384409-219384409 on Build GRCh38
rs11539913 Chr.2: 219384407-219384407 on Build GRCh38

View Full Product Details