Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219426231-219426741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DES
Entrez Gene ID
1674
Gene Name
desmin
Gene Aliases
CSM1, CSM2, LGMD2R
UniGene
Hs.594952
Gene Chromosome Location
Chr.2: 219418377-219426739 on Build GRCh38
Transcripts
NM_001927.3

SNP Detail

SNP ID Chromosome Location
rs757955535 Chr.2: 219426482-219426482 on Build GRCh38
rs935084030 Chr.2: 219426655-219426655 on Build GRCh38
rs887622719 Chr.2: 219426418-219426418 on Build GRCh38
rs745307832 Chr.2: 219426469-219426469 on Build GRCh38
rs901580524 Chr.2: 219426379-219426379 on Build GRCh38
rs73085265 Chr.2: 219426458-219426458 on Build GRCh38
rs181219659 Chr.2: 219426674-219426674 on Build GRCh38
rs548986110 Chr.2: 219426275-219426275 on Build GRCh38
rs140222667 Chr.2: 219426298-219426298 on Build GRCh38
rs980823712 Chr.2: 219426539-219426539 on Build GRCh38
rs1058616 Chr.2: 219426333-219426333 on Build GRCh38
rs116635264 Chr.2: 219426652-219426652 on Build GRCh38
rs886055655 Chr.2: 219426479-219426479 on Build GRCh38
rs547498920 Chr.2: 219426549-219426549 on Build GRCh38
rs138913201 Chr.2: 219426465-219426465 on Build GRCh38
rs969442170 Chr.2: 219426529-219426529 on Build GRCh38
rs988776615 Chr.2: 219426591-219426591 on Build GRCh38
rs1032888762 Chr.2: 219426548-219426548 on Build GRCh38
rs925513578 Chr.2: 219426270-219426270 on Build GRCh38
rs931755403 Chr.2: 219426381-219426381 on Build GRCh38
rs980897423 Chr.2: 219426269-219426269 on Build GRCh38
rs1040087255 Chr.2: 219426342-219426342 on Build GRCh38
rs941853932 Chr.2: 219426686-219426686 on Build GRCh38
rs760272041 Chr.2: 219426291-219426291 on Build GRCh38
rs2859788 Chr.2: 219426684-219426684 on Build GRCh38
rs371079685 Chr.2: 219426651-219426651 on Build GRCh38
rs897756868 Chr.2: 219426478-219426478 on Build GRCh38
rs113832558 Chr.2: 219426671-219426671 on Build GRCh38
rs150319065 Chr.2: 219426351-219426351 on Build GRCh38
rs988417423 Chr.2: 219426288-219426288 on Build GRCh38
rs746886224 Chr.2: 219426578-219426578 on Build GRCh38
rs914139708 Chr.2: 219426602-219426602 on Build GRCh38
rs912862221 Chr.2: 219426296-219426296 on Build GRCh38
rs886055654 Chr.2: 219426418-219426418 on Build GRCh38
rs887581721 Chr.2: 219426678-219426678 on Build GRCh38
rs1058876 Chr.2: 219426433-219426433 on Build GRCh38
rs1024964792 Chr.2: 219426518-219426518 on Build GRCh38
rs567741120 Chr.2: 219426637-219426637 on Build GRCh38
rs1036632098 Chr.2: 219426715-219426715 on Build GRCh38
rs975740337 Chr.2: 219426640-219426640 on Build GRCh38
rs551321742 Chr.2: 219426407-219426407 on Build GRCh38
rs768035073 Chr.2: 219426366-219426366 on Build GRCh38

View Full Product Details