Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219531915-219532407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC4
Entrez Gene ID
55515
Gene Name
acid sensing ion channel subunit family member 4
Gene Aliases
ACCN4, BNAC4
UniGene
Hs.87469
Gene Chromosome Location
Chr.2: 219514170-219538772 on Build GRCh38
Transcripts
NM_182847.2 XM_005246671.2 XM_017004439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016832 c.1166C>T p.P389L Chr.2: 219532058-219532058 on Build GRCh38
1016833 c.1184G>A p.G395E Chr.2: 219532076-219532076 on Build GRCh38
6010665 c.1118C>T p.S373L Chr.2: 219532010-219532010 on Build GRCh38
3578048 c.1291G>A p.E431K Chr.2: 219532369-219532369 on Build GRCh38
1016834 c.1201G>T p.G401C Chr.2: 219532093-219532093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202037132 Chr.2: 219532025-219532025 on Build GRCh38
rs750769790 Chr.2: 219532126-219532126 on Build GRCh38
rs1038129946 Chr.2: 219532146-219532146 on Build GRCh38
rs776637130 Chr.2: 219532067-219532067 on Build GRCh38
rs980004807 Chr.2: 219531997-219531997 on Build GRCh38
rs759497969 Chr.2: 219532183-219532183 on Build GRCh38
rs770276072 Chr.2: 219532041-219532041 on Build GRCh38
rs760931120 Chr.2: 219532365-219532365 on Build GRCh38
rs757090803 Chr.2: 219532348-219532348 on Build GRCh38
rs940674797 Chr.2: 219532048-219532048 on Build GRCh38
rs569648325 Chr.2: 219531959-219531959 on Build GRCh38
rs770536032 Chr.2: 219532306-219532306 on Build GRCh38
rs770338987 Chr.2: 219531953-219531953 on Build GRCh38
rs544029545 Chr.2: 219532186-219532186 on Build GRCh38
rs773833798 Chr.2: 219532053-219532053 on Build GRCh38
rs757985394 Chr.2: 219532294-219532294 on Build GRCh38
rs896996947 Chr.2: 219531944-219531944 on Build GRCh38
rs148895186 Chr.2: 219532372-219532372 on Build GRCh38
rs918244738 Chr.2: 219532347-219532347 on Build GRCh38
rs774414742 Chr.2: 219532185-219532185 on Build GRCh38
rs746400183 Chr.2: 219531947-219531947 on Build GRCh38
rs59761991 Chr.2: 219532148-219532148 on Build GRCh38
rs765130281 Chr.2: 219532184-219532184 on Build GRCh38
rs546484806 Chr.2: 219532262-219532262 on Build GRCh38
rs766406205 Chr.2: 219532002-219532002 on Build GRCh38
rs780664613 Chr.2: 219532044-219532044 on Build GRCh38
rs750336176 Chr.2: 219531985-219531985 on Build GRCh38
rs767798894 Chr.2: 219532122-219532122 on Build GRCh38
rs745712688 Chr.2: 219532342-219532342 on Build GRCh38
rs758520189 Chr.2: 219532008-219532008 on Build GRCh38
rs202019494 Chr.2: 219532166-219532166 on Build GRCh38
rs376578334 Chr.2: 219531945-219531945 on Build GRCh38
rs1001480270 Chr.2: 219532182-219532182 on Build GRCh38
rs576114197 Chr.2: 219532366-219532366 on Build GRCh38
rs374509292 Chr.2: 219532286-219532286 on Build GRCh38
rs148791410 Chr.2: 219532063-219532063 on Build GRCh38
rs34784127 Chr.2: 219532350-219532350 on Build GRCh38
rs759831448 Chr.2: 219532379-219532379 on Build GRCh38
rs747603402 Chr.2: 219532288-219532288 on Build GRCh38
rs747700795 Chr.2: 219532160-219532160 on Build GRCh38
rs752706307 Chr.2: 219532007-219532007 on Build GRCh38
rs769234826 Chr.2: 219532303-219532303 on Build GRCh38
rs149591037 Chr.2: 219532256-219532256 on Build GRCh38
rs754395464 Chr.2: 219532142-219532142 on Build GRCh38
rs572070268 Chr.2: 219532056-219532056 on Build GRCh38
rs757444378 Chr.2: 219532114-219532114 on Build GRCh38
rs1034380385 Chr.2: 219532011-219532011 on Build GRCh38
rs371570850 Chr.2: 219532171-219532171 on Build GRCh38
rs774064154 Chr.2: 219531962-219531962 on Build GRCh38
rs113299906 Chr.2: 219532351-219532351 on Build GRCh38
rs776385892 Chr.2: 219532181-219532181 on Build GRCh38
rs200868720 Chr.2: 219532135-219532135 on Build GRCh38
rs756510718 Chr.2: 219532127-219532127 on Build GRCh38
rs745384849 Chr.2: 219532349-219532349 on Build GRCh38
rs776452320 Chr.2: 219532324-219532324 on Build GRCh38
rs747279893 Chr.2: 219531938-219531938 on Build GRCh38
rs199990846 Chr.2: 219532373-219532373 on Build GRCh38
rs1022031899 Chr.2: 219532150-219532150 on Build GRCh38
rs755919346 Chr.2: 219532080-219532080 on Build GRCh38
rs774836309 Chr.2: 219531958-219531958 on Build GRCh38
rs1017069120 Chr.2: 219531936-219531936 on Build GRCh38
rs761632057 Chr.2: 219531979-219531979 on Build GRCh38
rs1020282770 Chr.2: 219532195-219532195 on Build GRCh38
rs777615707 Chr.2: 219532169-219532169 on Build GRCh38
rs558548389 Chr.2: 219532049-219532049 on Build GRCh38
rs751690752 Chr.2: 219532099-219532099 on Build GRCh38
rs941037510 Chr.2: 219532313-219532313 on Build GRCh38
rs766790420 Chr.2: 219532196-219532196 on Build GRCh38
rs751813411 Chr.2: 219532013-219532013 on Build GRCh38
rs777793535 Chr.2: 219532010-219532010 on Build GRCh38
rs909316143 Chr.2: 219532028-219532028 on Build GRCh38
rs757872748 Chr.2: 219532027-219532027 on Build GRCh38
rs984904881 Chr.2: 219531980-219531980 on Build GRCh38
rs777526690 Chr.2: 219532302-219532302 on Build GRCh38
rs971340485 Chr.2: 219531957-219531957 on Build GRCh38
rs555094972 Chr.2: 219532140-219532140 on Build GRCh38
rs764258912 Chr.2: 219532086-219532086 on Build GRCh38
rs755480746 Chr.2: 219532274-219532274 on Build GRCh38
rs779630344 Chr.2: 219532282-219532282 on Build GRCh38
rs760667027 Chr.2: 219531986-219531986 on Build GRCh38
rs373384533 Chr.2: 219532189-219532189 on Build GRCh38
rs757571805 Chr.2: 219532020-219532020 on Build GRCh38
rs773111154 Chr.2: 219532062-219532062 on Build GRCh38
rs780774383 Chr.2: 219532209-219532209 on Build GRCh38
rs767672359 Chr.2: 219532188-219532188 on Build GRCh38
rs754205418 Chr.2: 219532270-219532270 on Build GRCh38
rs201323394 Chr.2: 219532055-219532055 on Build GRCh38
rs923465021 Chr.2: 219532257-219532257 on Build GRCh38
rs768213575 Chr.2: 219532050-219532050 on Build GRCh38
rs186898701 Chr.2: 219531946-219531946 on Build GRCh38
rs746734801 Chr.2: 219532170-219532170 on Build GRCh38
rs932034238 Chr.2: 219531955-219531955 on Build GRCh38
rs781311841 Chr.2: 219532336-219532337 on Build GRCh38

View Full Product Details