Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231397624-231398123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT7
Entrez Gene ID
93010
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7
Gene Aliases
beta3GnT7
UniGene
Hs.299329
Gene Chromosome Location
Chr.2: 231395624-231401164 on Build GRCh38
Transcripts
NM_145236.2 XM_005246953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4092325 c.84T>C p.S28S Chr.2: 231397803-231397803 on Build GRCh38
5354739 c.267C>T p.C89C Chr.2: 231397986-231397986 on Build GRCh38
5396467 c.186G>A p.W62* Chr.2: 231397905-231397905 on Build GRCh38
1245884 c.210G>T p.A70A Chr.2: 231397929-231397929 on Build GRCh38
4001540 c.63C>T p.A21A Chr.2: 231397782-231397782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758379647 Chr.2: 231397836-231397837 on Build GRCh38
rs550844855 Chr.2: 231397908-231397908 on Build GRCh38
rs373965347 Chr.2: 231397769-231397769 on Build GRCh38
rs773378467 Chr.2: 231397782-231397782 on Build GRCh38
rs757539825 Chr.2: 231398071-231398071 on Build GRCh38
rs371049738 Chr.2: 231397788-231397788 on Build GRCh38
rs765053112 Chr.2: 231398062-231398062 on Build GRCh38
rs751148913 Chr.2: 231397824-231397824 on Build GRCh38
rs746003805 Chr.2: 231398013-231398013 on Build GRCh38
rs151118590 Chr.2: 231397818-231397818 on Build GRCh38
rs759443242 Chr.2: 231398060-231398060 on Build GRCh38
rs772860715 Chr.2: 231398040-231398040 on Build GRCh38
rs369256532 Chr.2: 231397962-231397962 on Build GRCh38
rs754292525 Chr.2: 231397716-231397716 on Build GRCh38
rs780846082 Chr.2: 231397758-231397758 on Build GRCh38
rs761345051 Chr.2: 231397911-231397911 on Build GRCh38
rs532531850 Chr.2: 231397863-231397863 on Build GRCh38
rs375999431 Chr.2: 231397928-231397928 on Build GRCh38
rs745834879 Chr.2: 231397835-231397835 on Build GRCh38
rs772021983 Chr.2: 231397777-231397777 on Build GRCh38
rs369747651 Chr.2: 231398073-231398073 on Build GRCh38
rs766174080 Chr.2: 231397787-231397787 on Build GRCh38
rs574145605 Chr.2: 231397665-231397665 on Build GRCh38
rs770008689 Chr.2: 231398014-231398014 on Build GRCh38
rs765051779 Chr.2: 231397708-231397709 on Build GRCh38
rs963916805 Chr.2: 231397670-231397670 on Build GRCh38
rs1038352999 Chr.2: 231397808-231397808 on Build GRCh38
rs565171249 Chr.2: 231397828-231397828 on Build GRCh38
rs376778157 Chr.2: 231397951-231397951 on Build GRCh38
rs750337710 Chr.2: 231397914-231397914 on Build GRCh38
rs770489982 Chr.2: 231397939-231397939 on Build GRCh38
rs750001905 Chr.2: 231397971-231397971 on Build GRCh38
rs994189815 Chr.2: 231397647-231397647 on Build GRCh38
rs754749746 Chr.2: 231397936-231397936 on Build GRCh38
rs10445829 Chr.2: 231397708-231397708 on Build GRCh38
rs201032403 Chr.2: 231397693-231397693 on Build GRCh38
rs1056277149 Chr.2: 231397784-231397784 on Build GRCh38
rs760145688 Chr.2: 231397892-231397892 on Build GRCh38
rs373388615 Chr.2: 231397935-231397935 on Build GRCh38
rs757464144 Chr.2: 231397999-231397999 on Build GRCh38
rs774004851 Chr.2: 231397960-231397960 on Build GRCh38
rs776037524 Chr.2: 231397893-231397893 on Build GRCh38
rs761292125 Chr.2: 231397711-231397711 on Build GRCh38
rs367701147 Chr.2: 231397741-231397741 on Build GRCh38
rs750679692 Chr.2: 231398083-231398083 on Build GRCh38
rs754760232 Chr.2: 231397987-231397987 on Build GRCh38
rs754740686 Chr.2: 231397933-231397933 on Build GRCh38
rs754661716 Chr.2: 231397720-231397720 on Build GRCh38
rs763867229 Chr.2: 231397903-231397903 on Build GRCh38
rs116349534 Chr.2: 231397664-231397664 on Build GRCh38
rs749780489 Chr.2: 231398025-231398025 on Build GRCh38
rs779873331 Chr.2: 231397843-231397843 on Build GRCh38
rs780328214 Chr.2: 231398099-231398099 on Build GRCh38
rs757946281 Chr.2: 231397745-231397745 on Build GRCh38
rs748338089 Chr.2: 231397844-231397844 on Build GRCh38
rs781417639 Chr.2: 231398011-231398011 on Build GRCh38
rs765731280 Chr.2: 231397799-231397799 on Build GRCh38
rs755853136 Chr.2: 231397836-231397836 on Build GRCh38
rs374018323 Chr.2: 231398020-231398020 on Build GRCh38
rs777481184 Chr.2: 231397805-231397805 on Build GRCh38
rs747050205 Chr.2: 231397883-231397883 on Build GRCh38
rs779816208 Chr.2: 231397929-231397929 on Build GRCh38
rs967135781 Chr.2: 231397718-231397718 on Build GRCh38
rs1008479887 Chr.2: 231397864-231397864 on Build GRCh38
rs371847021 Chr.2: 231397794-231397794 on Build GRCh38
rs932132929 Chr.2: 231397679-231397679 on Build GRCh38
rs760108002 Chr.2: 231398044-231398044 on Build GRCh38
rs770918007 Chr.2: 231397886-231397886 on Build GRCh38
rs747832242 Chr.2: 231398038-231398038 on Build GRCh38
rs375194946 Chr.2: 231397816-231397816 on Build GRCh38
rs763320598 Chr.2: 231397953-231397953 on Build GRCh38
rs767791399 Chr.2: 231398072-231398072 on Build GRCh38
rs750315281 Chr.2: 231397757-231397758 on Build GRCh38
rs944106162 Chr.2: 231398061-231398061 on Build GRCh38
rs201156023 Chr.2: 231398039-231398039 on Build GRCh38
rs1018646542 Chr.2: 231397669-231397669 on Build GRCh38
rs116209946 Chr.2: 231397783-231397783 on Build GRCh38
rs200868742 Chr.2: 231397764-231397764 on Build GRCh38
rs916725138 Chr.2: 231397972-231397972 on Build GRCh38
rs376886234 Chr.2: 231397683-231397683 on Build GRCh38
rs781128304 Chr.2: 231397831-231397831 on Build GRCh38
rs199641918 Chr.2: 231397934-231397934 on Build GRCh38
rs372799734 Chr.2: 231397913-231397913 on Build GRCh38
rs765903072 Chr.2: 231398045-231398045 on Build GRCh38
rs778597902 Chr.2: 231397991-231397991 on Build GRCh38
rs762508875 Chr.2: 231397895-231397895 on Build GRCh38
rs776584310 Chr.2: 231398055-231398055 on Build GRCh38
rs769041298 Chr.2: 231398036-231398036 on Build GRCh38
rs766981564 Chr.2: 231397969-231397969 on Build GRCh38
rs775984337 Chr.2: 231397952-231397952 on Build GRCh38
rs765135137 Chr.2: 231397977-231397977 on Build GRCh38
rs757259930 Chr.2: 231397748-231397748 on Build GRCh38
rs764922477 Chr.2: 231397740-231397740 on Build GRCh38
rs375071671 Chr.2: 231397979-231397979 on Build GRCh38
rs544015296 Chr.2: 231397650-231397650 on Build GRCh38
rs752505664 Chr.2: 231398066-231398066 on Build GRCh38
rs377346766 Chr.2: 231398037-231398037 on Build GRCh38
rs753853523 Chr.2: 231397860-231397860 on Build GRCh38
rs755502756 Chr.2: 231398100-231398100 on Build GRCh38
rs746899567 Chr.2: 231397747-231397747 on Build GRCh38
rs200329884 Chr.2: 231397930-231397930 on Build GRCh38
rs769579473 Chr.2: 231397768-231397768 on Build GRCh38
rs751787225 Chr.2: 231397994-231397994 on Build GRCh38
rs992751419 Chr.2: 231397968-231397968 on Build GRCh38
rs755689972 Chr.2: 231397662-231397663 on Build GRCh38
rs566644892 Chr.2: 231397974-231397974 on Build GRCh38
rs748391272 Chr.2: 231397776-231397776 on Build GRCh38
rs756224089 Chr.2: 231398091-231398091 on Build GRCh38
rs887620920 Chr.2: 231397819-231397819 on Build GRCh38
rs766243017 Chr.2: 231397976-231397976 on Build GRCh38
rs768908837 Chr.2: 231397956-231397956 on Build GRCh38
rs373568057 Chr.2: 231397715-231397715 on Build GRCh38
rs777240589 Chr.2: 231397887-231397887 on Build GRCh38

View Full Product Details