Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231399545-231400075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT7
Entrez Gene ID
93010
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7
Gene Aliases
beta3GnT7
UniGene
Hs.299329
Gene Chromosome Location
Chr.2: 231395624-231401164 on Build GRCh38
Transcripts
NM_145236.2 XM_005246953.1

SNP Detail

SNP ID Chromosome Location
rs934145414 Chr.2: 231399721-231399721 on Build GRCh38
rs10445831 Chr.2: 231399673-231399673 on Build GRCh38
rs1053813769 Chr.2: 231399833-231399833 on Build GRCh38
rs1001601522 Chr.2: 231399846-231399846 on Build GRCh38
rs1014074526 Chr.2: 231399620-231399620 on Build GRCh38
rs951896758 Chr.2: 231399772-231399772 on Build GRCh38
rs892043575 Chr.2: 231399835-231399835 on Build GRCh38
rs891599189 Chr.2: 231399613-231399613 on Build GRCh38
rs754875590 Chr.2: 231399649-231399649 on Build GRCh38
rs114509189 Chr.2: 231399603-231399603 on Build GRCh38
rs543165436 Chr.2: 231399804-231399804 on Build GRCh38
rs749301912 Chr.2: 231399610-231399610 on Build GRCh38
rs1004717176 Chr.2: 231400018-231400018 on Build GRCh38
rs184575561 Chr.2: 231399845-231399845 on Build GRCh38
rs1024380645 Chr.2: 231399739-231399739 on Build GRCh38
rs911372380 Chr.2: 231399955-231399955 on Build GRCh38
rs553846695 Chr.2: 231399672-231399672 on Build GRCh38
rs1053355432 Chr.2: 231399634-231399634 on Build GRCh38
rs931691561 Chr.2: 231399825-231399825 on Build GRCh38
rs1001330429 Chr.2: 231399762-231399762 on Build GRCh38
rs565513632 Chr.2: 231400040-231400040 on Build GRCh38
rs149249587 Chr.2: 231399840-231399840 on Build GRCh38
rs1014246712 Chr.2: 231399693-231399693 on Build GRCh38
rs918850377 Chr.2: 231399697-231399697 on Build GRCh38
rs955638260 Chr.2: 231399935-231399935 on Build GRCh38
rs1027423798 Chr.2: 231399764-231399764 on Build GRCh38
rs935678847 Chr.2: 231399805-231399805 on Build GRCh38
rs966141931 Chr.2: 231399788-231399788 on Build GRCh38
rs187266314 Chr.2: 231399830-231399830 on Build GRCh38
rs980496081 Chr.2: 231399661-231399661 on Build GRCh38
rs1015258717 Chr.2: 231400022-231400022 on Build GRCh38
rs965955879 Chr.2: 231400039-231400039 on Build GRCh38
rs985643906 Chr.2: 231399826-231399826 on Build GRCh38
rs970074612 Chr.2: 231399751-231399751 on Build GRCh38
rs904202036 Chr.2: 231399571-231399571 on Build GRCh38
rs972941014 Chr.2: 231399678-231399678 on Build GRCh38
rs964563757 Chr.2: 231399971-231399971 on Build GRCh38
rs970189468 Chr.2: 231399652-231399652 on Build GRCh38
rs184285401 Chr.2: 231399789-231399789 on Build GRCh38
rs949940548 Chr.2: 231400044-231400044 on Build GRCh38
rs1017436771 Chr.2: 231399847-231399847 on Build GRCh38
rs1000831213 Chr.2: 231399628-231399628 on Build GRCh38
rs191783103 Chr.2: 231399832-231399832 on Build GRCh38
rs763212665 Chr.2: 231399980-231399980 on Build GRCh38
rs547734386 Chr.2: 231399774-231399774 on Build GRCh38
rs962668344 Chr.2: 231399867-231399867 on Build GRCh38
rs911618606 Chr.2: 231399757-231399757 on Build GRCh38
rs925730480 Chr.2: 231399997-231399997 on Build GRCh38
rs935664131 Chr.2: 231400002-231400002 on Build GRCh38
rs1001598614 Chr.2: 231399902-231399902 on Build GRCh38
rs759961028 Chr.2: 231400024-231400024 on Build GRCh38
rs987064489 Chr.2: 231399945-231399945 on Build GRCh38
rs905976500 Chr.2: 231399869-231399869 on Build GRCh38
rs994210180 Chr.2: 231399912-231399912 on Build GRCh38
rs907779600 Chr.2: 231399775-231399775 on Build GRCh38
rs189606183 Chr.2: 231399918-231399918 on Build GRCh38
rs1039214025 Chr.2: 231399780-231399780 on Build GRCh38
rs921688917 Chr.2: 231399815-231399815 on Build GRCh38

View Full Product Details