Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 236214481-236214985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB18
Entrez Gene ID
401036
Gene Name
ankyrin repeat and SOCS box containing 18
Gene Aliases
ASB-18
UniGene
Hs.639909
Gene Chromosome Location
Chr.2: 236194872-236264345 on Build GRCh38
Transcripts
NM_212556.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4790490 c.722G>T p.R241L Chr.2: 236214741-236214741 on Build GRCh38
4803361 c.941G>T p.R314L Chr.2: 236214522-236214522 on Build GRCh38
6019953 c.680C>T p.A227V Chr.2: 236214783-236214783 on Build GRCh38
4128225 c.831G>A p.A277A Chr.2: 236214632-236214632 on Build GRCh38
6038403 c.889C>T p.L297L Chr.2: 236214574-236214574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915740305 Chr.2: 236214867-236214867 on Build GRCh38
rs771803750 Chr.2: 236214515-236214515 on Build GRCh38
rs986815021 Chr.2: 236214649-236214649 on Build GRCh38
rs115967613 Chr.2: 236214877-236214877 on Build GRCh38
rs568492759 Chr.2: 236214611-236214611 on Build GRCh38
rs969395881 Chr.2: 236214917-236214917 on Build GRCh38
rs1057037261 Chr.2: 236214518-236214518 on Build GRCh38
rs889251051 Chr.2: 236214911-236214911 on Build GRCh38
rs763458414 Chr.2: 236214711-236214711 on Build GRCh38
rs6431437 Chr.2: 236214682-236214682 on Build GRCh38
rs1030515431 Chr.2: 236214634-236214634 on Build GRCh38
rs894411374 Chr.2: 236214522-236214522 on Build GRCh38
rs751662212 Chr.2: 236214795-236214795 on Build GRCh38
rs1012564677 Chr.2: 236214525-236214525 on Build GRCh38
rs555686516 Chr.2: 236214894-236214894 on Build GRCh38
rs1004290536 Chr.2: 236214775-236214775 on Build GRCh38
rs147888546 Chr.2: 236214598-236214598 on Build GRCh38
rs548914403 Chr.2: 236214575-236214575 on Build GRCh38
rs1030441984 Chr.2: 236214820-236214820 on Build GRCh38
rs968713937 Chr.2: 236214830-236214830 on Build GRCh38
rs868198216 Chr.2: 236214512-236214512 on Build GRCh38
rs533836717 Chr.2: 236214892-236214892 on Build GRCh38
rs1003961132 Chr.2: 236214610-236214610 on Build GRCh38
rs536010190 Chr.2: 236214878-236214878 on Build GRCh38
rs776825057 Chr.2: 236214526-236214526 on Build GRCh38
rs960062838 Chr.2: 236214951-236214951 on Build GRCh38
rs1008147595 Chr.2: 236214912-236214912 on Build GRCh38
rs888119390 Chr.2: 236214766-236214766 on Build GRCh38
rs13427406 Chr.2: 236214574-236214574 on Build GRCh38
rs760008402 Chr.2: 236214565-236214565 on Build GRCh38
rs751268528 Chr.2: 236214888-236214888 on Build GRCh38
rs540744027 Chr.2: 236214945-236214945 on Build GRCh38
rs540112321 Chr.2: 236214821-236214821 on Build GRCh38
rs1015800358 Chr.2: 236214616-236214616 on Build GRCh38
rs1015310630 Chr.2: 236214777-236214777 on Build GRCh38
rs900947410 Chr.2: 236214790-236214790 on Build GRCh38
rs1048828138 Chr.2: 236214754-236214754 on Build GRCh38
rs948652878 Chr.2: 236214753-236214753 on Build GRCh38
rs947414441 Chr.2: 236214677-236214677 on Build GRCh38
rs888139351 Chr.2: 236214591-236214591 on Build GRCh38
rs1049506666 Chr.2: 236214907-236214907 on Build GRCh38
rs969277285 Chr.2: 236214627-236214645 on Build GRCh38
rs563334671 Chr.2: 236214501-236214501 on Build GRCh38
rs570889823 Chr.2: 236214817-236214817 on Build GRCh38
rs953610373 Chr.2: 236214646-236214646 on Build GRCh38
rs547631783 Chr.2: 236214913-236214913 on Build GRCh38
rs938468414 Chr.2: 236214739-236214739 on Build GRCh38
rs778596701 Chr.2: 236214503-236214503 on Build GRCh38
rs529127050 Chr.2: 236214573-236214573 on Build GRCh38
rs749251191 Chr.2: 236214500-236214500 on Build GRCh38
rs747751719 Chr.2: 236214513-236214513 on Build GRCh38
rs939677572 Chr.2: 236214881-236214881 on Build GRCh38
rs374319025 Chr.2: 236214841-236214841 on Build GRCh38
rs999141653 Chr.2: 236214925-236214925 on Build GRCh38
rs992941985 Chr.2: 236214849-236214849 on Build GRCh38
rs914571361 Chr.2: 236214665-236214665 on Build GRCh38
rs900873712 Chr.2: 236214893-236214893 on Build GRCh38
rs1057518421 Chr.2: 236214769-236214769 on Build GRCh38
rs768212844 Chr.2: 236214502-236214502 on Build GRCh38
rs960093731 Chr.2: 236214846-236214846 on Build GRCh38
rs56247246 Chr.2: 236214741-236214741 on Build GRCh38
rs1022297168 Chr.2: 236214824-236214824 on Build GRCh38
rs976233442 Chr.2: 236214631-236214631 on Build GRCh38

View Full Product Details