Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240871229-240871727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGXT
Entrez Gene ID
189
Gene Name
alanine-glyoxylate aminotransferase
Gene Aliases
AGT, AGT1, AGXT1, PH1, SPAT, SPT, TLH6
UniGene
Hs.144567
Gene Chromosome Location
Chr.2: 240868745-240879119 on Build GRCh38
Transcripts
NM_000030.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244655 c.466G>A p.G156R Chr.2: 240871391-240871391 on Build GRCh38
6840786 c.467G>T p.G156V Chr.2: 240871392-240871392 on Build GRCh38
1018787 c.451C>T p.L151L Chr.2: 240871376-240871376 on Build GRCh38
5993816 c.486G>A p.V162V Chr.2: 240871411-240871411 on Build GRCh38
5396644 c.468G>A p.G156G Chr.2: 240871393-240871393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34572785 Chr.2: 240871606-240871606 on Build GRCh38
rs538338287 Chr.2: 240871583-240871583 on Build GRCh38
rs965707125 Chr.2: 240871435-240871435 on Build GRCh38
rs771809604 Chr.2: 240871363-240871363 on Build GRCh38
rs180177224 Chr.2: 240871385-240871385 on Build GRCh38
rs919786533 Chr.2: 240871546-240871546 on Build GRCh38
rs1054324964 Chr.2: 240871645-240871645 on Build GRCh38
rs1051315576 Chr.2: 240871254-240871254 on Build GRCh38
rs199879522 Chr.2: 240871468-240871468 on Build GRCh38
rs398122323 Chr.2: 240871337-240871338 on Build GRCh38
rs965407367 Chr.2: 240871296-240871296 on Build GRCh38
rs575497900 Chr.2: 240871294-240871294 on Build GRCh38
rs77667245 Chr.2: 240871567-240871567 on Build GRCh38
rs891120250 Chr.2: 240871285-240871285 on Build GRCh38
rs747774501 Chr.2: 240871360-240871360 on Build GRCh38
rs368587594 Chr.2: 240871308-240871308 on Build GRCh38
rs906515171 Chr.2: 240871668-240871668 on Build GRCh38
rs556634120 Chr.2: 240871663-240871663 on Build GRCh38
rs773166371 Chr.2: 240871405-240871405 on Build GRCh38
rs180177220 Chr.2: 240871370-240871370 on Build GRCh38
rs74895925 Chr.2: 240871333-240871333 on Build GRCh38
rs938286356 Chr.2: 240871572-240871572 on Build GRCh38
rs767782309 Chr.2: 240871488-240871488 on Build GRCh38
rs778832054 Chr.2: 240871427-240871427 on Build GRCh38
rs121908529 Chr.2: 240871433-240871433 on Build GRCh38
rs180177230 Chr.2: 240871422-240871422 on Build GRCh38
rs180177232 Chr.2: 240871444-240871444 on Build GRCh38
rs375123832 Chr.2: 240871354-240871354 on Build GRCh38
rs910879087 Chr.2: 240871575-240871575 on Build GRCh38
rs562139903 Chr.2: 240871464-240871464 on Build GRCh38
rs935615488 Chr.2: 240871604-240871604 on Build GRCh38
rs180177225 Chr.2: 240871398-240871398 on Build GRCh38
rs369523966 Chr.2: 240871345-240871345 on Build GRCh38
rs117045831 Chr.2: 240871269-240871269 on Build GRCh38
rs770462933 Chr.2: 240871448-240871448 on Build GRCh38
rs375481302 Chr.2: 240871332-240871332 on Build GRCh38
rs908604662 Chr.2: 240871519-240871519 on Build GRCh38
rs999556034 Chr.2: 240871370-240871370 on Build GRCh38
rs769101845 Chr.2: 240871469-240871469 on Build GRCh38
rs894195873 Chr.2: 240871258-240871258 on Build GRCh38
rs924223583 Chr.2: 240871602-240871602 on Build GRCh38
rs180177219 Chr.2: 240871347-240871347 on Build GRCh38
rs758695568 Chr.2: 240871302-240871302 on Build GRCh38
rs564788107 Chr.2: 240871667-240871667 on Build GRCh38
rs73106676 Chr.2: 240871320-240871320 on Build GRCh38
rs1012738802 Chr.2: 240871664-240871664 on Build GRCh38
rs746656296 Chr.2: 240871445-240871445 on Build GRCh38
rs970754032 Chr.2: 240871510-240871510 on Build GRCh38
rs1035634115 Chr.2: 240871523-240871523 on Build GRCh38
rs1021436520 Chr.2: 240871272-240871272 on Build GRCh38
rs552061241 Chr.2: 240871473-240871473 on Build GRCh38
rs772766803 Chr.2: 240871378-240871378 on Build GRCh38
rs180177231 Chr.2: 240871443-240871443 on Build GRCh38
rs543832032 Chr.2: 240871421-240871421 on Build GRCh38
rs752385061 Chr.2: 240871375-240871375 on Build GRCh38
rs1012336403 Chr.2: 240871503-240871503 on Build GRCh38
rs910890174 Chr.2: 240871460-240871460 on Build GRCh38
rs121908530 Chr.2: 240871391-240871391 on Build GRCh38
rs781107643 Chr.2: 240871321-240871321 on Build GRCh38
rs886055839 Chr.2: 240871452-240871452 on Build GRCh38
rs180177223 Chr.2: 240871382-240871382 on Build GRCh38
rs1053971057 Chr.2: 240871500-240871500 on Build GRCh38
rs765812098 Chr.2: 240871380-240871380 on Build GRCh38
rs753234546 Chr.2: 240871301-240871301 on Build GRCh38
rs556577436 Chr.2: 240871288-240871288 on Build GRCh38
rs770992934 Chr.2: 240871455-240871455 on Build GRCh38
rs751768516 Chr.2: 240871318-240871318 on Build GRCh38
rs200921230 Chr.2: 240871326-240871326 on Build GRCh38
rs121908524 Chr.2: 240871379-240871379 on Build GRCh38
rs1041263627 Chr.2: 240871589-240871589 on Build GRCh38
rs80015252 Chr.2: 240871501-240871501 on Build GRCh38
rs961189534 Chr.2: 240871532-240871532 on Build GRCh38
rs757583201 Chr.2: 240871408-240871408 on Build GRCh38
rs915590413 Chr.2: 240871629-240871629 on Build GRCh38
rs762170458 Chr.2: 240871474-240871474 on Build GRCh38
rs147601535 Chr.2: 240871414-240871414 on Build GRCh38
rs763316488 Chr.2: 240871545-240871545 on Build GRCh38
rs149569747 Chr.2: 240871390-240871390 on Build GRCh38
rs749247113 Chr.2: 240871336-240871336 on Build GRCh38
rs147497484 Chr.2: 240871409-240871409 on Build GRCh38
rs180177222 Chr.2: 240871374-240871374 on Build GRCh38
rs538533341 Chr.2: 240871277-240871277 on Build GRCh38
rs981364665 Chr.2: 240871399-240871399 on Build GRCh38
rs947030049 Chr.2: 240871651-240871651 on Build GRCh38
rs180177221 Chr.2: 240871372-240871379 on Build GRCh38
rs369031110 Chr.2: 240871516-240871516 on Build GRCh38
rs180177233 Chr.2: 240871444-240871445 on Build GRCh38
rs984219480 Chr.2: 240871507-240871507 on Build GRCh38
rs567006601 Chr.2: 240871496-240871496 on Build GRCh38
rs902894652 Chr.2: 240871680-240871680 on Build GRCh38
rs982656318 Chr.2: 240871560-240871560 on Build GRCh38
rs180177227 Chr.2: 240871406-240871406 on Build GRCh38
rs528938116 Chr.2: 240871416-240871416 on Build GRCh38
rs10196315 Chr.2: 240871540-240871540 on Build GRCh38
rs764404787 Chr.2: 240871689-240871689 on Build GRCh38

View Full Product Details