Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241686743-241687263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTYMK
Entrez Gene ID
1841
Gene Name
deoxythymidylate kinase
Gene Aliases
CDC8, PP3731, TMPK, TYMK
UniGene
Hs.471873
Gene Chromosome Location
Chr.2: 241675742-241686968 on Build GRCh38
Transcripts
NM_001165031.1 NM_001320902.1 NM_001320903.1 NM_001320904.1 NM_001320905.1 NM_012145.3 NR_033255.1 NR_135492.1 XM_017005100.1 XM_017005101.1

Gene Symbol
ING5
Entrez Gene ID
84289
Gene Name
inhibitor of growth family member 5
Gene Aliases
p28ING5
UniGene
Hs.529172
Gene Chromosome Location
Chr.2: 241687017-241729481 on Build GRCh38
Transcripts
NM_001165031.1 NM_001320902.1 NM_001320903.1 NM_001320904.1 NM_001320905.1 NM_012145.3 NR_033255.1 NR_135492.1 XM_017005100.1 XM_017005101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157797 c.6G>T p.A2A Chr.2: 241686778-241686778 on Build GRCh38
3771843 c.19G>T p.A7S Chr.2: 241686765-241686765 on Build GRCh38
4596823 c.16G>A p.G6R Chr.2: 241686768-241686768 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs920934271 Chr.2: 241687192-241687192 on Build GRCh38
rs35925884 Chr.2: 241687064-241687064 on Build GRCh38
rs1022442505 Chr.2: 241687011-241687011 on Build GRCh38
rs763538564 Chr.2: 241686768-241686768 on Build GRCh38
rs557840238 Chr.2: 241687113-241687113 on Build GRCh38
rs562230284 Chr.2: 241686858-241686858 on Build GRCh38
rs982454940 Chr.2: 241686947-241686947 on Build GRCh38
rs1029642359 Chr.2: 241687246-241687246 on Build GRCh38
rs890585857 Chr.2: 241686826-241686826 on Build GRCh38
rs540945389 Chr.2: 241686845-241686845 on Build GRCh38
rs777297560 Chr.2: 241686805-241686805 on Build GRCh38
rs1050950970 Chr.2: 241687023-241687023 on Build GRCh38
rs921887682 Chr.2: 241686861-241686861 on Build GRCh38
rs1056137798 Chr.2: 241686973-241686973 on Build GRCh38
rs540531352 Chr.2: 241687238-241687238 on Build GRCh38
rs529523110 Chr.2: 241686928-241686928 on Build GRCh38
rs971512862 Chr.2: 241687082-241687082 on Build GRCh38
rs767655288 Chr.2: 241686779-241686779 on Build GRCh38
rs953491157 Chr.2: 241687060-241687060 on Build GRCh38
rs771863067 Chr.2: 241686802-241686802 on Build GRCh38
rs551071652 Chr.2: 241686998-241686998 on Build GRCh38
rs780049385 Chr.2: 241686787-241686787 on Build GRCh38
rs755123869 Chr.2: 241686900-241686900 on Build GRCh38
rs1022472985 Chr.2: 241686796-241686796 on Build GRCh38
rs959247108 Chr.2: 241687058-241687058 on Build GRCh38
rs753839699 Chr.2: 241686789-241686789 on Build GRCh38
rs1033011125 Chr.2: 241687050-241687050 on Build GRCh38
rs1041800898 Chr.2: 241687032-241687032 on Build GRCh38
rs932584964 Chr.2: 241687019-241687019 on Build GRCh38
rs939238761 Chr.2: 241686968-241686968 on Build GRCh38
rs763591671 Chr.2: 241686821-241686821 on Build GRCh38
rs947706579 Chr.2: 241687145-241687145 on Build GRCh38
rs769019188 Chr.2: 241686823-241686823 on Build GRCh38
rs1052622322 Chr.2: 241687226-241687226 on Build GRCh38
rs939303360 Chr.2: 241687123-241687123 on Build GRCh38
rs756190575 Chr.2: 241686786-241686786 on Build GRCh38
rs750602620 Chr.2: 241686785-241686785 on Build GRCh38
rs941180071 Chr.2: 241686936-241686936 on Build GRCh38
rs567436297 Chr.2: 241687037-241687037 on Build GRCh38
rs1033707241 Chr.2: 241686879-241686879 on Build GRCh38
rs1024428171 Chr.2: 241687081-241687081 on Build GRCh38
rs879530696 Chr.2: 241687157-241687157 on Build GRCh38
rs921205575 Chr.2: 241687016-241687016 on Build GRCh38
rs566489435 Chr.2: 241686999-241686999 on Build GRCh38
rs917407522 Chr.2: 241686931-241686931 on Build GRCh38
rs573205676 Chr.2: 241687225-241687225 on Build GRCh38
rs1000472141 Chr.2: 241686872-241686872 on Build GRCh38
rs1029009130 Chr.2: 241687057-241687057 on Build GRCh38
rs778819243 Chr.2: 241686792-241686792 on Build GRCh38
rs754840879 Chr.2: 241686790-241686790 on Build GRCh38
rs909747223 Chr.2: 241686915-241686915 on Build GRCh38
rs766543742 Chr.2: 241686776-241686777 on Build GRCh38
rs567080285 Chr.2: 241687038-241687038 on Build GRCh38
rs138371310 Chr.2: 241687025-241687025 on Build GRCh38
rs774870983 Chr.2: 241686834-241686834 on Build GRCh38
rs34081371 Chr.2: 241687101-241687101 on Build GRCh38
rs968194268 Chr.2: 241686814-241686814 on Build GRCh38
rs932569114 Chr.2: 241687196-241687196 on Build GRCh38
rs555987144 Chr.2: 241687072-241687072 on Build GRCh38
rs1056769944 Chr.2: 241687127-241687127 on Build GRCh38
rs904059305 Chr.2: 241687024-241687024 on Build GRCh38
rs7596968 Chr.2: 241687083-241687083 on Build GRCh38
rs999236480 Chr.2: 241687203-241687203 on Build GRCh38
rs985252114 Chr.2: 241686909-241686909 on Build GRCh38
rs564539865 Chr.2: 241686807-241686807 on Build GRCh38
rs778308266 Chr.2: 241687100-241687100 on Build GRCh38
rs1036949568 Chr.2: 241686949-241686949 on Build GRCh38
rs867302139 Chr.2: 241687144-241687144 on Build GRCh38
rs974186656 Chr.2: 241687152-241687152 on Build GRCh38
rs966284367 Chr.2: 241687036-241687036 on Build GRCh38
rs550650577 Chr.2: 241686964-241686964 on Build GRCh38
rs747919089 Chr.2: 241686800-241686800 on Build GRCh38
rs770620579 Chr.2: 241686808-241686808 on Build GRCh38
rs1012373058 Chr.2: 241686794-241686794 on Build GRCh38
rs776272909 Chr.2: 241686819-241686819 on Build GRCh38
rs1031141280 Chr.2: 241687034-241687034 on Build GRCh38
rs770502073 Chr.2: 241686764-241686764 on Build GRCh38
rs751834968 Chr.2: 241686775-241686775 on Build GRCh38
rs1050959696 Chr.2: 241687210-241687210 on Build GRCh38
rs932102555 Chr.2: 241686868-241686868 on Build GRCh38
rs997808197 Chr.2: 241687243-241687243 on Build GRCh38
rs533104086 Chr.2: 241686995-241686995 on Build GRCh38
rs992106349 Chr.2: 241686923-241686923 on Build GRCh38
rs978360754 Chr.2: 241686841-241686841 on Build GRCh38
rs762141486 Chr.2: 241686778-241686778 on Build GRCh38
rs376486491 Chr.2: 241686766-241686766 on Build GRCh38
rs568973396 Chr.2: 241686967-241686967 on Build GRCh38
rs764583881 Chr.2: 241686773-241686773 on Build GRCh38
rs528759558 Chr.2: 241686843-241686843 on Build GRCh38
rs34214302 Chr.2: 241687007-241687007 on Build GRCh38
rs903613062 Chr.2: 241687186-241687186 on Build GRCh38
rs748494708 Chr.2: 241687041-241687041 on Build GRCh38

View Full Product Details