Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 95865-96397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB125
Entrez Gene ID
245938
Gene Name
defensin beta 125
Gene Aliases
DEFB-25
UniGene
Hs.380220
Gene Chromosome Location
Chr.20: 87672-96573 on Build GRCh38
Transcripts
NM_153325.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3549333 c.425C>T p.S142F Chr.20: 96371-96371 on Build GRCh38
6533028 c.276A>C p.P92P Chr.20: 96222-96222 on Build GRCh38
3673168 c.394A>T p.T132S Chr.20: 96340-96340 on Build GRCh38
3549329 c.200G>A p.R67Q Chr.20: 96146-96146 on Build GRCh38
6094021 c.241G>T p.D81Y Chr.20: 96187-96187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747592085 Chr.20: 96319-96319 on Build GRCh38
rs774994048 Chr.20: 96304-96304 on Build GRCh38
rs751532285 Chr.20: 96075-96075 on Build GRCh38
rs765443181 Chr.20: 96260-96260 on Build GRCh38
rs960455369 Chr.20: 96127-96127 on Build GRCh38
rs200870238 Chr.20: 96149-96149 on Build GRCh38
rs754005070 Chr.20: 96142-96142 on Build GRCh38
rs144284374 Chr.20: 96274-96274 on Build GRCh38
rs925218716 Chr.20: 96255-96255 on Build GRCh38
rs148698629 Chr.20: 96125-96125 on Build GRCh38
rs565892940 Chr.20: 96103-96103 on Build GRCh38
rs980471437 Chr.20: 95914-95914 on Build GRCh38
rs564645511 Chr.20: 95892-95892 on Build GRCh38
rs778407594 Chr.20: 96318-96318 on Build GRCh38
rs772464690 Chr.20: 96115-96115 on Build GRCh38
rs537226093 Chr.20: 96261-96261 on Build GRCh38
rs774092161 Chr.20: 96069-96069 on Build GRCh38
rs763390419 Chr.20: 96029-96029 on Build GRCh38
rs375564330 Chr.20: 96197-96197 on Build GRCh38
rs778362608 Chr.20: 96179-96179 on Build GRCh38
rs578173037 Chr.20: 96257-96257 on Build GRCh38
rs202219607 Chr.20: 96085-96085 on Build GRCh38
rs758251612 Chr.20: 96001-96001 on Build GRCh38
rs750692266 Chr.20: 96052-96052 on Build GRCh38
rs969665579 Chr.20: 96240-96240 on Build GRCh38
rs890000950 Chr.20: 96078-96078 on Build GRCh38
rs762285182 Chr.20: 96108-96108 on Build GRCh38
rs376536497 Chr.20: 95999-95999 on Build GRCh38
rs745496891 Chr.20: 96017-96017 on Build GRCh38
rs771370558 Chr.20: 96342-96342 on Build GRCh38
rs371782324 Chr.20: 95988-95988 on Build GRCh38
rs760912688 Chr.20: 96056-96056 on Build GRCh38
rs139650748 Chr.20: 96155-96155 on Build GRCh38
rs140898692 Chr.20: 96337-96337 on Build GRCh38
rs767658093 Chr.20: 95982-95982 on Build GRCh38
rs564320474 Chr.20: 96013-96013 on Build GRCh38
rs748878197 Chr.20: 96174-96174 on Build GRCh38
rs146549701 Chr.20: 96293-96293 on Build GRCh38
rs992117781 Chr.20: 96277-96277 on Build GRCh38
rs750637651 Chr.20: 95984-95984 on Build GRCh38
rs187581419 Chr.20: 95994-95994 on Build GRCh38
rs532933005 Chr.20: 96081-96081 on Build GRCh38
rs370092526 Chr.20: 96062-96062 on Build GRCh38
rs748762744 Chr.20: 96095-96095 on Build GRCh38
rs773518041 Chr.20: 96121-96121 on Build GRCh38
rs771807843 Chr.20: 96143-96143 on Build GRCh38
rs767492858 Chr.20: 96271-96271 on Build GRCh38
rs750171266 Chr.20: 96162-96162 on Build GRCh38
rs748066968 Chr.20: 95991-95991 on Build GRCh38
rs751870543 Chr.20: 95978-95978 on Build GRCh38
rs778648489 Chr.20: 96112-96112 on Build GRCh38
rs775021767 Chr.20: 96020-96020 on Build GRCh38
rs561362746 Chr.20: 96009-96009 on Build GRCh38
rs768046229 Chr.20: 96096-96096 on Build GRCh38
rs139068554 Chr.20: 96279-96279 on Build GRCh38
rs867765515 Chr.20: 96219-96219 on Build GRCh38
rs766063826 Chr.20: 96285-96285 on Build GRCh38
rs116934569 Chr.20: 96059-96059 on Build GRCh38
rs369692500 Chr.20: 96243-96243 on Build GRCh38
rs45528036 Chr.20: 96372-96372 on Build GRCh38
rs762230159 Chr.20: 96050-96050 on Build GRCh38
rs756853525 Chr.20: 96297-96297 on Build GRCh38
rs868868710 Chr.20: 96023-96023 on Build GRCh38
rs767854746 Chr.20: 96051-96051 on Build GRCh38
rs151148014 Chr.20: 96145-96145 on Build GRCh38
rs769435585 Chr.20: 96094-96094 on Build GRCh38
rs769223826 Chr.20: 96019-96019 on Build GRCh38
rs373770106 Chr.20: 96083-96083 on Build GRCh38
rs911393706 Chr.20: 96364-96364 on Build GRCh38
rs369118073 Chr.20: 95959-95959 on Build GRCh38
rs374363647 Chr.20: 96338-96338 on Build GRCh38
rs773571509 Chr.20: 96229-96229 on Build GRCh38
rs145802690 Chr.20: 96048-96048 on Build GRCh38
rs756206352 Chr.20: 95987-95987 on Build GRCh38
rs558280500 Chr.20: 96245-96245 on Build GRCh38
rs751058215 Chr.20: 96294-96329 on Build GRCh38
rs948002945 Chr.20: 95894-95894 on Build GRCh38
rs766392612 Chr.20: 96058-96058 on Build GRCh38
rs764394071 Chr.20: 95968-95968 on Build GRCh38
rs937361143 Chr.20: 95951-95952 on Build GRCh38
rs745543814 Chr.20: 96093-96093 on Build GRCh38
rs776824971 Chr.20: 96367-96367 on Build GRCh38
rs757216800 Chr.20: 96146-96146 on Build GRCh38
rs917355535 Chr.20: 95954-95954 on Build GRCh38
rs768203979 Chr.20: 96177-96177 on Build GRCh38
rs781107020 Chr.20: 96092-96092 on Build GRCh38
rs755723306 Chr.20: 96163-96163 on Build GRCh38
rs6111385 Chr.20: 96321-96321 on Build GRCh38
rs993824024 Chr.20: 96098-96098 on Build GRCh38
rs991503469 Chr.20: 95953-95957 on Build GRCh38
rs748677322 Chr.20: 96183-96183 on Build GRCh38
rs761171088 Chr.20: 96232-96232 on Build GRCh38
rs201575332 Chr.20: 96049-96049 on Build GRCh38
rs200259658 Chr.20: 96336-96336 on Build GRCh38
rs926211300 Chr.20: 95917-95917 on Build GRCh38
rs779821283 Chr.20: 96169-96169 on Build GRCh38
rs147882888 Chr.20: 96166-96166 on Build GRCh38
rs41276376 Chr.20: 96130-96130 on Build GRCh38
rs770080919 Chr.20: 96373-96373 on Build GRCh38
rs753566285 Chr.20: 96294-96294 on Build GRCh38

View Full Product Details