Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10038483-10039032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKEF1
Entrez Gene ID
63926
Gene Name
ankyrin repeat and EF-hand domain containing 1
Gene Aliases
ANKRD5
UniGene
Hs.70903
Gene Chromosome Location
Chr.20: 10034999-10056764 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Gene Symbol
SNAP25-AS1
Entrez Gene ID
100131208
Gene Name
SNAP25 antisense RNA 1
Gene Aliases
-
UniGene
Hs.658178
Gene Chromosome Location
Chr.20: 10023812-10219506 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1637694 c.194G>A p.S65N Chr.20: 10038495-10038495 on Build GRCh38
1182940 c.280G>T p.E94* Chr.20: 10038581-10038581 on Build GRCh38
6991056 c.319A>G p.M107V Chr.20: 10038620-10038620 on Build GRCh38
3544024 c.234C>T p.D78D Chr.20: 10038535-10038535 on Build GRCh38
3840378 c.199C>T p.L67F Chr.20: 10038500-10038500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977128097 Chr.20: 10038513-10038513 on Build GRCh38
rs751281479 Chr.20: 10038530-10038530 on Build GRCh38
rs931979340 Chr.20: 10038970-10038970 on Build GRCh38
rs893847928 Chr.20: 10038952-10038952 on Build GRCh38
rs757413606 Chr.20: 10038720-10038720 on Build GRCh38
rs1031402548 Chr.20: 10038955-10038955 on Build GRCh38
rs758934091 Chr.20: 10038578-10038578 on Build GRCh38
rs71334885 Chr.20: 10038838-10038838 on Build GRCh38
rs971017478 Chr.20: 10038905-10038907 on Build GRCh38
rs531381069 Chr.20: 10038873-10038873 on Build GRCh38
rs766210721 Chr.20: 10038659-10038659 on Build GRCh38
rs760280062 Chr.20: 10038658-10038658 on Build GRCh38
rs991397831 Chr.20: 10038799-10038799 on Build GRCh38
rs749901789 Chr.20: 10038654-10038654 on Build GRCh38
rs6087109 Chr.20: 10038761-10038761 on Build GRCh38
rs929313119 Chr.20: 10038667-10038667 on Build GRCh38
rs747235271 Chr.20: 10038557-10038557 on Build GRCh38
rs776126169 Chr.20: 10038607-10038607 on Build GRCh38
rs763641219 Chr.20: 10038529-10038529 on Build GRCh38
rs1040368815 Chr.20: 10038602-10038602 on Build GRCh38
rs144446604 Chr.20: 10038830-10038830 on Build GRCh38
rs1029159051 Chr.20: 10038630-10038630 on Build GRCh38
rs989986380 Chr.20: 10038998-10038998 on Build GRCh38
rs973079694 Chr.20: 10038752-10038752 on Build GRCh38
rs754856631 Chr.20: 10038555-10038555 on Build GRCh38
rs139004770 Chr.20: 10038626-10038626 on Build GRCh38
rs539927439 Chr.20: 10038621-10038621 on Build GRCh38
rs773935316 Chr.20: 10038636-10038636 on Build GRCh38
rs548784313 Chr.20: 10038754-10038754 on Build GRCh38
rs766973371 Chr.20: 10038648-10038648 on Build GRCh38
rs745989495 Chr.20: 10038584-10038584 on Build GRCh38
rs747180684 Chr.20: 10038585-10038586 on Build GRCh38
rs368975724 Chr.20: 10038618-10038618 on Build GRCh38
rs949846094 Chr.20: 10038860-10038860 on Build GRCh38
rs767028510 Chr.20: 10038537-10038537 on Build GRCh38
rs200004813 Chr.20: 10038670-10038670 on Build GRCh38
rs962114474 Chr.20: 10038742-10038742 on Build GRCh38
rs779779505 Chr.20: 10039002-10039002 on Build GRCh38
rs887878415 Chr.20: 10038706-10038706 on Build GRCh38
rs773305271 Chr.20: 10038564-10038564 on Build GRCh38
rs998533652 Chr.20: 10038951-10038951 on Build GRCh38
rs763426983 Chr.20: 10038527-10038527 on Build GRCh38
rs778383812 Chr.20: 10038614-10038614 on Build GRCh38
rs777208036 Chr.20: 10038520-10038520 on Build GRCh38
rs771197219 Chr.20: 10038508-10038508 on Build GRCh38
rs188323188 Chr.20: 10038730-10038730 on Build GRCh38
rs7260784 Chr.20: 10038521-10038521 on Build GRCh38
rs775689262 Chr.20: 10038523-10038523 on Build GRCh38
rs756689534 Chr.20: 10038536-10038536 on Build GRCh38
rs780077900 Chr.20: 10038554-10038554 on Build GRCh38
rs935440929 Chr.20: 10038884-10038884 on Build GRCh38
rs761476237 Chr.20: 10038637-10038637 on Build GRCh38
rs35900659 Chr.20: 10038896-10038896 on Build GRCh38
rs958611814 Chr.20: 10038782-10038782 on Build GRCh38
rs895390432 Chr.20: 10038808-10038808 on Build GRCh38
rs1050628398 Chr.20: 10038975-10038975 on Build GRCh38
rs750310984 Chr.20: 10038546-10038546 on Build GRCh38
rs114094702 Chr.20: 10038596-10038596 on Build GRCh38
rs777850957 Chr.20: 10038556-10038556 on Build GRCh38
rs763224154 Chr.20: 10038624-10038624 on Build GRCh38
rs977673840 Chr.20: 10038869-10038869 on Build GRCh38
rs1006368509 Chr.20: 10038726-10038726 on Build GRCh38
rs756036453 Chr.20: 10038547-10038547 on Build GRCh38
rs1009038772 Chr.20: 10039005-10039005 on Build GRCh38
rs6087110 Chr.20: 10038984-10038984 on Build GRCh38
rs771072670 Chr.20: 10038569-10038569 on Build GRCh38
rs73897518 Chr.20: 10038692-10038692 on Build GRCh38

View Full Product Details