Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18813634-18814142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf78
Entrez Gene ID
100128496
Gene Name
chromosome 20 open reading frame 78
Gene Aliases
dJ1068E13.1
UniGene
Hs.664484
Gene Chromosome Location
Chr.20: 18809351-18830203 on Build GRCh38
Transcripts
NM_001242671.2 NM_001302818.1 NM_178483.2

Gene Symbol
SCP2D1
Entrez Gene ID
140856
Gene Name
SCP2 sterol binding domain containing 1
Gene Aliases
C20orf79, HSD22, dJ1068E13.2
UniGene
Hs.112794
Gene Chromosome Location
Chr.20: 18813726-18814392 on Build GRCh38
Transcripts
NM_001242671.2 NM_001302818.1 NM_178483.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5653578 c.255C>T p.T85T Chr.20: 18814070-18814070 on Build GRCh38
5644285 c.167delG p.E57fs*32 Chr.20: 18813982-18813982 on Build GRCh38
5578174 c.67G>A p.E23K Chr.20: 18813882-18813882 on Build GRCh38
5391645 c.95C>T p.A32V Chr.20: 18813910-18813910 on Build GRCh38
2704730 c.290C>T p.P97L Chr.20: 18814105-18814105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772324911 Chr.20: 18814100-18814100 on Build GRCh38
rs776432081 Chr.20: 18813870-18813870 on Build GRCh38
rs142475609 Chr.20: 18813930-18813930 on Build GRCh38
rs138668529 Chr.20: 18814020-18814020 on Build GRCh38
rs7352233 Chr.20: 18813883-18813883 on Build GRCh38
rs761978410 Chr.20: 18814014-18814014 on Build GRCh38
rs773937666 Chr.20: 18813915-18813915 on Build GRCh38
rs192783577 Chr.20: 18813984-18813984 on Build GRCh38
rs780421406 Chr.20: 18813803-18813803 on Build GRCh38
rs879329259 Chr.20: 18813733-18813733 on Build GRCh38
rs988791011 Chr.20: 18813717-18813717 on Build GRCh38
rs781006204 Chr.20: 18813813-18813813 on Build GRCh38
rs768912104 Chr.20: 18813999-18813999 on Build GRCh38
rs930923636 Chr.20: 18813660-18813660 on Build GRCh38
rs766902800 Chr.20: 18814044-18814044 on Build GRCh38
rs996710311 Chr.20: 18813699-18813699 on Build GRCh38
rs748896831 Chr.20: 18813813-18813813 on Build GRCh38
rs368681702 Chr.20: 18813833-18813833 on Build GRCh38
rs1016915375 Chr.20: 18813687-18813687 on Build GRCh38
rs908017068 Chr.20: 18814033-18814033 on Build GRCh38
rs760825661 Chr.20: 18813659-18813659 on Build GRCh38
rs374697243 Chr.20: 18814019-18814019 on Build GRCh38
rs758013817 Chr.20: 18813948-18813948 on Build GRCh38
rs772139970 Chr.20: 18814016-18814016 on Build GRCh38
rs143971555 Chr.20: 18813983-18813983 on Build GRCh38
rs773230277 Chr.20: 18814105-18814105 on Build GRCh38
rs765712789 Chr.20: 18813939-18813940 on Build GRCh38
rs777271829 Chr.20: 18813959-18813959 on Build GRCh38
rs765659732 Chr.20: 18814061-18814061 on Build GRCh38
rs778688716 Chr.20: 18813943-18813943 on Build GRCh38
rs760775866 Chr.20: 18814106-18814106 on Build GRCh38
rs138003057 Chr.20: 18813713-18813713 on Build GRCh38
rs750922065 Chr.20: 18813905-18813905 on Build GRCh38
rs200564762 Chr.20: 18814063-18814063 on Build GRCh38
rs903776476 Chr.20: 18813817-18813817 on Build GRCh38
rs41308066 Chr.20: 18813773-18813773 on Build GRCh38
rs968731014 Chr.20: 18814001-18814001 on Build GRCh38
rs781169917 Chr.20: 18814087-18814087 on Build GRCh38
rs756453842 Chr.20: 18813792-18813792 on Build GRCh38
rs374964954 Chr.20: 18813945-18813945 on Build GRCh38
rs79778412 Chr.20: 18813987-18813987 on Build GRCh38
rs570598719 Chr.20: 18813882-18813882 on Build GRCh38
rs201406631 Chr.20: 18813818-18813818 on Build GRCh38
rs531449512 Chr.20: 18813685-18813685 on Build GRCh38
rs547630005 Chr.20: 18813784-18813784 on Build GRCh38
rs764298632 Chr.20: 18813878-18813878 on Build GRCh38
rs760002689 Chr.20: 18814055-18814055 on Build GRCh38
rs750846012 Chr.20: 18813786-18813786 on Build GRCh38
rs746925152 Chr.20: 18813841-18813841 on Build GRCh38
rs1020268431 Chr.20: 18813720-18813720 on Build GRCh38
rs774575969 Chr.20: 18814004-18814004 on Build GRCh38
rs879075876 Chr.20: 18813703-18813703 on Build GRCh38
rs200917149 Chr.20: 18813970-18813970 on Build GRCh38
rs1053839 Chr.20: 18814110-18814110 on Build GRCh38
rs568111212 Chr.20: 18813750-18813750 on Build GRCh38
rs757831197 Chr.20: 18813770-18813770 on Build GRCh38
rs748382845 Chr.20: 18814099-18814099 on Build GRCh38
rs925190696 Chr.20: 18813769-18813769 on Build GRCh38
rs1053834 Chr.20: 18814070-18814070 on Build GRCh38
rs59741823 Chr.20: 18813656-18813656 on Build GRCh38
rs954764108 Chr.20: 18813712-18813712 on Build GRCh38
rs761177327 Chr.20: 18813909-18813909 on Build GRCh38
rs751079747 Chr.20: 18814081-18814081 on Build GRCh38
rs139167289 Chr.20: 18813901-18813901 on Build GRCh38
rs773306131 Chr.20: 18813825-18813825 on Build GRCh38
rs779754662 Chr.20: 18814098-18814098 on Build GRCh38
rs764475930 Chr.20: 18813691-18813691 on Build GRCh38
rs752403005 Chr.20: 18814062-18814062 on Build GRCh38
rs1050346638 Chr.20: 18814067-18814067 on Build GRCh38
rs917683341 Chr.20: 18813797-18813797 on Build GRCh38
rs940848927 Chr.20: 18814048-18814048 on Build GRCh38
rs575292008 Chr.20: 18813897-18813897 on Build GRCh38
rs772186682 Chr.20: 18813824-18813824 on Build GRCh38
rs556133893 Chr.20: 18813968-18813968 on Build GRCh38
rs968680039 Chr.20: 18813726-18813726 on Build GRCh38
rs749940712 Chr.20: 18813844-18813844 on Build GRCh38
rs763146845 Chr.20: 18813871-18813871 on Build GRCh38
rs757786265 Chr.20: 18813785-18813785 on Build GRCh38
rs868096249 Chr.20: 18813924-18813924 on Build GRCh38
rs372940690 Chr.20: 18813927-18813927 on Build GRCh38
rs41308631 Chr.20: 18813819-18813819 on Build GRCh38
rs745753112 Chr.20: 18814090-18814090 on Build GRCh38
rs766942012 Chr.20: 18814072-18814072 on Build GRCh38
rs564180497 Chr.20: 18813674-18813674 on Build GRCh38
rs144518160 Chr.20: 18813881-18813881 on Build GRCh38
rs747654523 Chr.20: 18813823-18813823 on Build GRCh38
rs745647775 Chr.20: 18813998-18813998 on Build GRCh38
rs931986466 Chr.20: 18813767-18813767 on Build GRCh38
rs757184912 Chr.20: 18814083-18814083 on Build GRCh38
rs979408731 Chr.20: 18813751-18813751 on Build GRCh38
rs372267357 Chr.20: 18814119-18814119 on Build GRCh38
rs1032266276 Chr.20: 18814036-18814036 on Build GRCh38
rs769605850 Chr.20: 18814092-18814092 on Build GRCh38
rs754743457 Chr.20: 18813933-18813933 on Build GRCh38
rs566343087 Chr.20: 18814118-18814127 on Build GRCh38
rs754193096 Chr.20: 18814054-18814054 on Build GRCh38

View Full Product Details