Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23364306-23364806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZF1
Entrez Gene ID
64412
Gene Name
GDNF inducible zinc finger protein 1
Gene Aliases
ZBTB23, ZNF336
UniGene
Hs.709462
Gene Chromosome Location
Chr.20: 23361585-23373062 on Build GRCh38
Transcripts
NM_001317012.1 NM_001317019.1 NM_022482.4 XM_005260800.2 XM_005260802.3 XM_011529321.2 XM_011529322.1 XM_017028017.1 XM_017028018.1 XM_017028019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1239042 c.260T>C p.F87S Chr.20: 23364643-23364643 on Build GRCh38
6677282 c.107G>A p.S36N Chr.20: 23364490-23364490 on Build GRCh38
1410784 c.138C>A p.F46L Chr.20: 23364521-23364521 on Build GRCh38
7260294 c.359A>T p.D120V Chr.20: 23364742-23364742 on Build GRCh38
1410785 c.313C>T p.R105W Chr.20: 23364696-23364696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745818348 Chr.20: 23364457-23364457 on Build GRCh38
rs774092249 Chr.20: 23364588-23364588 on Build GRCh38
rs760155848 Chr.20: 23364522-23364522 on Build GRCh38
rs767776459 Chr.20: 23364665-23364665 on Build GRCh38
rs200516081 Chr.20: 23364740-23364740 on Build GRCh38
rs183798033 Chr.20: 23364331-23364331 on Build GRCh38
rs576902241 Chr.20: 23364392-23364392 on Build GRCh38
rs774832417 Chr.20: 23364465-23364465 on Build GRCh38
rs150491728 Chr.20: 23364473-23364473 on Build GRCh38
rs866030971 Chr.20: 23364372-23364372 on Build GRCh38
rs762535046 Chr.20: 23364355-23364355 on Build GRCh38
rs374008128 Chr.20: 23364382-23364382 on Build GRCh38
rs777850882 Chr.20: 23364539-23364539 on Build GRCh38
rs560543912 Chr.20: 23364585-23364585 on Build GRCh38
rs756606093 Chr.20: 23364387-23364387 on Build GRCh38
rs776605405 Chr.20: 23364346-23364346 on Build GRCh38
rs146392173 Chr.20: 23364548-23364548 on Build GRCh38
rs772908144 Chr.20: 23364339-23364339 on Build GRCh38
rs779791091 Chr.20: 23364693-23364693 on Build GRCh38
rs377379164 Chr.20: 23364644-23364644 on Build GRCh38
rs780098479 Chr.20: 23364581-23364581 on Build GRCh38
rs143111256 Chr.20: 23364663-23364663 on Build GRCh38
rs374755807 Chr.20: 23364696-23364696 on Build GRCh38
rs946913529 Chr.20: 23364393-23364393 on Build GRCh38
rs189316929 Chr.20: 23364760-23364760 on Build GRCh38
rs756987103 Chr.20: 23364556-23364556 on Build GRCh38
rs148101388 Chr.20: 23364415-23364415 on Build GRCh38
rs374275229 Chr.20: 23364632-23364632 on Build GRCh38
rs775150641 Chr.20: 23364354-23364354 on Build GRCh38
rs138988702 Chr.20: 23364606-23364606 on Build GRCh38
rs988499404 Chr.20: 23364765-23364765 on Build GRCh38
rs754403991 Chr.20: 23364396-23364396 on Build GRCh38
rs370629303 Chr.20: 23364629-23364629 on Build GRCh38
rs372760791 Chr.20: 23364510-23364510 on Build GRCh38
rs367955768 Chr.20: 23364701-23364701 on Build GRCh38
rs770197032 Chr.20: 23364776-23364776 on Build GRCh38
rs200302078 Chr.20: 23364351-23364351 on Build GRCh38
rs976318007 Chr.20: 23364633-23364633 on Build GRCh38
rs773053982 Chr.20: 23364757-23364757 on Build GRCh38
rs138315434 Chr.20: 23364479-23364479 on Build GRCh38
rs747704995 Chr.20: 23364705-23364705 on Build GRCh38
rs370627382 Chr.20: 23364695-23364695 on Build GRCh38
rs759041068 Chr.20: 23364348-23364348 on Build GRCh38
rs769298576 Chr.20: 23364458-23364458 on Build GRCh38
rs545992812 Chr.20: 23364610-23364610 on Build GRCh38
rs780987756 Chr.20: 23364559-23364559 on Build GRCh38
rs765834776 Chr.20: 23364464-23364464 on Build GRCh38
rs758513587 Chr.20: 23364422-23364422 on Build GRCh38
rs769598052 Chr.20: 23364576-23364576 on Build GRCh38
rs749264121 Chr.20: 23364697-23364697 on Build GRCh38
rs779082542 Chr.20: 23364436-23364444 on Build GRCh38
rs6076060 Chr.20: 23364432-23364432 on Build GRCh38
rs563699571 Chr.20: 23364600-23364600 on Build GRCh38
rs746842036 Chr.20: 23364341-23364341 on Build GRCh38
rs376361468 Chr.20: 23364698-23364698 on Build GRCh38
rs771854846 Chr.20: 23364337-23364337 on Build GRCh38
rs746210854 Chr.20: 23364768-23364768 on Build GRCh38
rs140549788 Chr.20: 23364456-23364456 on Build GRCh38
rs750206786 Chr.20: 23364668-23364668 on Build GRCh38
rs770831542 Chr.20: 23364343-23364343 on Build GRCh38
rs763847181 Chr.20: 23364356-23364356 on Build GRCh38
rs754352875 Chr.20: 23364511-23364511 on Build GRCh38
rs149633064 Chr.20: 23364491-23364491 on Build GRCh38
rs752921068 Chr.20: 23364416-23364416 on Build GRCh38
rs758561145 Chr.20: 23364537-23364537 on Build GRCh38
rs745765108 Chr.20: 23364570-23364570 on Build GRCh38
rs753311654 Chr.20: 23364641-23364641 on Build GRCh38
rs199754225 Chr.20: 23364398-23364398 on Build GRCh38
rs540788122 Chr.20: 23364533-23364533 on Build GRCh38
rs879374128 Chr.20: 23364655-23364655 on Build GRCh38
rs770778427 Chr.20: 23364436-23364436 on Build GRCh38
rs764171758 Chr.20: 23364650-23364650 on Build GRCh38
rs748548388 Chr.20: 23364767-23364770 on Build GRCh38
rs750803117 Chr.20: 23364334-23364334 on Build GRCh38
rs756005236 Chr.20: 23364677-23364677 on Build GRCh38
rs768219558 Chr.20: 23364586-23364586 on Build GRCh38
rs375983173 Chr.20: 23364532-23364532 on Build GRCh38
rs778103641 Chr.20: 23364431-23364431 on Build GRCh38

View Full Product Details