Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31308428-31308927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB116
Entrez Gene ID
245930
Gene Name
defensin beta 116
Gene Aliases
DEFB-16
UniGene
Hs.580703
Gene Chromosome Location
Chr.20: 31303212-31308585 on Build GRCh38
Transcripts
NM_001037731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3545001 c.5C>T p.S2L Chr.20: 31308581-31308581 on Build GRCh38
6228764 c.32T>C p.I11T Chr.20: 31308554-31308554 on Build GRCh38
5689216 c.18C>T p.P6P Chr.20: 31308568-31308568 on Build GRCh38
4817300 c.55C>T p.Q19* Chr.20: 31308531-31308531 on Build GRCh38
6558638 c.9C>A p.V3V Chr.20: 31308577-31308577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778999910 Chr.20: 31308775-31308775 on Build GRCh38
rs898330462 Chr.20: 31308776-31308776 on Build GRCh38
rs1006654162 Chr.20: 31308739-31308739 on Build GRCh38
rs749785831 Chr.20: 31308468-31308468 on Build GRCh38
rs780176637 Chr.20: 31308579-31308579 on Build GRCh38
rs375031983 Chr.20: 31308616-31308616 on Build GRCh38
rs568378283 Chr.20: 31308887-31308887 on Build GRCh38
rs764869378 Chr.20: 31308499-31308499 on Build GRCh38
rs769709337 Chr.20: 31308623-31308623 on Build GRCh38
rs755301625 Chr.20: 31308590-31308590 on Build GRCh38
rs748264491 Chr.20: 31308551-31308551 on Build GRCh38
rs760580804 Chr.20: 31308489-31308489 on Build GRCh38
rs763499188 Chr.20: 31308575-31308575 on Build GRCh38
rs928100972 Chr.20: 31308595-31308595 on Build GRCh38
rs775626653 Chr.20: 31308624-31308624 on Build GRCh38
rs183829648 Chr.20: 31308618-31308618 on Build GRCh38
rs376747448 Chr.20: 31308550-31308550 on Build GRCh38
rs6119768 Chr.20: 31308530-31308530 on Build GRCh38
rs764445975 Chr.20: 31308621-31308624 on Build GRCh38
rs756351237 Chr.20: 31308577-31308577 on Build GRCh38
rs980231335 Chr.20: 31308871-31308871 on Build GRCh38
rs754896592 Chr.20: 31308498-31308498 on Build GRCh38
rs997855972 Chr.20: 31308542-31308542 on Build GRCh38
rs971560979 Chr.20: 31308806-31308806 on Build GRCh38
rs777646640 Chr.20: 31308613-31308613 on Build GRCh38
rs773536225 Chr.20: 31308555-31308555 on Build GRCh38
rs554645221 Chr.20: 31308862-31308862 on Build GRCh38
rs200042260 Chr.20: 31308504-31308504 on Build GRCh38
rs372498579 Chr.20: 31308525-31308525 on Build GRCh38
rs902636751 Chr.20: 31308668-31308668 on Build GRCh38
rs879616381 Chr.20: 31308537-31308537 on Build GRCh38
rs959554797 Chr.20: 31308830-31308830 on Build GRCh38
rs376994268 Chr.20: 31308503-31308503 on Build GRCh38
rs772038210 Chr.20: 31308554-31308554 on Build GRCh38
rs148563954 Chr.20: 31308629-31308629 on Build GRCh38
rs780501111 Chr.20: 31308580-31308580 on Build GRCh38
rs749344111 Chr.20: 31308654-31308654 on Build GRCh38
rs552662348 Chr.20: 31308704-31308704 on Build GRCh38
rs994051803 Chr.20: 31308781-31308781 on Build GRCh38
rs752652533 Chr.20: 31308570-31308570 on Build GRCh38
rs1029398947 Chr.20: 31308543-31308543 on Build GRCh38
rs1006831706 Chr.20: 31308700-31308700 on Build GRCh38
rs540450843 Chr.20: 31308471-31308471 on Build GRCh38
rs981184961 Chr.20: 31308808-31308808 on Build GRCh38
rs760746434 Chr.20: 31308558-31308558 on Build GRCh38
rs990934575 Chr.20: 31308882-31308882 on Build GRCh38
rs754998901 Chr.20: 31308684-31308684 on Build GRCh38
rs922156500 Chr.20: 31308597-31308632 on Build GRCh38
rs891453082 Chr.20: 31308706-31308706 on Build GRCh38
rs567040481 Chr.20: 31308676-31308676 on Build GRCh38
rs962610243 Chr.20: 31308768-31308768 on Build GRCh38
rs867343316 Chr.20: 31308602-31308602 on Build GRCh38
rs756559816 Chr.20: 31308531-31308531 on Build GRCh38
rs114153561 Chr.20: 31308677-31308677 on Build GRCh38
rs200029152 Chr.20: 31308877-31308877 on Build GRCh38
rs992853738 Chr.20: 31308812-31308812 on Build GRCh38
rs780228144 Chr.20: 31308534-31308534 on Build GRCh38
rs1050781320 Chr.20: 31308717-31308717 on Build GRCh38
rs768944135 Chr.20: 31308453-31308453 on Build GRCh38
rs6121357 Chr.20: 31308497-31308497 on Build GRCh38
rs574641497 Chr.20: 31308458-31308458 on Build GRCh38
rs978500590 Chr.20: 31308903-31308903 on Build GRCh38
rs769397185 Chr.20: 31308469-31308470 on Build GRCh38
rs767282930 Chr.20: 31308483-31308483 on Build GRCh38
rs779461835 Chr.20: 31308593-31308593 on Build GRCh38
rs765316849 Chr.20: 31308568-31308568 on Build GRCh38
rs950693630 Chr.20: 31308786-31308786 on Build GRCh38
rs762056119 Chr.20: 31308482-31308482 on Build GRCh38
rs144824349 Chr.20: 31308512-31308512 on Build GRCh38
rs746729803 Chr.20: 31308897-31308897 on Build GRCh38
rs370984956 Chr.20: 31308825-31308825 on Build GRCh38
rs962349676 Chr.20: 31308524-31308524 on Build GRCh38
rs754155550 Chr.20: 31308582-31308582 on Build GRCh38
rs775907336 Chr.20: 31308518-31308518 on Build GRCh38
rs774351342 Chr.20: 31308474-31308474 on Build GRCh38
rs1025267380 Chr.20: 31308782-31308782 on Build GRCh38
rs201371407 Chr.20: 31308614-31308614 on Build GRCh38
rs773273644 Chr.20: 31308488-31308488 on Build GRCh38
rs773087940 Chr.20: 31308634-31308634 on Build GRCh38
rs946986505 Chr.20: 31308894-31308894 on Build GRCh38
rs759566931 Chr.20: 31308565-31308565 on Build GRCh38
rs145509874 Chr.20: 31308708-31308708 on Build GRCh38
rs770765146 Chr.20: 31308561-31308561 on Build GRCh38
rs766071680 Chr.20: 31308490-31308490 on Build GRCh38
rs917361899 Chr.20: 31308651-31308651 on Build GRCh38
rs1057242395 Chr.20: 31308455-31308455 on Build GRCh38
rs972663244 Chr.20: 31308609-31308609 on Build GRCh38
rs370016029 Chr.20: 31308576-31308576 on Build GRCh38
rs763165282 Chr.20: 31308616-31308617 on Build GRCh38
rs915474181 Chr.20: 31308615-31308615 on Build GRCh38
rs868771272 Chr.20: 31308574-31308574 on Build GRCh38
rs368326400 Chr.20: 31308508-31308508 on Build GRCh38
rs531243939 Chr.20: 31308563-31308563 on Build GRCh38
rs970519170 Chr.20: 31308868-31308868 on Build GRCh38
rs201949154 Chr.20: 31308535-31308535 on Build GRCh38
rs372157777 Chr.20: 31308485-31308485 on Build GRCh38
rs1029496231 Chr.20: 31308470-31308470 on Build GRCh38
rs777524184 Chr.20: 31308519-31308519 on Build GRCh38
rs373060353 Chr.20: 31308612-31308612 on Build GRCh38
rs989017947 Chr.20: 31308899-31308899 on Build GRCh38
rs998461709 Chr.20: 31308463-31308463 on Build GRCh38
rs6121358 Chr.20: 31308510-31308510 on Build GRCh38
rs183515741 Chr.20: 31308652-31308652 on Build GRCh38

View Full Product Details