Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31665793-31666293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L1
Entrez Gene ID
598
Gene Name
BCL2 like 1
Gene Aliases
BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52
UniGene
Hs.516966
Gene Chromosome Location
Chr.20: 31664452-31723999 on Build GRCh38
Transcripts
NM_001191.3 NM_001317919.1 NM_001317920.1 NM_001317921.1 NM_001322239.1 NM_001322240.1 NM_001322242.1 NM_138578.2 NR_134257.1 XM_011528964.2 XM_017027993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
723188 c.678G>C p.L226L Chr.20: 31665973-31665973 on Build GRCh38
723190 c.679G>T p.G227C Chr.20: 31665972-31665972 on Build GRCh38
6919377 c.634C>T p.R212C Chr.20: 31666017-31666017 on Build GRCh38
6987550 c.654G>A p.M218I Chr.20: 31665997-31665997 on Build GRCh38
3991621 c.573T>C p.F191F Chr.20: 31666078-31666078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768665664 Chr.20: 31666025-31666025 on Build GRCh38
rs939286454 Chr.20: 31666093-31666093 on Build GRCh38
rs112926293 Chr.20: 31666134-31666134 on Build GRCh38
rs143094422 Chr.20: 31666092-31666092 on Build GRCh38
rs772077664 Chr.20: 31665996-31665996 on Build GRCh38
rs780264826 Chr.20: 31665984-31665984 on Build GRCh38
rs543917106 Chr.20: 31665988-31665988 on Build GRCh38
rs950656912 Chr.20: 31665826-31665826 on Build GRCh38
rs1038777780 Chr.20: 31666136-31666136 on Build GRCh38
rs189288861 Chr.20: 31666261-31666261 on Build GRCh38
rs749320983 Chr.20: 31666071-31666071 on Build GRCh38
rs762443576 Chr.20: 31666197-31666197 on Build GRCh38
rs1022080146 Chr.20: 31665835-31665835 on Build GRCh38
rs757599194 Chr.20: 31666048-31666048 on Build GRCh38
rs11550472 Chr.20: 31665974-31665974 on Build GRCh38
rs756307657 Chr.20: 31665968-31665968 on Build GRCh38
rs577378728 Chr.20: 31666069-31666069 on Build GRCh38
rs765845311 Chr.20: 31665914-31665914 on Build GRCh38
rs774540387 Chr.20: 31666193-31666193 on Build GRCh38
rs992719994 Chr.20: 31665828-31665828 on Build GRCh38
rs886118122 Chr.20: 31666207-31666207 on Build GRCh38
rs769154919 Chr.20: 31665907-31665907 on Build GRCh38
rs969871111 Chr.20: 31665870-31665870 on Build GRCh38
rs11550475 Chr.20: 31665824-31665824 on Build GRCh38
rs781506995 Chr.20: 31665943-31665943 on Build GRCh38
rs1024061730 Chr.20: 31665832-31665832 on Build GRCh38
rs759194807 Chr.20: 31666026-31666026 on Build GRCh38
rs895041150 Chr.20: 31666169-31666169 on Build GRCh38
rs7362890 Chr.20: 31665983-31665983 on Build GRCh38
rs989916889 Chr.20: 31665903-31665903 on Build GRCh38
rs751808059 Chr.20: 31665938-31665938 on Build GRCh38
rs1014089566 Chr.20: 31666210-31666210 on Build GRCh38
rs764588065 Chr.20: 31665930-31665930 on Build GRCh38
rs375110827 Chr.20: 31666051-31666051 on Build GRCh38
rs1012109210 Chr.20: 31666205-31666205 on Build GRCh38
rs751021951 Chr.20: 31666057-31666057 on Build GRCh38
rs201204270 Chr.20: 31665985-31665985 on Build GRCh38
rs200117616 Chr.20: 31666054-31666054 on Build GRCh38
rs903637824 Chr.20: 31666223-31666223 on Build GRCh38
rs898941505 Chr.20: 31666140-31666140 on Build GRCh38
rs558849192 Chr.20: 31666011-31666011 on Build GRCh38
rs41293088 Chr.20: 31666215-31666215 on Build GRCh38
rs763223337 Chr.20: 31666016-31666016 on Build GRCh38
rs577055474 Chr.20: 31666017-31666017 on Build GRCh38
rs945582919 Chr.20: 31666024-31666024 on Build GRCh38
rs760138522 Chr.20: 31665905-31665905 on Build GRCh38
rs1030657544 Chr.20: 31665913-31665913 on Build GRCh38
rs755249882 Chr.20: 31666078-31666078 on Build GRCh38
rs560897598 Chr.20: 31666266-31666266 on Build GRCh38
rs768692210 Chr.20: 31665987-31665987 on Build GRCh38
rs753539518 Chr.20: 31665922-31665922 on Build GRCh38
rs930721821 Chr.20: 31666043-31666043 on Build GRCh38
rs764743032 Chr.20: 31666037-31666037 on Build GRCh38
rs754613701 Chr.20: 31665923-31665923 on Build GRCh38
rs536243459 Chr.20: 31665840-31665840 on Build GRCh38
rs560082530 Chr.20: 31666206-31666206 on Build GRCh38
rs773174390 Chr.20: 31666003-31666003 on Build GRCh38
rs967769834 Chr.20: 31665889-31665889 on Build GRCh38
rs936677262 Chr.20: 31665887-31665887 on Build GRCh38
rs150204038 Chr.20: 31665964-31665964 on Build GRCh38
rs999241330 Chr.20: 31666225-31666225 on Build GRCh38
rs751979681 Chr.20: 31666047-31666047 on Build GRCh38
rs980376640 Chr.20: 31666258-31666258 on Build GRCh38
rs756681825 Chr.20: 31666060-31666060 on Build GRCh38
rs554584920 Chr.20: 31665890-31665890 on Build GRCh38
rs1031086621 Chr.20: 31666265-31666265 on Build GRCh38
rs746342082 Chr.20: 31665957-31665957 on Build GRCh38
rs757800093 Chr.20: 31665941-31665941 on Build GRCh38
rs926738457 Chr.20: 31665885-31665885 on Build GRCh38
rs980839387 Chr.20: 31665881-31665881 on Build GRCh38
rs994493000 Chr.20: 31666177-31666177 on Build GRCh38

View Full Product Details