Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32010363-32010858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2L
Entrez Gene ID
140706
Gene Name
CCM2 like scaffolding protein
Gene Aliases
C20orf160, dJ310O13.5
UniGene
Hs.382151
Gene Chromosome Location
Chr.20: 32010379-32032182 on Build GRCh38
Transcripts
NM_080625.3 XM_006723706.3 XM_011528566.1 XM_011528568.2 XM_011528569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389671 c.13G>T p.V5F Chr.20: 32010467-32010467 on Build GRCh38
5805441 c.8A>G p.Y3C Chr.20: 32010462-32010462 on Build GRCh38
3963455 c.3G>T p.M1I Chr.20: 32010457-32010457 on Build GRCh38
3963456 c.3G>T p.M1I Chr.20: 32010457-32010457 on Build GRCh38
4536448 c.22G>A p.G8R Chr.20: 32010476-32010476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543571284 Chr.20: 32010587-32010587 on Build GRCh38
rs923279144 Chr.20: 32010434-32010438 on Build GRCh38
rs776266841 Chr.20: 32010509-32010509 on Build GRCh38
rs962759721 Chr.20: 32010586-32010586 on Build GRCh38
rs1051946966 Chr.20: 32010816-32010816 on Build GRCh38
rs142123448 Chr.20: 32010799-32010799 on Build GRCh38
rs1006021475 Chr.20: 32010551-32010555 on Build GRCh38
rs201157058 Chr.20: 32010486-32010486 on Build GRCh38
rs13039469 Chr.20: 32010544-32010544 on Build GRCh38
rs960226444 Chr.20: 32010449-32010462 on Build GRCh38
rs1037549904 Chr.20: 32010553-32010553 on Build GRCh38
rs1011678642 Chr.20: 32010413-32010413 on Build GRCh38
rs897689635 Chr.20: 32010561-32010561 on Build GRCh38
rs985502206 Chr.20: 32010579-32010579 on Build GRCh38
rs968440649 Chr.20: 32010674-32010674 on Build GRCh38
rs957116750 Chr.20: 32010690-32010690 on Build GRCh38
rs897636873 Chr.20: 32010815-32010815 on Build GRCh38
rs979535124 Chr.20: 32010677-32010677 on Build GRCh38
rs547702169 Chr.20: 32010738-32010738 on Build GRCh38
rs868144141 Chr.20: 32010560-32010560 on Build GRCh38
rs986134924 Chr.20: 32010775-32010775 on Build GRCh38
rs549059798 Chr.20: 32010766-32010766 on Build GRCh38
rs893179631 Chr.20: 32010395-32010395 on Build GRCh38
rs796191955 Chr.20: 32010642-32010642 on Build GRCh38
rs372405341 Chr.20: 32010427-32010427 on Build GRCh38
rs973880166 Chr.20: 32010631-32010631 on Build GRCh38
rs375564243 Chr.20: 32010456-32010456 on Build GRCh38
rs73906718 Chr.20: 32010686-32010686 on Build GRCh38
rs577964289 Chr.20: 32010410-32010410 on Build GRCh38
rs918412593 Chr.20: 32010657-32010657 on Build GRCh38
rs981245831 Chr.20: 32010688-32010688 on Build GRCh38
rs937285255 Chr.20: 32010701-32010701 on Build GRCh38
rs903438134 Chr.20: 32010550-32010550 on Build GRCh38
rs369950230 Chr.20: 32010502-32010502 on Build GRCh38
rs1041858251 Chr.20: 32010785-32010785 on Build GRCh38
rs572380471 Chr.20: 32010539-32010539 on Build GRCh38
rs773729023 Chr.20: 32010491-32010491 on Build GRCh38
rs13045124 Chr.20: 32010545-32010545 on Build GRCh38
rs566448966 Chr.20: 32010740-32010740 on Build GRCh38
rs73906716 Chr.20: 32010582-32010582 on Build GRCh38
rs372361959 Chr.20: 32010507-32010507 on Build GRCh38
rs368759046 Chr.20: 32010556-32010556 on Build GRCh38
rs376443491 Chr.20: 32010699-32010699 on Build GRCh38
rs766619889 Chr.20: 32010687-32010687 on Build GRCh38
rs925499415 Chr.20: 32010412-32010412 on Build GRCh38
rs1020445546 Chr.20: 32010478-32010478 on Build GRCh38
rs139652666 Chr.20: 32010744-32010744 on Build GRCh38
rs117798563 Chr.20: 32010569-32010569 on Build GRCh38
rs558038839 Chr.20: 32010802-32010802 on Build GRCh38
rs569104569 Chr.20: 32010784-32010784 on Build GRCh38
rs993423451 Chr.20: 32010568-32010568 on Build GRCh38
rs376430290 Chr.20: 32010555-32010555 on Build GRCh38

View Full Product Details