Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32018773-32019276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2L
Entrez Gene ID
140706
Gene Name
CCM2 like scaffolding protein
Gene Aliases
C20orf160, dJ310O13.5
UniGene
Hs.382151
Gene Chromosome Location
Chr.20: 32010379-32032182 on Build GRCh38
Transcripts
NM_080625.3 XM_006723706.3 XM_011528566.1 XM_011528567.2 XM_011528568.2 XM_011528569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5674387 c.591C>A p.T197T Chr.20: 32019067-32019067 on Build GRCh38
5682840 c.566T>G p.V189G Chr.20: 32019042-32019042 on Build GRCh38
4646017 c.479A>G p.D160G Chr.20: 32018955-32018955 on Build GRCh38
5679657 c.624T>G p.G208G Chr.20: 32019100-32019100 on Build GRCh38
4134420 c.780G>A p.A260A Chr.20: 32019256-32019256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056668757 Chr.20: 32018823-32018823 on Build GRCh38
rs746058580 Chr.20: 32018978-32018978 on Build GRCh38
rs1019943992 Chr.20: 32018898-32018899 on Build GRCh38
rs930613519 Chr.20: 32018854-32018854 on Build GRCh38
rs892999090 Chr.20: 32019005-32019005 on Build GRCh38
rs930629206 Chr.20: 32018879-32018879 on Build GRCh38
rs907932970 Chr.20: 32018897-32018897 on Build GRCh38
rs974673858 Chr.20: 32018872-32018872 on Build GRCh38
rs981840317 Chr.20: 32019209-32019209 on Build GRCh38
rs896724111 Chr.20: 32018824-32018824 on Build GRCh38
rs755937904 Chr.20: 32018926-32018926 on Build GRCh38
rs1056635764 Chr.20: 32018922-32018922 on Build GRCh38
rs995289129 Chr.20: 32019087-32019091 on Build GRCh38
rs983696056 Chr.20: 32018958-32018958 on Build GRCh38
rs909867 Chr.20: 32018801-32018801 on Build GRCh38
rs971388811 Chr.20: 32019200-32019200 on Build GRCh38
rs372054476 Chr.20: 32018861-32018861 on Build GRCh38
rs908137061 Chr.20: 32019172-32019172 on Build GRCh38
rs943670712 Chr.20: 32019042-32019042 on Build GRCh38
rs549097248 Chr.20: 32018838-32018838 on Build GRCh38
rs985212843 Chr.20: 32018889-32018889 on Build GRCh38
rs1018461636 Chr.20: 32018911-32018911 on Build GRCh38
rs1001777114 Chr.20: 32018892-32018892 on Build GRCh38
rs930876817 Chr.20: 32018947-32018947 on Build GRCh38
rs964127762 Chr.20: 32019037-32019037 on Build GRCh38
rs1010144980 Chr.20: 32019017-32019017 on Build GRCh38
rs560923978 Chr.20: 32018844-32018844 on Build GRCh38
rs1054691682 Chr.20: 32019002-32019002 on Build GRCh38
rs950109974 Chr.20: 32018975-32018975 on Build GRCh38
rs1008312405 Chr.20: 32019029-32019029 on Build GRCh38
rs988644978 Chr.20: 32018851-32018851 on Build GRCh38
rs905950571 Chr.20: 32018986-32018986 on Build GRCh38
rs190244994 Chr.20: 32019094-32019094 on Build GRCh38
rs965929100 Chr.20: 32018867-32018867 on Build GRCh38
rs910786041 Chr.20: 32018895-32018895 on Build GRCh38
rs141969168 Chr.20: 32018810-32018810 on Build GRCh38
rs1032893948 Chr.20: 32018893-32018893 on Build GRCh38
rs912051855 Chr.20: 32018828-32018828 on Build GRCh38
rs907849774 Chr.20: 32018971-32018971 on Build GRCh38
rs886533162 Chr.20: 32018809-32018809 on Build GRCh38
rs779813482 Chr.20: 32019229-32019240 on Build GRCh38
rs796342232 Chr.20: 32019117-32019117 on Build GRCh38
rs571435366 Chr.20: 32019103-32019103 on Build GRCh38
rs538538318 Chr.20: 32019256-32019256 on Build GRCh38
rs549343031 Chr.20: 32018908-32018908 on Build GRCh38
rs1012535224 Chr.20: 32018837-32018837 on Build GRCh38
rs1045776327 Chr.20: 32018980-32018980 on Build GRCh38
rs940797178 Chr.20: 32018914-32018914 on Build GRCh38
rs929476187 Chr.20: 32019100-32019100 on Build GRCh38
rs886463496 Chr.20: 32018863-32018863 on Build GRCh38
rs937475086 Chr.20: 32019001-32019001 on Build GRCh38
rs921856492 Chr.20: 32018876-32018876 on Build GRCh38
rs920803544 Chr.20: 32019095-32019095 on Build GRCh38
rs927354499 Chr.20: 32019222-32019222 on Build GRCh38
rs1017991553 Chr.20: 32019036-32019036 on Build GRCh38
rs956002055 Chr.20: 32018848-32018848 on Build GRCh38
rs1000276220 Chr.20: 32018841-32018841 on Build GRCh38
rs562260253 Chr.20: 32018887-32018888 on Build GRCh38
rs955053673 Chr.20: 32018907-32018907 on Build GRCh38
rs890773980 Chr.20: 32019028-32019028 on Build GRCh38
rs1054575301 Chr.20: 32019233-32019238 on Build GRCh38
rs920798720 Chr.20: 32018933-32018933 on Build GRCh38
rs749268099 Chr.20: 32019233-32019235 on Build GRCh38
rs952224232 Chr.20: 32019106-32019106 on Build GRCh38
rs937405199 Chr.20: 32019232-32019233 on Build GRCh38
rs1020226354 Chr.20: 32019025-32019025 on Build GRCh38
rs775248451 Chr.20: 32019133-32019133 on Build GRCh38
rs905488243 Chr.20: 32018840-32018840 on Build GRCh38

View Full Product Details