Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32030938-32031442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2L
Entrez Gene ID
140706
Gene Name
CCM2 like scaffolding protein
Gene Aliases
C20orf160, dJ310O13.5
UniGene
Hs.382151
Gene Chromosome Location
Chr.20: 32010379-32032182 on Build GRCh38
Transcripts
NM_080625.3 XM_006723706.3 XM_011528566.1 XM_011528567.2 XM_011528568.2 XM_011528569.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5951744 c.1614G>A p.T538T Chr.20: 32031212-32031212 on Build GRCh38
7238702 c.1439A>G p.Y480C Chr.20: 32031037-32031037 on Build GRCh38
4445590 c.1486C>T p.L496F Chr.20: 32031084-32031084 on Build GRCh38
3707687 c.1452C>T p.F484F Chr.20: 32031050-32031050 on Build GRCh38
5150949 c.1466G>C p.G489A Chr.20: 32031064-32031064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552472243 Chr.20: 32031249-32031249 on Build GRCh38
rs1019391106 Chr.20: 32031305-32031305 on Build GRCh38
rs751832452 Chr.20: 32031236-32031237 on Build GRCh38
rs996687499 Chr.20: 32031327-32031327 on Build GRCh38
rs982716017 Chr.20: 32031182-32031182 on Build GRCh38
rs1017649503 Chr.20: 32031099-32031099 on Build GRCh38
rs919863944 Chr.20: 32031153-32031153 on Build GRCh38
rs771144429 Chr.20: 32031282-32031282 on Build GRCh38
rs1047472940 Chr.20: 32031003-32031003 on Build GRCh38
rs540720404 Chr.20: 32031092-32031092 on Build GRCh38
rs1053361875 Chr.20: 32031253-32031253 on Build GRCh38
rs747035577 Chr.20: 32031262-32031262 on Build GRCh38
rs963758564 Chr.20: 32031318-32031318 on Build GRCh38
rs762569054 Chr.20: 32031011-32031011 on Build GRCh38
rs1026022355 Chr.20: 32031419-32031419 on Build GRCh38
rs751323701 Chr.20: 32031056-32031056 on Build GRCh38
rs574083033 Chr.20: 32031123-32031123 on Build GRCh38
rs908569083 Chr.20: 32031191-32031191 on Build GRCh38
rs938803052 Chr.20: 32031020-32031020 on Build GRCh38
rs563364213 Chr.20: 32031069-32031069 on Build GRCh38
rs948901504 Chr.20: 32031228-32031228 on Build GRCh38
rs956392155 Chr.20: 32031121-32031121 on Build GRCh38
rs975216903 Chr.20: 32031143-32031143 on Build GRCh38
rs983132150 Chr.20: 32031371-32031371 on Build GRCh38
rs992759287 Chr.20: 32031417-32031417 on Build GRCh38
rs899105138 Chr.20: 32031392-32031393 on Build GRCh38
rs758573932 Chr.20: 32031246-32031246 on Build GRCh38
rs943258589 Chr.20: 32031374-32031374 on Build GRCh38
rs916004064 Chr.20: 32031418-32031418 on Build GRCh38
rs1038965192 Chr.20: 32031375-32031375 on Build GRCh38
rs755396923 Chr.20: 32031231-32031231 on Build GRCh38
rs141874653 Chr.20: 32030993-32030993 on Build GRCh38
rs552082280 Chr.20: 32031248-32031248 on Build GRCh38
rs78827511 Chr.20: 32031256-32031257 on Build GRCh38
rs994559176 Chr.20: 32031113-32031113 on Build GRCh38
rs1000543428 Chr.20: 32031097-32031097 on Build GRCh38
rs1007874905 Chr.20: 32031293-32031293 on Build GRCh38
rs890360872 Chr.20: 32031054-32031054 on Build GRCh38
rs539753399 Chr.20: 32031060-32031060 on Build GRCh38
rs1031125678 Chr.20: 32031115-32031115 on Build GRCh38
rs890299211 Chr.20: 32031358-32031358 on Build GRCh38
rs952567166 Chr.20: 32031356-32031356 on Build GRCh38
rs45565933 Chr.20: 32031034-32031034 on Build GRCh38
rs926651151 Chr.20: 32030963-32030963 on Build GRCh38
rs867090556 Chr.20: 32031169-32031169 on Build GRCh38
rs1043171969 Chr.20: 32031245-32031245 on Build GRCh38
rs867616586 Chr.20: 32031103-32031103 on Build GRCh38
rs533444431 Chr.20: 32031158-32031158 on Build GRCh38
rs915971637 Chr.20: 32031227-32031227 on Build GRCh38
rs1027273977 Chr.20: 32031339-32031339 on Build GRCh38
rs756967887 Chr.20: 32031333-32031333 on Build GRCh38
rs913987764 Chr.20: 32031288-32031288 on Build GRCh38
rs1007484682 Chr.20: 32031086-32031086 on Build GRCh38
rs139704146 Chr.20: 32031239-32031240 on Build GRCh38
rs963096274 Chr.20: 32031100-32031100 on Build GRCh38
rs751232656 Chr.20: 32031255-32031255 on Build GRCh38
rs764167579 Chr.20: 32031231-32031232 on Build GRCh38
rs766846316 Chr.20: 32031059-32031059 on Build GRCh38
rs749216690 Chr.20: 32031240-32031248 on Build GRCh38
rs368034293 Chr.20: 32031127-32031127 on Build GRCh38
rs758481947 Chr.20: 32030959-32030959 on Build GRCh38
rs908434989 Chr.20: 32031373-32031373 on Build GRCh38
rs192307170 Chr.20: 32031131-32031131 on Build GRCh38
rs919820221 Chr.20: 32030965-32030965 on Build GRCh38
rs1043140075 Chr.20: 32031422-32031422 on Build GRCh38
rs938680540 Chr.20: 32031193-32031193 on Build GRCh38
rs184780954 Chr.20: 32031393-32031393 on Build GRCh38
rs562123754 Chr.20: 32031111-32031111 on Build GRCh38
rs755250848 Chr.20: 32031077-32031077 on Build GRCh38
rs948765995 Chr.20: 32031420-32031420 on Build GRCh38
rs960046952 Chr.20: 32031377-32031377 on Build GRCh38
rs775682874 Chr.20: 32030964-32030964 on Build GRCh38
rs763480670 Chr.20: 32031053-32031053 on Build GRCh38
rs1051589350 Chr.20: 32031032-32031032 on Build GRCh38
rs1057166421 Chr.20: 32031202-32031202 on Build GRCh38
rs781349101 Chr.20: 32031312-32031312 on Build GRCh38
rs767586605 Chr.20: 32030979-32030979 on Build GRCh38
rs994484157 Chr.20: 32031416-32031416 on Build GRCh38
rs573320466 Chr.20: 32031072-32031072 on Build GRCh38
rs6061112 Chr.20: 32030984-32030984 on Build GRCh38

View Full Product Details