Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32358019-32358518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL1
Entrez Gene ID
171023
Gene Name
additional sex combs like 1, transcriptional regulator
Gene Aliases
BOPS, MDS
UniGene
Hs.374043
Gene Chromosome Location
Chr.20: 32358062-32439319 on Build GRCh38
Transcripts
NM_001164603.1 NM_015338.5 XM_006723727.3 XM_017027706.1

SNP Detail

SNP ID Chromosome Location
rs981019402 Chr.20: 32358173-32358173 on Build GRCh38
rs529676500 Chr.20: 32358411-32358411 on Build GRCh38
rs984787241 Chr.20: 32358106-32358106 on Build GRCh38
rs898858977 Chr.20: 32358398-32358398 on Build GRCh38
rs1007900979 Chr.20: 32358357-32358357 on Build GRCh38
rs973102868 Chr.20: 32358050-32358050 on Build GRCh38
rs140315242 Chr.20: 32358370-32358370 on Build GRCh38
rs1010051085 Chr.20: 32358093-32358093 on Build GRCh38
rs570644507 Chr.20: 32358077-32358077 on Build GRCh38
rs747051397 Chr.20: 32358233-32358233 on Build GRCh38
rs1054364865 Chr.20: 32358083-32358083 on Build GRCh38
rs563366395 Chr.20: 32358492-32358492 on Build GRCh38
rs1010538767 Chr.20: 32358222-32358222 on Build GRCh38
rs1053913436 Chr.20: 32358231-32358231 on Build GRCh38
rs769954548 Chr.20: 32358109-32358109 on Build GRCh38
rs1052037476 Chr.20: 32358304-32358304 on Build GRCh38
rs1038935859 Chr.20: 32358391-32358391 on Build GRCh38
rs546820789 Chr.20: 32358308-32358308 on Build GRCh38
rs757599168 Chr.20: 32358395-32358395 on Build GRCh38
rs142222436 Chr.20: 32358098-32358098 on Build GRCh38
rs528594862 Chr.20: 32358107-32358107 on Build GRCh38
rs1053861657 Chr.20: 32358100-32358100 on Build GRCh38
rs891735207 Chr.20: 32358091-32358091 on Build GRCh38
rs949810592 Chr.20: 32358061-32358061 on Build GRCh38
rs1044166328 Chr.20: 32358139-32358139 on Build GRCh38
rs371960756 Chr.20: 32358104-32358104 on Build GRCh38
rs948204520 Chr.20: 32358134-32358134 on Build GRCh38
rs905171319 Chr.20: 32358094-32358094 on Build GRCh38
rs886056584 Chr.20: 32358384-32358384 on Build GRCh38
rs935638756 Chr.20: 32358185-32358185 on Build GRCh38
rs973155522 Chr.20: 32358125-32358125 on Build GRCh38
rs949669168 Chr.20: 32358163-32358164 on Build GRCh38
rs540894675 Chr.20: 32358286-32358286 on Build GRCh38
rs994459675 Chr.20: 32358463-32358463 on Build GRCh38
rs1025578632 Chr.20: 32358493-32358493 on Build GRCh38
rs1040297348 Chr.20: 32358239-32358239 on Build GRCh38
rs949446801 Chr.20: 32358089-32358089 on Build GRCh38
rs952040896 Chr.20: 32358105-32358105 on Build GRCh38
rs780760915 Chr.20: 32358293-32358293 on Build GRCh38
rs945446906 Chr.20: 32358293-32358296 on Build GRCh38
rs548312050 Chr.20: 32358491-32358491 on Build GRCh38
rs575480431 Chr.20: 32358114-32358114 on Build GRCh38
rs149124543 Chr.20: 32358080-32358080 on Build GRCh38
rs922507322 Chr.20: 32358204-32358205 on Build GRCh38
rs918056235 Chr.20: 32358162-32358162 on Build GRCh38
rs114950455 Chr.20: 32358199-32358199 on Build GRCh38
rs556994910 Chr.20: 32358110-32358110 on Build GRCh38
rs758778723 Chr.20: 32358147-32358147 on Build GRCh38
rs996180790 Chr.20: 32358102-32358102 on Build GRCh38
rs962585352 Chr.20: 32358115-32358115 on Build GRCh38
rs890273009 Chr.20: 32358305-32358305 on Build GRCh38
rs777936226 Chr.20: 32358198-32358198 on Build GRCh38
rs918045089 Chr.20: 32358128-32358128 on Build GRCh38
rs965916878 Chr.20: 32358101-32358101 on Build GRCh38
rs535050178 Chr.20: 32358079-32358079 on Build GRCh38
rs564835956 Chr.20: 32358171-32358171 on Build GRCh38
rs1044177678 Chr.20: 32358068-32358068 on Build GRCh38
rs891476869 Chr.20: 32358219-32358219 on Build GRCh38
rs73241264 Chr.20: 32358122-32358122 on Build GRCh38
rs530763476 Chr.20: 32358494-32358494 on Build GRCh38

View Full Product Details