Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32358495-32359027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL1
Entrez Gene ID
171023
Gene Name
additional sex combs like 1, transcriptional regulator
Gene Aliases
BOPS, MDS
UniGene
Hs.374043
Gene Chromosome Location
Chr.20: 32358062-32439319 on Build GRCh38
Transcripts
NM_001164603.1 NM_015338.5 XM_006723727.3 XM_011528648.2 XM_017027706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6906087 c.25_27delAAG p.K9del Chr.20: 32358800-32358802 on Build GRCh38
6302973 c.57+6G>A p.? Chr.20: 32358838-32358838 on Build GRCh38
6970240 c.31C>T p.R11C Chr.20: 32358806-32358806 on Build GRCh38
4991355 c.24G>A p.K8K Chr.20: 32358799-32358799 on Build GRCh38
6939009 c.43G>C p.E15Q Chr.20: 32358818-32358818 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs911978295 Chr.20: 32358694-32358695 on Build GRCh38
rs913865340 Chr.20: 32358543-32358543 on Build GRCh38
rs886080863 Chr.20: 32358635-32358635 on Build GRCh38
rs1029022160 Chr.20: 32358988-32358988 on Build GRCh38
rs868083458 Chr.20: 32358707-32358707 on Build GRCh38
rs957205367 Chr.20: 32358589-32358589 on Build GRCh38
rs966722192 Chr.20: 32358684-32358684 on Build GRCh38
rs958279688 Chr.20: 32358680-32358680 on Build GRCh38
rs1024041213 Chr.20: 32358676-32358677 on Build GRCh38
rs976188740 Chr.20: 32358892-32358892 on Build GRCh38
rs943230941 Chr.20: 32358714-32358714 on Build GRCh38
rs1001244893 Chr.20: 32358874-32358874 on Build GRCh38
rs771767100 Chr.20: 32358772-32358772 on Build GRCh38
rs886056591 Chr.20: 32358682-32358682 on Build GRCh38
rs987982555 Chr.20: 32358685-32358686 on Build GRCh38
rs1021198194 Chr.20: 32358893-32358893 on Build GRCh38
rs930461650 Chr.20: 32358910-32358910 on Build GRCh38
rs907779561 Chr.20: 32358938-32358938 on Build GRCh38
rs967152855 Chr.20: 32358894-32358894 on Build GRCh38
rs996002104 Chr.20: 32358983-32358983 on Build GRCh38
rs1047387529 Chr.20: 32358617-32358617 on Build GRCh38
rs886056589 Chr.20: 32358598-32358598 on Build GRCh38
rs911741322 Chr.20: 32358568-32358569 on Build GRCh38
rs989489003 Chr.20: 32358523-32358531 on Build GRCh38
rs1038966097 Chr.20: 32358598-32358598 on Build GRCh38
rs776025782 Chr.20: 32358878-32358878 on Build GRCh38
rs775600332 Chr.20: 32358989-32358989 on Build GRCh38
rs949975924 Chr.20: 32358981-32358981 on Build GRCh38
rs763356478 Chr.20: 32358750-32358751 on Build GRCh38
rs983373711 Chr.20: 32358916-32358916 on Build GRCh38
rs540053795 Chr.20: 32359002-32359002 on Build GRCh38
rs552069090 Chr.20: 32358540-32358540 on Build GRCh38
rs886129274 Chr.20: 32358801-32358801 on Build GRCh38
rs568996228 Chr.20: 32358973-32358973 on Build GRCh38
rs901672968 Chr.20: 32358968-32358968 on Build GRCh38
rs1038849626 Chr.20: 32358718-32358718 on Build GRCh38
rs886056590 Chr.20: 32358617-32358617 on Build GRCh38
rs757765039 Chr.20: 32358849-32358850 on Build GRCh38
rs570751501 Chr.20: 32358679-32358679 on Build GRCh38
rs954034889 Chr.20: 32358659-32358686 on Build GRCh38
rs534633106 Chr.20: 32358683-32358683 on Build GRCh38
rs920439839 Chr.20: 32358905-32358905 on Build GRCh38
rs886056588 Chr.20: 32358555-32358555 on Build GRCh38
rs1011190827 Chr.20: 32358885-32358885 on Build GRCh38
rs376599709 Chr.20: 32358903-32358903 on Build GRCh38
rs1051827037 Chr.20: 32358560-32358560 on Build GRCh38
rs886056594 Chr.20: 32358688-32358688 on Build GRCh38
rs778480123 Chr.20: 32358736-32358736 on Build GRCh38
rs781330454 Chr.20: 32358873-32358874 on Build GRCh38
rs1020071543 Chr.20: 32358685-32358685 on Build GRCh38
rs557072958 Chr.20: 32358904-32358904 on Build GRCh38
rs958059333 Chr.20: 32358519-32358519 on Build GRCh38
rs746263656 Chr.20: 32358874-32358874 on Build GRCh38
rs578092988 Chr.20: 32358903-32358903 on Build GRCh38
rs920387361 Chr.20: 32358749-32358749 on Build GRCh38
rs764597521 Chr.20: 32358754-32358756 on Build GRCh38
rs989709157 Chr.20: 32358682-32358682 on Build GRCh38
rs200198574 Chr.20: 32358851-32358851 on Build GRCh38
rs994676294 Chr.20: 32358616-32358616 on Build GRCh38
rs770114307 Chr.20: 32358875-32358875 on Build GRCh38
rs971345309 Chr.20: 32358677-32358677 on Build GRCh38
rs886056586 Chr.20: 32358537-32358537 on Build GRCh38
rs1011318285 Chr.20: 32358603-32358603 on Build GRCh38
rs886056596 Chr.20: 32358800-32358802 on Build GRCh38
rs899077070 Chr.20: 32358600-32358600 on Build GRCh38
rs188788333 Chr.20: 32358884-32358884 on Build GRCh38
rs886056592 Chr.20: 32358684-32358684 on Build GRCh38
rs763506849 Chr.20: 32358880-32358880 on Build GRCh38
rs1045404951 Chr.20: 32358986-32358986 on Build GRCh38
rs747543538 Chr.20: 32358752-32358752 on Build GRCh38
rs752094508 Chr.20: 32358789-32358791 on Build GRCh38
rs949746726 Chr.20: 32358839-32358839 on Build GRCh38
rs945471862 Chr.20: 32358555-32358555 on Build GRCh38
rs1034161551 Chr.20: 32358583-32358583 on Build GRCh38
rs943197946 Chr.20: 32358577-32358577 on Build GRCh38
rs886056587 Chr.20: 32358553-32358553 on Build GRCh38
rs886056595 Chr.20: 32358744-32358744 on Build GRCh38
rs886056593 Chr.20: 32358699-32358700 on Build GRCh38

View Full Product Details