Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32779772-32780264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3B
Entrez Gene ID
1789
Gene Name
DNA methyltransferase 3 beta
Gene Aliases
ICF, ICF1, M.HsaIIIB
UniGene
Hs.643024
Gene Chromosome Location
Chr.20: 32762385-32809356 on Build GRCh38
Transcripts
NM_001207055.1 NM_001207056.1 NM_006892.3 NM_175848.1 NM_175849.1 NM_175850.2 XM_011528653.2 XM_011528654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4134438 c.27C>T p.S9S Chr.20: 32780142-32780142 on Build GRCh38
7269224 c.30+4_30+5delAG p.? Chr.20: 32780149-32780150 on Build GRCh38
3730726 c.30+5delG p.? Chr.20: 32780150-32780150 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370406742 Chr.20: 32780190-32780190 on Build GRCh38
rs535911872 Chr.20: 32779978-32779978 on Build GRCh38
rs940209667 Chr.20: 32779813-32779813 on Build GRCh38
rs1013608796 Chr.20: 32779930-32779930 on Build GRCh38
rs6058885 Chr.20: 32780142-32780142 on Build GRCh38
rs781414104 Chr.20: 32780067-32780067 on Build GRCh38
rs759146619 Chr.20: 32780118-32780118 on Build GRCh38
rs985895540 Chr.20: 32780044-32780044 on Build GRCh38
rs1009479964 Chr.20: 32780165-32780166 on Build GRCh38
rs941561778 Chr.20: 32780054-32780054 on Build GRCh38
rs909998929 Chr.20: 32779807-32779807 on Build GRCh38
rs779107629 Chr.20: 32780167-32780167 on Build GRCh38
rs770099038 Chr.20: 32780086-32780086 on Build GRCh38
rs898768830 Chr.20: 32780061-32780061 on Build GRCh38
rs941712747 Chr.20: 32780051-32780051 on Build GRCh38
rs973198192 Chr.20: 32779816-32779816 on Build GRCh38
rs772085974 Chr.20: 32780175-32780175 on Build GRCh38
rs754269795 Chr.20: 32780045-32780045 on Build GRCh38
rs746076116 Chr.20: 32780085-32780085 on Build GRCh38
rs367677856 Chr.20: 32780178-32780178 on Build GRCh38
rs1032274451 Chr.20: 32779994-32779994 on Build GRCh38
rs999480396 Chr.20: 32780201-32780201 on Build GRCh38
rs557508006 Chr.20: 32780036-32780036 on Build GRCh38
rs985259645 Chr.20: 32780042-32780042 on Build GRCh38
rs189183505 Chr.20: 32780075-32780075 on Build GRCh38
rs1024087524 Chr.20: 32779935-32779935 on Build GRCh38
rs994307700 Chr.20: 32779821-32779821 on Build GRCh38
rs776425195 Chr.20: 32780195-32780195 on Build GRCh38
rs534958197 Chr.20: 32779929-32779929 on Build GRCh38
rs1022563720 Chr.20: 32780147-32780147 on Build GRCh38
rs896597060 Chr.20: 32779877-32779877 on Build GRCh38
rs1038960790 Chr.20: 32780072-32780072 on Build GRCh38
rs950011853 Chr.20: 32779826-32779826 on Build GRCh38
rs760211805 Chr.20: 32780114-32780114 on Build GRCh38
rs570640182 Chr.20: 32779899-32779899 on Build GRCh38
rs752017376 Chr.20: 32780123-32780123 on Build GRCh38
rs955104117 Chr.20: 32780017-32780017 on Build GRCh38
rs1010717734 Chr.20: 32780144-32780144 on Build GRCh38
rs376242248 Chr.20: 32780159-32780159 on Build GRCh38
rs878879044 Chr.20: 32779841-32779841 on Build GRCh38
rs540989285 Chr.20: 32779800-32779800 on Build GRCh38
rs372737106 Chr.20: 32780088-32780088 on Build GRCh38
rs760239224 Chr.20: 32780184-32780184 on Build GRCh38
rs368735412 Chr.20: 32780172-32780172 on Build GRCh38
rs766182686 Chr.20: 32780116-32780116 on Build GRCh38
rs977726249 Chr.20: 32780015-32780015 on Build GRCh38
rs748030022 Chr.20: 32780171-32780171 on Build GRCh38
rs879121054 Chr.20: 32780219-32780219 on Build GRCh38
rs755162607 Chr.20: 32780113-32780114 on Build GRCh38
rs185118162 Chr.20: 32779973-32779973 on Build GRCh38
rs753346954 Chr.20: 32780117-32780117 on Build GRCh38
rs959269541 Chr.20: 32779950-32779950 on Build GRCh38
rs761612686 Chr.20: 32780099-32780099 on Build GRCh38
rs907253494 Chr.20: 32780095-32780095 on Build GRCh38
rs539431309 Chr.20: 32780113-32780113 on Build GRCh38
rs552176507 Chr.20: 32779818-32779818 on Build GRCh38
rs969421463 Chr.20: 32779948-32779948 on Build GRCh38
rs368554438 Chr.20: 32780087-32780087 on Build GRCh38
rs1044445262 Chr.20: 32779990-32779990 on Build GRCh38
rs1025742634 Chr.20: 32779870-32779870 on Build GRCh38
rs771750028 Chr.20: 32780101-32780101 on Build GRCh38
rs954530244 Chr.20: 32780235-32780235 on Build GRCh38
rs781384415 Chr.20: 32780138-32780138 on Build GRCh38
rs764624409 Chr.20: 32780122-32780122 on Build GRCh38
rs898707803 Chr.20: 32779808-32779808 on Build GRCh38
rs930209406 Chr.20: 32780063-32780063 on Build GRCh38
rs756282732 Chr.20: 32780153-32780153 on Build GRCh38
rs573451454 Chr.20: 32780162-32780162 on Build GRCh38
rs540921199 Chr.20: 32780229-32780229 on Build GRCh38
rs933090778 Chr.20: 32780035-32780035 on Build GRCh38
rs757853408 Chr.20: 32780132-32780132 on Build GRCh38
rs780286949 Chr.20: 32780157-32780157 on Build GRCh38
rs966576839 Chr.20: 32780191-32780191 on Build GRCh38
rs749523817 Chr.20: 32780080-32780080 on Build GRCh38
rs922264632 Chr.20: 32780016-32780016 on Build GRCh38
rs774277730 Chr.20: 32780091-32780091 on Build GRCh38
rs546475094 Chr.20: 32779958-32779958 on Build GRCh38
rs1038618769 Chr.20: 32779806-32779814 on Build GRCh38
rs1055915301 Chr.20: 32780102-32780102 on Build GRCh38
rs917355990 Chr.20: 32779822-32779822 on Build GRCh38
rs747356795 Chr.20: 32780066-32780066 on Build GRCh38
rs923087043 Chr.20: 32780027-32780027 on Build GRCh38

View Full Product Details