Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32780231-32780722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3B
Entrez Gene ID
1789
Gene Name
DNA methyltransferase 3 beta
Gene Aliases
ICF, ICF1, M.HsaIIIB
UniGene
Hs.643024
Gene Chromosome Location
Chr.20: 32762385-32809356 on Build GRCh38
Transcripts
NM_001207055.1 NM_001207056.1 NM_006892.3 NM_175848.1 NM_175849.1 NM_175850.2 XM_011528653.2 XM_011528654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6921202 c.88C>T p.Q30* Chr.20: 32780411-32780411 on Build GRCh38
6921203 c.124C>T p.Q42* Chr.20: 32780411-32780411 on Build GRCh38
3405020 c.165C>T p.T55T Chr.20: 32780452-32780452 on Build GRCh38
2889470 c.37G>T p.D13Y Chr.20: 32780360-32780360 on Build GRCh38
6647841 c.84C>T p.S28S Chr.20: 32780407-32780407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530131895 Chr.20: 32780519-32780519 on Build GRCh38
rs963434160 Chr.20: 32780252-32780252 on Build GRCh38
rs770176873 Chr.20: 32780496-32780496 on Build GRCh38
rs576300808 Chr.20: 32780318-32780318 on Build GRCh38
rs754241012 Chr.20: 32780387-32780387 on Build GRCh38
rs139418486 Chr.20: 32780677-32780677 on Build GRCh38
rs766920119 Chr.20: 32780424-32780424 on Build GRCh38
rs759924115 Chr.20: 32780433-32780433 on Build GRCh38
rs555607862 Chr.20: 32780606-32780606 on Build GRCh38
rs529125414 Chr.20: 32780500-32780500 on Build GRCh38
rs534189689 Chr.20: 32780594-32780594 on Build GRCh38
rs370943056 Chr.20: 32780501-32780501 on Build GRCh38
rs121908945 Chr.20: 32780411-32780411 on Build GRCh38
rs752972556 Chr.20: 32780440-32780440 on Build GRCh38
rs937385003 Chr.20: 32780372-32780372 on Build GRCh38
rs765224048 Chr.20: 32780673-32780673 on Build GRCh38
rs955051695 Chr.20: 32780251-32780251 on Build GRCh38
rs376079908 Chr.20: 32780515-32780515 on Build GRCh38
rs189508107 Chr.20: 32780616-32780616 on Build GRCh38
rs750715627 Chr.20: 32780289-32780289 on Build GRCh38
rs748967232 Chr.20: 32780469-32780469 on Build GRCh38
rs774816105 Chr.20: 32780380-32780380 on Build GRCh38
rs370942739 Chr.20: 32780279-32780279 on Build GRCh38
rs867493361 Chr.20: 32780652-32780652 on Build GRCh38
rs768234407 Chr.20: 32780472-32780472 on Build GRCh38
rs781115406 Chr.20: 32780399-32780399 on Build GRCh38
rs374862682 Chr.20: 32780273-32780273 on Build GRCh38
rs373594568 Chr.20: 32780368-32780368 on Build GRCh38
rs141756182 Chr.20: 32780382-32780382 on Build GRCh38
rs1006528662 Chr.20: 32780488-32780488 on Build GRCh38
rs755258723 Chr.20: 32780320-32780320 on Build GRCh38
rs375474052 Chr.20: 32780687-32780687 on Build GRCh38
rs775216151 Chr.20: 32780408-32780408 on Build GRCh38
rs374913902 Chr.20: 32780438-32780438 on Build GRCh38
rs144581857 Chr.20: 32780659-32780659 on Build GRCh38
rs369669631 Chr.20: 32780489-32780489 on Build GRCh38
rs4571133 Chr.20: 32780511-32780511 on Build GRCh38
rs973115622 Chr.20: 32780259-32780259 on Build GRCh38
rs730823 Chr.20: 32780365-32780365 on Build GRCh38
rs759054819 Chr.20: 32780268-32780268 on Build GRCh38
rs1041851718 Chr.20: 32780603-32780603 on Build GRCh38
rs758218721 Chr.20: 32780340-32780340 on Build GRCh38
rs201455430 Chr.20: 32780337-32780337 on Build GRCh38
rs761058273 Chr.20: 32780516-32780516 on Build GRCh38
rs751741367 Chr.20: 32780448-32780448 on Build GRCh38
rs377301092 Chr.20: 32780452-32780452 on Build GRCh38
rs757326781 Chr.20: 32780398-32780398 on Build GRCh38
rs991473250 Chr.20: 32780563-32780563 on Build GRCh38
rs758536558 Chr.20: 32780369-32780369 on Build GRCh38
rs779770844 Chr.20: 32780468-32780468 on Build GRCh38
rs1569687 Chr.20: 32780582-32780582 on Build GRCh38
rs773892206 Chr.20: 32780291-32780291 on Build GRCh38
rs935128885 Chr.20: 32780622-32780622 on Build GRCh38
rs564154904 Chr.20: 32780425-32780425 on Build GRCh38
rs768199189 Chr.20: 32780419-32780419 on Build GRCh38
rs115990549 Chr.20: 32780275-32780275 on Build GRCh38
rs747677882 Chr.20: 32780481-32780481 on Build GRCh38
rs1053875662 Chr.20: 32780634-32780634 on Build GRCh38
rs758603280 Chr.20: 32780445-32780445 on Build GRCh38
rs761138255 Chr.20: 32780423-32780423 on Build GRCh38
rs777511594 Chr.20: 32780395-32780395 on Build GRCh38
rs992426406 Chr.20: 32780564-32780564 on Build GRCh38
rs530552033 Chr.20: 32780383-32780383 on Build GRCh38
rs150200553 Chr.20: 32780447-32780447 on Build GRCh38
rs147580720 Chr.20: 32780362-32780362 on Build GRCh38
rs995204658 Chr.20: 32780683-32780683 on Build GRCh38
rs778370717 Chr.20: 32780475-32780475 on Build GRCh38
rs757214677 Chr.20: 32780451-32780451 on Build GRCh38
rs143743194 Chr.20: 32780389-32780389 on Build GRCh38
rs375171362 Chr.20: 32780366-32780366 on Build GRCh38
rs770859652 Chr.20: 32780363-32780363 on Build GRCh38
rs184776217 Chr.20: 32780577-32780577 on Build GRCh38
rs752677666 Chr.20: 32780393-32780393 on Build GRCh38
rs754046991 Chr.20: 32780304-32780304 on Build GRCh38
rs981644166 Chr.20: 32780528-32780528 on Build GRCh38
rs180865290 Chr.20: 32780632-32780632 on Build GRCh38
rs539966301 Chr.20: 32780545-32780545 on Build GRCh38
rs773918660 Chr.20: 32780420-32780420 on Build GRCh38
rs949594205 Chr.20: 32780311-32780311 on Build GRCh38
rs769293365 Chr.20: 32780405-32780405 on Build GRCh38
rs756476916 Chr.20: 32780294-32780294 on Build GRCh38
rs200773466 Chr.20: 32780307-32780307 on Build GRCh38
rs568228701 Chr.20: 32780492-32780492 on Build GRCh38
rs778827030 Chr.20: 32780333-32780333 on Build GRCh38
rs780191958 Chr.20: 32780296-32780296 on Build GRCh38
rs1047298429 Chr.20: 32780278-32780278 on Build GRCh38
rs759757544 Chr.20: 32780495-32780495 on Build GRCh38
rs367766007 Chr.20: 32780390-32780390 on Build GRCh38
rs138805251 Chr.20: 32780455-32780455 on Build GRCh38
rs781313570 Chr.20: 32780454-32780454 on Build GRCh38
rs755951894 Chr.20: 32780457-32780457 on Build GRCh38
rs151128145 Chr.20: 32780396-32780396 on Build GRCh38
rs546883331 Chr.20: 32780484-32780484 on Build GRCh38
rs181671547 Chr.20: 32780544-32780544 on Build GRCh38
rs201790923 Chr.20: 32780404-32780404 on Build GRCh38
rs368407604 Chr.20: 32780360-32780360 on Build GRCh38
rs758506171 Chr.20: 32780394-32780394 on Build GRCh38
rs974233540 Chr.20: 32780339-32780339 on Build GRCh38

View Full Product Details