Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32781195-32781594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3B
Entrez Gene ID
1789
Gene Name
DNA methyltransferase 3 beta
Gene Aliases
ICF, ICF1, M.HsaIIIB
UniGene
Hs.643024
Gene Chromosome Location
Chr.20: 32762385-32809356 on Build GRCh38
Transcripts
NM_001207055.1 NM_001207056.1 NM_006892.3 NM_175848.1 NM_175849.1 NM_175850.2 XM_011528653.2 XM_011528654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025776 c.187T>C p.S63P Chr.20: 32781361-32781361 on Build GRCh38
6919872 c.182G>A p.R61Q Chr.20: 32781356-32781356 on Build GRCh38
6647872 c.197G>A p.R66Q Chr.20: 32781371-32781371 on Build GRCh38
6843499 c.145C>T p.R49* Chr.20: 32781355-32781355 on Build GRCh38
1025778 c.240+1G>A p.? Chr.20: 32781415-32781415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373973931 Chr.20: 32781438-32781438 on Build GRCh38
rs938323831 Chr.20: 32781552-32781552 on Build GRCh38
rs752329190 Chr.20: 32781392-32781392 on Build GRCh38
rs200738364 Chr.20: 32781331-32781331 on Build GRCh38
rs768937892 Chr.20: 32781350-32781350 on Build GRCh38
rs780432886 Chr.20: 32781443-32781443 on Build GRCh38
rs1057305735 Chr.20: 32781239-32781239 on Build GRCh38
rs949900284 Chr.20: 32781413-32781413 on Build GRCh38
rs760640013 Chr.20: 32781370-32781370 on Build GRCh38
rs34005307 Chr.20: 32781227-32781228 on Build GRCh38
rs1021222720 Chr.20: 32781560-32781560 on Build GRCh38
rs769132393 Chr.20: 32781457-32781457 on Build GRCh38
rs182909590 Chr.20: 32781514-32781514 on Build GRCh38
rs200957571 Chr.20: 32781451-32781451 on Build GRCh38
rs1013470342 Chr.20: 32781273-32781273 on Build GRCh38
rs1056810716 Chr.20: 32781555-32781555 on Build GRCh38
rs377138079 Chr.20: 32781344-32781344 on Build GRCh38
rs756075182 Chr.20: 32781306-32781306 on Build GRCh38
rs919773729 Chr.20: 32781442-32781442 on Build GRCh38
rs548919805 Chr.20: 32781474-32781474 on Build GRCh38
rs1002563562 Chr.20: 32781225-32781225 on Build GRCh38
rs777319209 Chr.20: 32781322-32781322 on Build GRCh38
rs964839947 Chr.20: 32781407-32781407 on Build GRCh38
rs560602182 Chr.20: 32781427-32781427 on Build GRCh38
rs771169367 Chr.20: 32781281-32781281 on Build GRCh38
rs200349528 Chr.20: 32781345-32781345 on Build GRCh38
rs17123590 Chr.20: 32781371-32781371 on Build GRCh38
rs551310722 Chr.20: 32781400-32781400 on Build GRCh38
rs767507418 Chr.20: 32781303-32781303 on Build GRCh38
rs190353959 Chr.20: 32781320-32781320 on Build GRCh38
rs754734640 Chr.20: 32781375-32781375 on Build GRCh38
rs756606814 Chr.20: 32781439-32781439 on Build GRCh38
rs544561079 Chr.20: 32781368-32781368 on Build GRCh38
rs145694804 Chr.20: 32781369-32781369 on Build GRCh38
rs746379478 Chr.20: 32781332-32781332 on Build GRCh38
rs567369870 Chr.20: 32781487-32781487 on Build GRCh38
rs908198175 Chr.20: 32781488-32781488 on Build GRCh38
rs753624270 Chr.20: 32781309-32781309 on Build GRCh38
rs750373045 Chr.20: 32781305-32781305 on Build GRCh38
rs376894979 Chr.20: 32781471-32781471 on Build GRCh38
rs533183175 Chr.20: 32781393-32781393 on Build GRCh38
rs779868570 Chr.20: 32781307-32781307 on Build GRCh38
rs1009865663 Chr.20: 32781535-32781535 on Build GRCh38
rs778307198 Chr.20: 32781330-32781331 on Build GRCh38
rs543579055 Chr.20: 32781492-32781492 on Build GRCh38
rs1032660063 Chr.20: 32781243-32781243 on Build GRCh38
rs968920052 Chr.20: 32781301-32781301 on Build GRCh38
rs746722947 Chr.20: 32781264-32781264 on Build GRCh38
rs3835238 Chr.20: 32781331-32781331 on Build GRCh38
rs766736344 Chr.20: 32781412-32781412 on Build GRCh38
rs764811687 Chr.20: 32781377-32781377 on Build GRCh38
rs1011153215 Chr.20: 32781265-32781265 on Build GRCh38
rs201695828 Chr.20: 32781314-32781314 on Build GRCh38
rs777231550 Chr.20: 32781414-32781414 on Build GRCh38
rs966867954 Chr.20: 32781295-32781295 on Build GRCh38
rs1021801664 Chr.20: 32781282-32781282 on Build GRCh38
rs762185930 Chr.20: 32781353-32781353 on Build GRCh38
rs1001626697 Chr.20: 32781236-32781236 on Build GRCh38
rs6057643 Chr.20: 32781483-32781483 on Build GRCh38

View Full Product Details