Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32788763-32789274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3B
Entrez Gene ID
1789
Gene Name
DNA methyltransferase 3 beta
Gene Aliases
ICF, ICF1, M.HsaIIIB
UniGene
Hs.643024
Gene Chromosome Location
Chr.20: 32762385-32809356 on Build GRCh38
Transcripts
NM_001207055.1 NM_001207056.1 NM_006892.3 NM_175848.1 NM_175849.1 NM_175850.2 XM_011528653.2 XM_011528654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
418989 c.845C>G p.S282C Chr.20: 32789008-32789008 on Build GRCh38
95831 c.813G>A p.E271E Chr.20: 32789012-32789012 on Build GRCh38
4506465 c.684C>T p.L228L Chr.20: 32788883-32788883 on Build GRCh38
6038451 c.846C>T p.S282S Chr.20: 32789009-32789009 on Build GRCh38
1025791 c.685G>A p.V229M Chr.20: 32788884-32788884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935624265 Chr.20: 32788797-32788797 on Build GRCh38
rs565659570 Chr.20: 32788982-32788982 on Build GRCh38
rs376006476 Chr.20: 32788854-32788854 on Build GRCh38
rs368419589 Chr.20: 32788821-32788821 on Build GRCh38
rs945731547 Chr.20: 32789206-32789206 on Build GRCh38
rs1011386107 Chr.20: 32788801-32788801 on Build GRCh38
rs764811612 Chr.20: 32788922-32788922 on Build GRCh38
rs760505841 Chr.20: 32788878-32788878 on Build GRCh38
rs181311493 Chr.20: 32789193-32789193 on Build GRCh38
rs763490983 Chr.20: 32788925-32788925 on Build GRCh38
rs779489353 Chr.20: 32788957-32788957 on Build GRCh38
rs763840638 Chr.20: 32789026-32789026 on Build GRCh38
rs121908947 Chr.20: 32789007-32789007 on Build GRCh38
rs776562712 Chr.20: 32788987-32788987 on Build GRCh38
rs746146250 Chr.20: 32788824-32788826 on Build GRCh38
rs762732247 Chr.20: 32788924-32788924 on Build GRCh38
rs754201724 Chr.20: 32788954-32788954 on Build GRCh38
rs557352923 Chr.20: 32788991-32788991 on Build GRCh38
rs146869906 Chr.20: 32788928-32788928 on Build GRCh38
rs996321864 Chr.20: 32788962-32788962 on Build GRCh38
rs918019174 Chr.20: 32789171-32789171 on Build GRCh38
rs558666547 Chr.20: 32789186-32789186 on Build GRCh38
rs6119961 Chr.20: 32789118-32789118 on Build GRCh38
rs1014966921 Chr.20: 32789022-32789022 on Build GRCh38
rs761422208 Chr.20: 32789033-32789033 on Build GRCh38
rs1021818100 Chr.20: 32788814-32788814 on Build GRCh38
rs770784575 Chr.20: 32788984-32788984 on Build GRCh38
rs901993836 Chr.20: 32788941-32788941 on Build GRCh38
rs140518698 Chr.20: 32789009-32789009 on Build GRCh38
rs759527333 Chr.20: 32788994-32788994 on Build GRCh38
rs370526016 Chr.20: 32789020-32789020 on Build GRCh38
rs953639938 Chr.20: 32789227-32789227 on Build GRCh38
rs750967946 Chr.20: 32789028-32789028 on Build GRCh38
rs576409141 Chr.20: 32789130-32789130 on Build GRCh38
rs762465821 Chr.20: 32789021-32789021 on Build GRCh38
rs537597783 Chr.20: 32789172-32789172 on Build GRCh38
rs768933791 Chr.20: 32788806-32788806 on Build GRCh38
rs901604398 Chr.20: 32789233-32789233 on Build GRCh38
rs201520258 Chr.20: 32788931-32788931 on Build GRCh38
rs992273117 Chr.20: 32789156-32789156 on Build GRCh38
rs762250690 Chr.20: 32789205-32789205 on Build GRCh38
rs1029223008 Chr.20: 32789191-32789191 on Build GRCh38
rs964977606 Chr.20: 32788909-32788909 on Build GRCh38
rs780710615 Chr.20: 32788938-32788938 on Build GRCh38
rs1054134807 Chr.20: 32788836-32788836 on Build GRCh38
rs779295951 Chr.20: 32788837-32788837 on Build GRCh38
rs188514528 Chr.20: 32788976-32788976 on Build GRCh38
rs185784169 Chr.20: 32789223-32789223 on Build GRCh38
rs1054511815 Chr.20: 32789204-32789204 on Build GRCh38
rs373811098 Chr.20: 32788883-32788883 on Build GRCh38
rs767198587 Chr.20: 32789037-32789037 on Build GRCh38
rs867559860 Chr.20: 32788920-32788920 on Build GRCh38
rs6119285 Chr.20: 32789189-32789189 on Build GRCh38
rs375192760 Chr.20: 32788818-32788818 on Build GRCh38
rs776366575 Chr.20: 32788895-32788895 on Build GRCh38
rs774981841 Chr.20: 32788812-32788812 on Build GRCh38
rs370105694 Chr.20: 32788838-32788838 on Build GRCh38
rs891529660 Chr.20: 32788880-32788880 on Build GRCh38
rs949838499 Chr.20: 32789183-32789183 on Build GRCh38
rs200921151 Chr.20: 32788919-32788919 on Build GRCh38
rs748289618 Chr.20: 32788967-32788967 on Build GRCh38
rs1042889047 Chr.20: 32789080-32789080 on Build GRCh38
rs754396044 Chr.20: 32789050-32789050 on Build GRCh38
rs772383693 Chr.20: 32788975-32788975 on Build GRCh38
rs199883886 Chr.20: 32788923-32788923 on Build GRCh38
rs575221231 Chr.20: 32789225-32789225 on Build GRCh38
rs773384575 Chr.20: 32788858-32788858 on Build GRCh38
rs754750423 Chr.20: 32788961-32788962 on Build GRCh38
rs927167478 Chr.20: 32789190-32789190 on Build GRCh38

View Full Product Details